Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 58168525-58168924 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSNK2A2
Entrez Gene ID
1459
Gene Name
casein kinase 2 alpha 2
Gene Aliases
CK2A2, CK2alpha', CSNK2A1
UniGene
Hs.82201
Gene Chromosome Location
Chr.16: 58157907-58198116 on Build GRCh38
Transcripts
NM_001896.2 XM_005255800.3 XM_005255801.3 XM_017022945.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6839079 c.496G>C p.D166H Chr.16: 58168627-58168627 on Build GRCh38
6295830 c.505C>A p.Q169K Chr.16: 58168618-58168618 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs945852848 Chr.16: 58168884-58168884 on Build GRCh38
rs988118317 Chr.16: 58168899-58168899 on Build GRCh38
rs553892235 Chr.16: 58168865-58168865 on Build GRCh38
rs1003606797 Chr.16: 58168870-58168870 on Build GRCh38
rs756466707 Chr.16: 58168711-58168711 on Build GRCh38
rs762261740 Chr.16: 58168730-58168730 on Build GRCh38
rs899044863 Chr.16: 58168830-58168830 on Build GRCh38
rs780597425 Chr.16: 58168723-58168723 on Build GRCh38
rs539597583 Chr.16: 58168749-58168749 on Build GRCh38
rs570639813 Chr.16: 58168569-58168569 on Build GRCh38
rs536757753 Chr.16: 58168818-58168818 on Build GRCh38
rs572532862 Chr.16: 58168806-58168806 on Build GRCh38
rs372932882 Chr.16: 58168582-58168582 on Build GRCh38
rs746782575 Chr.16: 58168596-58168596 on Build GRCh38
rs759058120 Chr.16: 58168737-58168737 on Build GRCh38
rs921690733 Chr.16: 58168748-58168748 on Build GRCh38
rs144651178 Chr.16: 58168754-58168754 on Build GRCh38
rs534783008 Chr.16: 58168701-58168701 on Build GRCh38
rs774916682 Chr.16: 58168728-58168728 on Build GRCh38
rs879123154 Chr.16: 58168672-58168672 on Build GRCh38
rs771211913 Chr.16: 58168664-58168664 on Build GRCh38
rs753551189 Chr.16: 58168735-58168735 on Build GRCh38
rs937117278 Chr.16: 58168861-58168864 on Build GRCh38
rs1035058555 Chr.16: 58168873-58168873 on Build GRCh38
rs769939605 Chr.16: 58168700-58168700 on Build GRCh38
rs926916677 Chr.16: 58168842-58168843 on Build GRCh38
rs953492302 Chr.16: 58168727-58168727 on Build GRCh38
rs956961750 Chr.16: 58168879-58168879 on Build GRCh38
rs781398318 Chr.16: 58168666-58168666 on Build GRCh38
rs369576559 Chr.16: 58168719-58168719 on Build GRCh38
rs375805834 Chr.16: 58168709-58168709 on Build GRCh38
rs779176682 Chr.16: 58168741-58168741 on Build GRCh38
rs890152347 Chr.16: 58168801-58168801 on Build GRCh38
rs1004547291 Chr.16: 58168802-58168802 on Build GRCh38
rs985246920 Chr.16: 58168740-58168740 on Build GRCh38
rs746032185 Chr.16: 58168687-58168687 on Build GRCh38
rs1023397005 Chr.16: 58168839-58168839 on Build GRCh38
rs569216926 Chr.16: 58168747-58168747 on Build GRCh38
rs995087644 Chr.16: 58168835-58168835 on Build GRCh38

View Full Product Details