Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 61655378-61655877 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDH8
Entrez Gene ID
1006
Gene Name
cadherin 8
Gene Aliases
Nbla04261
UniGene
Hs.368322
Gene Chromosome Location
Chr.16: 61640435-62036835 on Build GRCh38
Transcripts
NM_001796.4 XM_005255760.3 XM_017022829.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
972086 c.1831G>A p.E611K Chr.16: 61655545-61655545 on Build GRCh38
6349665 c.1857C>G p.L619L Chr.16: 61655519-61655519 on Build GRCh38
167883 c.1813G>A p.V605I Chr.16: 61655563-61655563 on Build GRCh38
471892 c.1806C>G p.D602E Chr.16: 61655570-61655570 on Build GRCh38
218499 c.1850T>A p.I617N Chr.16: 61655526-61655526 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs140833890 Chr.16: 61655574-61655574 on Build GRCh38
rs1000565029 Chr.16: 61655465-61655465 on Build GRCh38
rs952700467 Chr.16: 61655647-61655647 on Build GRCh38
rs200664753 Chr.16: 61655767-61655767 on Build GRCh38
rs749548045 Chr.16: 61655641-61655641 on Build GRCh38
rs745899192 Chr.16: 61655619-61655619 on Build GRCh38
rs780041984 Chr.16: 61655418-61655418 on Build GRCh38
rs751958648 Chr.16: 61655581-61655581 on Build GRCh38
rs756217884 Chr.16: 61655762-61655762 on Build GRCh38
rs552393725 Chr.16: 61655453-61655453 on Build GRCh38
rs780387534 Chr.16: 61655633-61655633 on Build GRCh38
rs892187391 Chr.16: 61655511-61655511 on Build GRCh38
rs755649849 Chr.16: 61655450-61655450 on Build GRCh38
rs377569521 Chr.16: 61655760-61655760 on Build GRCh38
rs867920571 Chr.16: 61655681-61655681 on Build GRCh38
rs778497935 Chr.16: 61655463-61655463 on Build GRCh38
rs112856325 Chr.16: 61655547-61655547 on Build GRCh38
rs569963221 Chr.16: 61655446-61655446 on Build GRCh38
rs774280986 Chr.16: 61655436-61655436 on Build GRCh38
rs181502944 Chr.16: 61655444-61655444 on Build GRCh38
rs764747942 Chr.16: 61655690-61655690 on Build GRCh38
rs781297807 Chr.16: 61655497-61655497 on Build GRCh38
rs202247121 Chr.16: 61655573-61655573 on Build GRCh38
rs111810282 Chr.16: 61655829-61655829 on Build GRCh38
rs145143780 Chr.16: 61655661-61655661 on Build GRCh38
rs149052342 Chr.16: 61655775-61655775 on Build GRCh38
rs969167994 Chr.16: 61655816-61655816 on Build GRCh38
rs760455016 Chr.16: 61655663-61655663 on Build GRCh38
rs377367876 Chr.16: 61655572-61655572 on Build GRCh38
rs543586963 Chr.16: 61655536-61655536 on Build GRCh38
rs908620486 Chr.16: 61655710-61655710 on Build GRCh38
rs185749706 Chr.16: 61655550-61655550 on Build GRCh38
rs761638294 Chr.16: 61655530-61655530 on Build GRCh38
rs766269925 Chr.16: 61655546-61655546 on Build GRCh38
rs771390154 Chr.16: 61655486-61655486 on Build GRCh38
rs903809316 Chr.16: 61655840-61655840 on Build GRCh38
rs938765647 Chr.16: 61655724-61655725 on Build GRCh38
rs866984859 Chr.16: 61655680-61655680 on Build GRCh38
rs754629645 Chr.16: 61655461-61655461 on Build GRCh38
rs201367392 Chr.16: 61655549-61655549 on Build GRCh38
rs778088256 Chr.16: 61655576-61655576 on Build GRCh38
rs781605014 Chr.16: 61655592-61655592 on Build GRCh38
rs141172673 Chr.16: 61655449-61655449 on Build GRCh38
rs762559369 Chr.16: 61655696-61655696 on Build GRCh38
rs769050652 Chr.16: 61655650-61655650 on Build GRCh38
rs772027090 Chr.16: 61655534-61655534 on Build GRCh38
rs757572651 Chr.16: 61655746-61655746 on Build GRCh38
rs759020906 Chr.16: 61655684-61655684 on Build GRCh38
rs147771827 Chr.16: 61655432-61655432 on Build GRCh38
rs34124175 Chr.16: 61655693-61655693 on Build GRCh38
rs144069542 Chr.16: 61655474-61655474 on Build GRCh38
rs769764183 Chr.16: 61655629-61655629 on Build GRCh38
rs369865177 Chr.16: 61655477-61655477 on Build GRCh38
rs746983273 Chr.16: 61655527-61655527 on Build GRCh38
rs868495017 Chr.16: 61655626-61655626 on Build GRCh38
rs770039944 Chr.16: 61655519-61655519 on Build GRCh38
rs777937491 Chr.16: 61655584-61655584 on Build GRCh38
rs760363205 Chr.16: 61655767-61655768 on Build GRCh38
rs371269358 Chr.16: 61655752-61655752 on Build GRCh38
rs560315079 Chr.16: 61655841-61655841 on Build GRCh38
rs927635086 Chr.16: 61655445-61655445 on Build GRCh38
rs763801431 Chr.16: 61655709-61655709 on Build GRCh38
rs374720488 Chr.16: 61655470-61655470 on Build GRCh38
rs753339953 Chr.16: 61655459-61655459 on Build GRCh38
rs759779542 Chr.16: 61655419-61655419 on Build GRCh38
rs373281473 Chr.16: 61655537-61655537 on Build GRCh38
rs776447082 Chr.16: 61655672-61655672 on Build GRCh38
rs745998335 Chr.16: 61655500-61655500 on Build GRCh38
rs748499678 Chr.16: 61655466-61655466 on Build GRCh38
rs1004181969 Chr.16: 61655651-61655651 on Build GRCh38
rs751936754 Chr.16: 61655745-61655745 on Build GRCh38
rs553486677 Chr.16: 61655848-61655848 on Build GRCh38
rs377335189 Chr.16: 61655667-61655667 on Build GRCh38
rs150293920 Chr.16: 61655526-61655526 on Build GRCh38
rs920201133 Chr.16: 61655621-61655621 on Build GRCh38
rs146415895 Chr.16: 61655563-61655563 on Build GRCh38
rs374533261 Chr.16: 61655544-61655544 on Build GRCh38

View Full Product Details