Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 61726969-61727491 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDH8
Entrez Gene ID
1006
Gene Name
cadherin 8
Gene Aliases
Nbla04261
UniGene
Hs.368322
Gene Chromosome Location
Chr.16: 61640435-62036835 on Build GRCh38
Transcripts
NM_001796.4 XM_005255760.3 XM_017022829.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1661078 c.1473C>A p.D491E Chr.16: 61727157-61727157 on Build GRCh38
3510703 c.1494C>T p.S498S Chr.16: 61727136-61727136 on Build GRCh38
2993192 c.1440A>G p.V480V Chr.16: 61727190-61727190 on Build GRCh38
6002213 c.1536+6T>C p.? Chr.16: 61727088-61727088 on Build GRCh38
193192 c.1488C>T p.F496F Chr.16: 61727142-61727142 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs542600689 Chr.16: 61727390-61727390 on Build GRCh38
rs752861917 Chr.16: 61727015-61727015 on Build GRCh38
rs764252618 Chr.16: 61727189-61727189 on Build GRCh38
rs777230396 Chr.16: 61727065-61727065 on Build GRCh38
rs919497646 Chr.16: 61727290-61727290 on Build GRCh38
rs746683292 Chr.16: 61727141-61727141 on Build GRCh38
rs778260497 Chr.16: 61727043-61727043 on Build GRCh38
rs200934735 Chr.16: 61727100-61727100 on Build GRCh38
rs1020239649 Chr.16: 61727452-61727452 on Build GRCh38
rs760088513 Chr.16: 61727070-61727070 on Build GRCh38
rs763106444 Chr.16: 61727085-61727085 on Build GRCh38
rs747555647 Chr.16: 61727129-61727129 on Build GRCh38
rs750157697 Chr.16: 61727027-61727027 on Build GRCh38
rs764905971 Chr.16: 61727075-61727075 on Build GRCh38
rs777400821 Chr.16: 61727250-61727250 on Build GRCh38
rs752710948 Chr.16: 61727236-61727236 on Build GRCh38
rs1056510110 Chr.16: 61727409-61727409 on Build GRCh38
rs762899072 Chr.16: 61727185-61727185 on Build GRCh38
rs942170975 Chr.16: 61727451-61727451 on Build GRCh38
rs565353354 Chr.16: 61727390-61727391 on Build GRCh38
rs757799764 Chr.16: 61727248-61727248 on Build GRCh38
rs371471776 Chr.16: 61727099-61727099 on Build GRCh38
rs753621691 Chr.16: 61727202-61727202 on Build GRCh38
rs201384725 Chr.16: 61727048-61727048 on Build GRCh38
rs967068615 Chr.16: 61727131-61727131 on Build GRCh38
rs376009677 Chr.16: 61727047-61727047 on Build GRCh38
rs755904579 Chr.16: 61727030-61727030 on Build GRCh38
rs778113916 Chr.16: 61727025-61727025 on Build GRCh38
rs1050166449 Chr.16: 61727354-61727354 on Build GRCh38
rs867143514 Chr.16: 61727107-61727107 on Build GRCh38
rs563873222 Chr.16: 61727353-61727353 on Build GRCh38
rs776261356 Chr.16: 61727226-61727227 on Build GRCh38
rs908973882 Chr.16: 61727422-61727422 on Build GRCh38
rs972324295 Chr.16: 61727246-61727246 on Build GRCh38
rs140846478 Chr.16: 61727389-61727390 on Build GRCh38
rs913126767 Chr.16: 61727268-61727268 on Build GRCh38
rs770747257 Chr.16: 61727142-61727142 on Build GRCh38
rs765207880 Chr.16: 61727000-61727000 on Build GRCh38
rs185572745 Chr.16: 61727386-61727386 on Build GRCh38
rs753677900 Chr.16: 61727108-61727108 on Build GRCh38
rs781004813 Chr.16: 61727267-61727267 on Build GRCh38
rs138335076 Chr.16: 61727434-61727434 on Build GRCh38
rs538593171 Chr.16: 61727196-61727196 on Build GRCh38
rs866223525 Chr.16: 61727312-61727312 on Build GRCh38
rs777130460 Chr.16: 61727148-61727148 on Build GRCh38
rs770141191 Chr.16: 61727177-61727177 on Build GRCh38
rs755069578 Chr.16: 61726999-61726999 on Build GRCh38
rs895676731 Chr.16: 61727421-61727421 on Build GRCh38
rs374298300 Chr.16: 61727136-61727136 on Build GRCh38
rs758542382 Chr.16: 61727018-61727018 on Build GRCh38
rs776066801 Chr.16: 61727180-61727180 on Build GRCh38
rs945694686 Chr.16: 61727026-61727026 on Build GRCh38
rs1049070689 Chr.16: 61727361-61727361 on Build GRCh38
rs771617779 Chr.16: 61727135-61727135 on Build GRCh38
rs779805931 Chr.16: 61727103-61727103 on Build GRCh38
rs751684596 Chr.16: 61727194-61727194 on Build GRCh38
rs751883489 Chr.16: 61727336-61727336 on Build GRCh38
rs1032785863 Chr.16: 61727159-61727159 on Build GRCh38
rs746583218 Chr.16: 61727265-61727265 on Build GRCh38
rs754869691 Chr.16: 61727118-61727118 on Build GRCh38
rs1034179462 Chr.16: 61727153-61727153 on Build GRCh38
rs755267462 Chr.16: 61727292-61727292 on Build GRCh38
rs765017822 Chr.16: 61727208-61727208 on Build GRCh38
rs1050536395 Chr.16: 61727062-61727062 on Build GRCh38
rs985870634 Chr.16: 61727183-61727183 on Build GRCh38
rs978409824 Chr.16: 61727157-61727157 on Build GRCh38
rs781000085 Chr.16: 61727323-61727323 on Build GRCh38
rs768691738 Chr.16: 61727042-61727042 on Build GRCh38
rs749242596 Chr.16: 61727036-61727036 on Build GRCh38
rs771613272 Chr.16: 61727045-61727045 on Build GRCh38
rs975761301 Chr.16: 61727318-61727318 on Build GRCh38
rs150448972 Chr.16: 61727127-61727127 on Build GRCh38
rs761994642 Chr.16: 61727201-61727201 on Build GRCh38
rs938105161 Chr.16: 61727394-61727394 on Build GRCh38
rs967713809 Chr.16: 61727309-61727309 on Build GRCh38
rs754819538 Chr.16: 61727207-61727207 on Build GRCh38
rs752339349 Chr.16: 61727079-61727079 on Build GRCh38
rs764152984 Chr.16: 61727086-61727086 on Build GRCh38
rs760001992 Chr.16: 61727154-61727154 on Build GRCh38

View Full Product Details