Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 66609318-66609812 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CMTM3
Entrez Gene ID
123920
Gene Name
CKLF like MARVEL transmembrane domain containing 3
Gene Aliases
BNAS2, CKLFSF3
UniGene
Hs.298198
Gene Chromosome Location
Chr.16: 66604032-66613892 on Build GRCh38
Transcripts
NM_144601.3 NM_181553.2 NR_037613.1 NR_037619.1 XM_006721131.1 XM_011522861.1 XM_017022954.1

Gene Symbol
CMTM4
Entrez Gene ID
146223
Gene Name
CKLF like MARVEL transmembrane domain containing 4
Gene Aliases
CKLFSF4
UniGene
Hs.643961
Gene Chromosome Location
Chr.16: 66598178-66696707 on Build GRCh38
Transcripts
NM_144601.3 NM_181553.2 NR_037613.1 NR_037619.1 XM_006721131.1 XM_011522861.1 XM_017022954.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1378784 c.313C>T p.R105C Chr.16: 66609444-66609444 on Build GRCh38
1378785 c.364G>A p.A122T Chr.16: 66609495-66609495 on Build GRCh38
42790 c.336C>T p.I112I Chr.16: 66609467-66609467 on Build GRCh38
972193 c.304-2A>G p.? Chr.16: 66609433-66609433 on Build GRCh38
4061681 c.375G>A p.S125S Chr.16: 66609506-66609506 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765724187 Chr.16: 66609464-66609464 on Build GRCh38
rs760050557 Chr.16: 66609391-66609391 on Build GRCh38
rs767898016 Chr.16: 66609559-66609559 on Build GRCh38
rs974156047 Chr.16: 66609654-66609654 on Build GRCh38
rs1043479325 Chr.16: 66609767-66609779 on Build GRCh38
rs763529011 Chr.16: 66609536-66609536 on Build GRCh38
rs374945743 Chr.16: 66609445-66609445 on Build GRCh38
rs1033133363 Chr.16: 66609681-66609681 on Build GRCh38
rs780008717 Chr.16: 66609428-66609428 on Build GRCh38
rs554990064 Chr.16: 66609384-66609384 on Build GRCh38
rs774090424 Chr.16: 66609359-66609359 on Build GRCh38
rs564292546 Chr.16: 66609692-66609692 on Build GRCh38
rs201534476 Chr.16: 66609483-66609483 on Build GRCh38
rs961140369 Chr.16: 66609634-66609634 on Build GRCh38
rs777282641 Chr.16: 66609505-66609505 on Build GRCh38
rs968243920 Chr.16: 66609373-66609373 on Build GRCh38
rs749749982 Chr.16: 66609778-66609778 on Build GRCh38
rs776957337 Chr.16: 66609632-66609632 on Build GRCh38
rs71392154 Chr.16: 66609588-66609588 on Build GRCh38
rs535310604 Chr.16: 66609358-66609358 on Build GRCh38
rs71376802 Chr.16: 66609387-66609387 on Build GRCh38
rs890943696 Chr.16: 66609506-66609506 on Build GRCh38
rs372961322 Chr.16: 66609424-66609424 on Build GRCh38
rs766268585 Chr.16: 66609488-66609488 on Build GRCh38
rs771727928 Chr.16: 66609446-66609446 on Build GRCh38
rs749166146 Chr.16: 66609435-66609435 on Build GRCh38
rs758364559 Chr.16: 66609732-66609732 on Build GRCh38
rs1031286828 Chr.16: 66609623-66609623 on Build GRCh38
rs544377670 Chr.16: 66609688-66609688 on Build GRCh38
rs762022648 Chr.16: 66609790-66609790 on Build GRCh38
rs967021550 Chr.16: 66609569-66609569 on Build GRCh38
rs1014066228 Chr.16: 66609352-66609352 on Build GRCh38
rs957124743 Chr.16: 66609682-66609682 on Build GRCh38
rs948083873 Chr.16: 66609650-66609650 on Build GRCh38
rs768288339 Chr.16: 66609356-66609356 on Build GRCh38
rs12925258 Chr.16: 66609693-66609693 on Build GRCh38
rs372483559 Chr.16: 66609495-66609495 on Build GRCh38
rs746580386 Chr.16: 66609747-66609747 on Build GRCh38
rs114592261 Chr.16: 66609344-66609344 on Build GRCh38
rs992316112 Chr.16: 66609641-66609641 on Build GRCh38
rs774634862 Chr.16: 66609541-66609541 on Build GRCh38
rs758200035 Chr.16: 66609775-66609776 on Build GRCh38
rs752802447 Chr.16: 66609469-66609469 on Build GRCh38
rs765315953 Chr.16: 66609667-66609668 on Build GRCh38
rs760346957 Chr.16: 66609456-66609456 on Build GRCh38
rs768776663 Chr.16: 66609443-66609443 on Build GRCh38
rs760032543 Chr.16: 66609463-66609463 on Build GRCh38
rs903007565 Chr.16: 66609653-66609653 on Build GRCh38
rs764254878 Chr.16: 66609475-66609475 on Build GRCh38
rs530837986 Chr.16: 66609567-66609567 on Build GRCh38
rs750309642 Chr.16: 66609747-66609748 on Build GRCh38
rs758635322 Chr.16: 66609406-66609406 on Build GRCh38
rs757801687 Chr.16: 66609503-66609503 on Build GRCh38
rs200389814 Chr.16: 66609533-66609533 on Build GRCh38
rs778469660 Chr.16: 66609444-66609444 on Build GRCh38
rs769993909 Chr.16: 66609777-66609777 on Build GRCh38
rs545909802 Chr.16: 66609487-66609487 on Build GRCh38
rs557142222 Chr.16: 66609455-66609455 on Build GRCh38
rs764013496 Chr.16: 66609540-66609540 on Build GRCh38
rs777261968 Chr.16: 66609745-66609745 on Build GRCh38
rs762899233 Chr.16: 66609470-66609470 on Build GRCh38
rs573186551 Chr.16: 66609524-66609524 on Build GRCh38
rs770637677 Chr.16: 66609514-66609514 on Build GRCh38
rs745555556 Chr.16: 66609768-66609768 on Build GRCh38
rs1056944664 Chr.16: 66609392-66609392 on Build GRCh38
rs930197138 Chr.16: 66609548-66609548 on Build GRCh38
rs766764761 Chr.16: 66609385-66609385 on Build GRCh38
rs1055818377 Chr.16: 66609599-66609599 on Build GRCh38
rs746598469 Chr.16: 66609509-66609509 on Build GRCh38
rs916761734 Chr.16: 66609647-66609647 on Build GRCh38
rs770223705 Chr.16: 66609525-66609525 on Build GRCh38
rs748151460 Chr.16: 66609502-66609502 on Build GRCh38
rs111953008 Chr.16: 66609642-66609642 on Build GRCh38
rs568635141 Chr.16: 66609386-66609386 on Build GRCh38
rs774548967 Chr.16: 66609789-66609789 on Build GRCh38
rs780865235 Chr.16: 66609764-66609764 on Build GRCh38
rs752522325 Chr.16: 66609710-66609710 on Build GRCh38
rs939574913 Chr.16: 66609387-66609387 on Build GRCh38
rs1047407278 Chr.16: 66609717-66609717 on Build GRCh38
rs181826390 Chr.16: 66609774-66609774 on Build GRCh38
rs149217026 Chr.16: 66609494-66609494 on Build GRCh38
rs756882898 Chr.16: 66609748-66609748 on Build GRCh38
rs777894557 Chr.16: 66609411-66609411 on Build GRCh38
rs769263896 Chr.16: 66609783-66609783 on Build GRCh38
rs537288167 Chr.16: 66609412-66609412 on Build GRCh38
rs143994929 Chr.16: 66609457-66609457 on Build GRCh38
rs537078418 Chr.16: 66609405-66609405 on Build GRCh38
rs754202973 Chr.16: 66609639-66609639 on Build GRCh38
rs888541640 Chr.16: 66609583-66609583 on Build GRCh38
rs965714608 Chr.16: 66609728-66609728 on Build GRCh38
rs762110397 Chr.16: 66609545-66609545 on Build GRCh38
rs146435793 Chr.16: 66609458-66609458 on Build GRCh38
rs140101654 Chr.16: 66609566-66609566 on Build GRCh38

View Full Product Details