Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 67200090-67200579 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELMO3
Entrez Gene ID
79767
Gene Name
engulfment and cell motility 3
Gene Aliases
CED-12, CED12, ELMO-3
UniGene
Hs.377416
Gene Chromosome Location
Chr.16: 67199125-67204024 on Build GRCh38
Transcripts
NM_024712.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4571921 c.584T>A p.V195E Chr.16: 67200462-67200462 on Build GRCh38
5480384 c.468C>A p.A156A Chr.16: 67200257-67200257 on Build GRCh38
6991607 c.620T>C p.M207T Chr.16: 67200498-67200498 on Build GRCh38
5731310 c.526C>T p.R176C Chr.16: 67200315-67200315 on Build GRCh38
7249193 c.442A>G p.N148D Chr.16: 67200231-67200231 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763962241 Chr.16: 67200475-67200475 on Build GRCh38
rs780370445 Chr.16: 67200373-67200373 on Build GRCh38
rs762064736 Chr.16: 67200233-67200233 on Build GRCh38
rs775151130 Chr.16: 67200472-67200472 on Build GRCh38
rs749926112 Chr.16: 67200479-67200479 on Build GRCh38
rs1012759432 Chr.16: 67200556-67200556 on Build GRCh38
rs780343244 Chr.16: 67200435-67200435 on Build GRCh38
rs374005379 Chr.16: 67200369-67200369 on Build GRCh38
rs771217814 Chr.16: 67200470-67200470 on Build GRCh38
rs764403831 Chr.16: 67200217-67200217 on Build GRCh38
rs749610608 Chr.16: 67200442-67200442 on Build GRCh38
rs565642405 Chr.16: 67200502-67200502 on Build GRCh38
rs772442840 Chr.16: 67200382-67200382 on Build GRCh38
rs751021533 Chr.16: 67200353-67200353 on Build GRCh38
rs746057451 Chr.16: 67200516-67200516 on Build GRCh38
rs543222303 Chr.16: 67200153-67200153 on Build GRCh38
rs142718318 Chr.16: 67200406-67200416 on Build GRCh38
rs772715477 Chr.16: 67200559-67200559 on Build GRCh38
rs374854997 Chr.16: 67200314-67200314 on Build GRCh38
rs1030666836 Chr.16: 67200419-67200419 on Build GRCh38
rs963357038 Chr.16: 67200460-67200460 on Build GRCh38
rs755219354 Chr.16: 67200525-67200526 on Build GRCh38
rs774310307 Chr.16: 67200378-67200378 on Build GRCh38
rs369252219 Chr.16: 67200338-67200338 on Build GRCh38
rs181679927 Chr.16: 67200529-67200529 on Build GRCh38
rs894354171 Chr.16: 67200267-67200267 on Build GRCh38
rs553420551 Chr.16: 67200461-67200461 on Build GRCh38
rs771941795 Chr.16: 67200555-67200555 on Build GRCh38
rs755417338 Chr.16: 67200489-67200489 on Build GRCh38
rs756709609 Chr.16: 67200422-67200422 on Build GRCh38
rs753663503 Chr.16: 67200232-67200233 on Build GRCh38
rs754963278 Chr.16: 67200146-67200146 on Build GRCh38
rs773614482 Chr.16: 67200383-67200383 on Build GRCh38
rs758960655 Chr.16: 67200505-67200505 on Build GRCh38
rs200393647 Chr.16: 67200351-67200351 on Build GRCh38
rs755762919 Chr.16: 67200431-67200431 on Build GRCh38
rs200290034 Chr.16: 67200255-67200255 on Build GRCh38
rs755854857 Chr.16: 67200469-67200469 on Build GRCh38
rs1002653034 Chr.16: 67200221-67200221 on Build GRCh38
rs370574106 Chr.16: 67200380-67200380 on Build GRCh38
rs749279793 Chr.16: 67200144-67200144 on Build GRCh38
rs942162562 Chr.16: 67200302-67200302 on Build GRCh38
rs780785375 Chr.16: 67200359-67200359 on Build GRCh38
rs749830785 Chr.16: 67200374-67200374 on Build GRCh38
rs535312917 Chr.16: 67200427-67200427 on Build GRCh38
rs8059662 Chr.16: 67200376-67200376 on Build GRCh38
rs762905199 Chr.16: 67200215-67200215 on Build GRCh38
rs754174587 Chr.16: 67200487-67200487 on Build GRCh38
rs779400342 Chr.16: 67200508-67200508 on Build GRCh38
rs761708248 Chr.16: 67200476-67200476 on Build GRCh38
rs199678800 Chr.16: 67200509-67200509 on Build GRCh38
rs376353251 Chr.16: 67200506-67200506 on Build GRCh38
rs762771202 Chr.16: 67200473-67200473 on Build GRCh38
rs746647149 Chr.16: 67200173-67200173 on Build GRCh38
rs758350343 Chr.16: 67200341-67200341 on Build GRCh38
rs777306471 Chr.16: 67200281-67200281 on Build GRCh38
rs1026702533 Chr.16: 67200366-67200366 on Build GRCh38
rs8058861 Chr.16: 67200231-67200231 on Build GRCh38
rs902946180 Chr.16: 67200209-67200209 on Build GRCh38
rs910632086 Chr.16: 67200457-67200457 on Build GRCh38
rs757891562 Chr.16: 67200260-67200260 on Build GRCh38
rs867432068 Chr.16: 67200109-67200109 on Build GRCh38
rs764022939 Chr.16: 67200401-67200401 on Build GRCh38
rs201561338 Chr.16: 67200532-67200532 on Build GRCh38
rs762810962 Chr.16: 67200394-67200394 on Build GRCh38
rs760091521 Chr.16: 67200185-67200185 on Build GRCh38
rs765098402 Chr.16: 67200336-67200336 on Build GRCh38
rs894041193 Chr.16: 67200180-67200180 on Build GRCh38
rs575799078 Chr.16: 67200116-67200116 on Build GRCh38
rs778745882 Chr.16: 67200389-67200390 on Build GRCh38
rs745751311 Chr.16: 67200285-67200285 on Build GRCh38
rs763498567 Chr.16: 67200315-67200315 on Build GRCh38
rs570176557 Chr.16: 67200292-67200292 on Build GRCh38
rs531083390 Chr.16: 67200322-67200322 on Build GRCh38
rs755469444 Chr.16: 67200444-67200444 on Build GRCh38
rs763969040 Chr.16: 67200184-67200184 on Build GRCh38
rs765190397 Chr.16: 67200385-67200385 on Build GRCh38
rs990020119 Chr.16: 67200474-67200474 on Build GRCh38
rs753713994 Chr.16: 67200324-67200324 on Build GRCh38
rs533198861 Chr.16: 67200249-67200249 on Build GRCh38
rs770318596 Chr.16: 67200305-67200305 on Build GRCh38
rs757083531 Chr.16: 67200385-67200385 on Build GRCh38
rs201641837 Chr.16: 67200360-67200360 on Build GRCh38
rs200007690 Chr.16: 67200176-67200176 on Build GRCh38
rs772691254 Chr.16: 67200163-67200163 on Build GRCh38
rs541007473 Chr.16: 67200201-67200201 on Build GRCh38
rs775341356 Chr.16: 67200389-67200389 on Build GRCh38
rs910747977 Chr.16: 67200119-67200119 on Build GRCh38
rs780572116 Chr.16: 67200246-67200246 on Build GRCh38
rs202117358 Chr.16: 67200181-67200181 on Build GRCh38
rs760944768 Chr.16: 67200384-67200384 on Build GRCh38
rs796369040 Chr.16: 67200256-67200256 on Build GRCh38
rs747018436 Chr.16: 67200507-67200507 on Build GRCh38
rs754967465 Chr.16: 67200247-67200247 on Build GRCh38
rs756909661 Chr.16: 67200356-67200356 on Build GRCh38
rs747593103 Chr.16: 67200154-67200154 on Build GRCh38
rs775555328 Chr.16: 67200208-67200208 on Build GRCh38
rs373706784 Chr.16: 67200483-67200483 on Build GRCh38
rs781134129 Chr.16: 67200534-67200535 on Build GRCh38
rs372726536 Chr.16: 67200284-67200284 on Build GRCh38
rs376143115 Chr.16: 67200467-67200467 on Build GRCh38
rs751063874 Chr.16: 67200410-67200410 on Build GRCh38
rs201206921 Chr.16: 67200316-67200316 on Build GRCh38
rs751777848 Chr.16: 67200224-67200224 on Build GRCh38
rs373446239 Chr.16: 67200452-67200452 on Build GRCh38
rs771787266 Chr.16: 67200157-67200157 on Build GRCh38
rs549598699 Chr.16: 67200335-67200335 on Build GRCh38
rs779424155 Chr.16: 67200445-67200445 on Build GRCh38
rs201579675 Chr.16: 67200283-67200283 on Build GRCh38
rs774488380 Chr.16: 67200319-67200319 on Build GRCh38
rs370571157 Chr.16: 67200482-67200482 on Build GRCh38
rs774013584 Chr.16: 67200531-67200531 on Build GRCh38

View Full Product Details