Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 80543150-80543659 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DYNLRB2
Entrez Gene ID
83657
Gene Name
dynein light chain roadblock-type 2
Gene Aliases
DNCL2B, DNLC2B, ROBLD2
UniGene
Hs.98849
Gene Chromosome Location
Chr.16: 80540259-80550811 on Build GRCh38
Transcripts
NM_001305017.1 NM_130897.2 NR_130942.1 NR_130943.1 XM_006721295.3 XM_006721296.3 XM_011523370.2 NR_120307.1

Gene Symbol
LOC102724084
Entrez Gene ID
102724084
Gene Name
uncharacterized LOC102724084
Gene Aliases
-
UniGene
Hs.147884
Gene Chromosome Location
Chr.16: 80155958-80563135 on Build GRCh38
Transcripts
NM_001305017.1 NM_130897.2 NR_130942.1 NR_130943.1 XM_006721295.3 XM_006721296.3 XM_011523370.2 NR_120307.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5971939 c.18A>G p.E6E Chr.16: 80543290-80543290 on Build GRCh38
6145373 c.10G>T p.V4L Chr.16: 80543282-80543282 on Build GRCh38
4063036 c.29G>T p.R10M Chr.16: 80543301-80543301 on Build GRCh38
6650670 c.70A>C p.N24H Chr.16: 80543342-80543342 on Build GRCh38
3377997 c.24A>G p.L8L Chr.16: 80543296-80543296 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771629484 Chr.16: 80543322-80543322 on Build GRCh38
rs549750864 Chr.16: 80543299-80543299 on Build GRCh38
rs892551678 Chr.16: 80543608-80543608 on Build GRCh38
rs779412868 Chr.16: 80543395-80543395 on Build GRCh38
rs1040814111 Chr.16: 80543205-80543205 on Build GRCh38
rs145340983 Chr.16: 80543490-80543490 on Build GRCh38
rs986532636 Chr.16: 80543451-80543451 on Build GRCh38
rs192989595 Chr.16: 80543425-80543425 on Build GRCh38
rs767688367 Chr.16: 80543370-80543370 on Build GRCh38
rs554796645 Chr.16: 80543235-80543235 on Build GRCh38
rs765177592 Chr.16: 80543352-80543352 on Build GRCh38
rs905037229 Chr.16: 80543287-80543287 on Build GRCh38
rs113601170 Chr.16: 80543290-80543290 on Build GRCh38
rs13332289 Chr.16: 80543313-80543313 on Build GRCh38
rs781405892 Chr.16: 80543281-80543281 on Build GRCh38
rs372358788 Chr.16: 80543411-80543411 on Build GRCh38
rs780260536 Chr.16: 80543386-80543386 on Build GRCh38
rs199533524 Chr.16: 80543351-80543351 on Build GRCh38
rs772797116 Chr.16: 80543325-80543325 on Build GRCh38
rs149421698 Chr.16: 80543282-80543282 on Build GRCh38
rs762750635 Chr.16: 80543359-80543359 on Build GRCh38
rs566307552 Chr.16: 80543404-80543404 on Build GRCh38
rs370829650 Chr.16: 80543438-80543438 on Build GRCh38
rs1045121300 Chr.16: 80543609-80543609 on Build GRCh38
rs8049769 Chr.16: 80543201-80543201 on Build GRCh38
rs1005493141 Chr.16: 80543277-80543277 on Build GRCh38
rs370874644 Chr.16: 80543321-80543321 on Build GRCh38
rs757449611 Chr.16: 80543269-80543269 on Build GRCh38
rs545455831 Chr.16: 80543620-80543620 on Build GRCh38
rs796474737 Chr.16: 80543464-80543464 on Build GRCh38
rs956888664 Chr.16: 80543363-80543363 on Build GRCh38
rs888139762 Chr.16: 80543527-80543527 on Build GRCh38
rs900794679 Chr.16: 80543210-80543210 on Build GRCh38
rs760114193 Chr.16: 80543239-80543239 on Build GRCh38
rs764914966 Chr.16: 80543261-80543261 on Build GRCh38
rs949120549 Chr.16: 80543607-80543607 on Build GRCh38
rs752483808 Chr.16: 80543266-80543266 on Build GRCh38
rs929784174 Chr.16: 80543586-80543586 on Build GRCh38
rs771169833 Chr.16: 80543401-80543401 on Build GRCh38
rs756301749 Chr.16: 80543378-80543378 on Build GRCh38
rs750669262 Chr.16: 80543371-80543371 on Build GRCh38
rs982582152 Chr.16: 80543590-80543590 on Build GRCh38
rs767151835 Chr.16: 80543238-80543238 on Build GRCh38
rs184591343 Chr.16: 80543449-80543449 on Build GRCh38
rs988096447 Chr.16: 80543376-80543376 on Build GRCh38
rs769114763 Chr.16: 80543310-80543310 on Build GRCh38
rs1046746702 Chr.16: 80543500-80543500 on Build GRCh38
rs746842434 Chr.16: 80543393-80543393 on Build GRCh38
rs759276976 Chr.16: 80543379-80543380 on Build GRCh38
rs749780318 Chr.16: 80543309-80543309 on Build GRCh38
rs780773664 Chr.16: 80543301-80543301 on Build GRCh38
rs922367531 Chr.16: 80543476-80543476 on Build GRCh38
rs753920941 Chr.16: 80543389-80543389 on Build GRCh38
rs775925620 Chr.16: 80543542-80543542 on Build GRCh38
rs184768018 Chr.16: 80543602-80543602 on Build GRCh38
rs764654070 Chr.16: 80543625-80543625 on Build GRCh38
rs200686705 Chr.16: 80543335-80543335 on Build GRCh38
rs1003387603 Chr.16: 80543557-80543557 on Build GRCh38
rs554832795 Chr.16: 80543429-80543429 on Build GRCh38
rs754578081 Chr.16: 80543172-80543172 on Build GRCh38
rs986763062 Chr.16: 80543224-80543224 on Build GRCh38
rs188176701 Chr.16: 80543531-80543531 on Build GRCh38
rs754838370 Chr.16: 80543248-80543248 on Build GRCh38
rs766320458 Chr.16: 80543246-80543246 on Build GRCh38
rs929754011 Chr.16: 80543499-80543499 on Build GRCh38
rs377126729 Chr.16: 80543583-80543583 on Build GRCh38
rs773061156 Chr.16: 80543230-80543230 on Build GRCh38
rs760238854 Chr.16: 80543330-80543330 on Build GRCh38
rs748122622 Chr.16: 80543182-80543182 on Build GRCh38
rs751940801 Chr.16: 80543635-80543635 on Build GRCh38
rs148127589 Chr.16: 80543336-80543336 on Build GRCh38
rs577335261 Chr.16: 80543181-80543181 on Build GRCh38
rs752436279 Chr.16: 80543357-80543357 on Build GRCh38
rs919726349 Chr.16: 80543368-80543368 on Build GRCh38
rs944912023 Chr.16: 80543437-80543437 on Build GRCh38
rs968253793 Chr.16: 80543406-80543406 on Build GRCh38
rs1009666136 Chr.16: 80543433-80543433 on Build GRCh38
rs905444224 Chr.16: 80543410-80543410 on Build GRCh38
rs768896817 Chr.16: 80543226-80543226 on Build GRCh38
rs531267087 Chr.16: 80543236-80543236 on Build GRCh38
rs191179085 Chr.16: 80543541-80543541 on Build GRCh38
rs753765981 Chr.16: 80543247-80543247 on Build GRCh38
rs558957954 Chr.16: 80543504-80543504 on Build GRCh38

View Full Product Details