Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 81264514-81264990 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCO1
Entrez Gene ID
53630
Gene Name
beta-carotene oxygenase 1
Gene Aliases
BCDO, BCDO1, BCMO, BCMO1, BCO
UniGene
Hs.212172
Gene Chromosome Location
Chr.16: 81238642-81291142 on Build GRCh38
Transcripts
NM_017429.2 XM_011523109.1 XM_017023286.1 XM_017023287.1 XM_017023288.1 XM_017023289.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4063096 c.509C>T p.T170M Chr.16: 81264677-81264677 on Build GRCh38
5456575 c.489C>T p.Y163Y Chr.16: 81264657-81264657 on Build GRCh38
7185222 c.546A>G p.L182L Chr.16: 81264714-81264714 on Build GRCh38
3988687 c.515A>G p.H172R Chr.16: 81264683-81264683 on Build GRCh38
974082 c.616C>T p.P206S Chr.16: 81264784-81264784 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1000298862 Chr.16: 81264605-81264605 on Build GRCh38
rs72833320 Chr.16: 81264932-81264932 on Build GRCh38
rs369214485 Chr.16: 81264663-81264663 on Build GRCh38
rs774889298 Chr.16: 81264608-81264608 on Build GRCh38
rs927100268 Chr.16: 81264963-81264963 on Build GRCh38
rs746764244 Chr.16: 81264770-81264770 on Build GRCh38
rs113894835 Chr.16: 81264766-81264766 on Build GRCh38
rs759049294 Chr.16: 81264657-81264657 on Build GRCh38
rs745782751 Chr.16: 81264780-81264780 on Build GRCh38
rs991896131 Chr.16: 81264873-81264873 on Build GRCh38
rs941750832 Chr.16: 81264580-81264580 on Build GRCh38
rs764757293 Chr.16: 81264658-81264658 on Build GRCh38
rs552460042 Chr.16: 81264582-81264582 on Build GRCh38
rs899907454 Chr.16: 81264886-81264886 on Build GRCh38
rs542676422 Chr.16: 81264849-81264849 on Build GRCh38
rs566283561 Chr.16: 81264601-81264601 on Build GRCh38
rs541357816 Chr.16: 81264806-81264806 on Build GRCh38
rs960469944 Chr.16: 81264870-81264870 on Build GRCh38
rs770459452 Chr.16: 81264590-81264590 on Build GRCh38
rs574321964 Chr.16: 81264847-81264847 on Build GRCh38
rs747391927 Chr.16: 81264816-81264816 on Build GRCh38
rs756349704 Chr.16: 81264645-81264645 on Build GRCh38
rs777920537 Chr.16: 81264836-81264836 on Build GRCh38
rs139667928 Chr.16: 81264664-81264664 on Build GRCh38
rs375261365 Chr.16: 81264818-81264818 on Build GRCh38
rs180793034 Chr.16: 81264904-81264904 on Build GRCh38
rs1057400617 Chr.16: 81264570-81264570 on Build GRCh38
rs746352143 Chr.16: 81264589-81264589 on Build GRCh38
rs758437920 Chr.16: 81264830-81264830 on Build GRCh38
rs756673329 Chr.16: 81264675-81264675 on Build GRCh38
rs148919906 Chr.16: 81264662-81264662 on Build GRCh38
rs762379928 Chr.16: 81264800-81264800 on Build GRCh38
rs774899787 Chr.16: 81264725-81264725 on Build GRCh38
rs927018709 Chr.16: 81264808-81264808 on Build GRCh38
rs143043516 Chr.16: 81264696-81264696 on Build GRCh38
rs983240201 Chr.16: 81264538-81264538 on Build GRCh38
rs909560718 Chr.16: 81264670-81264670 on Build GRCh38
rs765677522 Chr.16: 81264823-81264823 on Build GRCh38
rs772250710 Chr.16: 81264699-81264699 on Build GRCh38
rs867118720 Chr.16: 81264931-81264931 on Build GRCh38
rs926231795 Chr.16: 81264923-81264923 on Build GRCh38
rs112981006 Chr.16: 81264747-81264747 on Build GRCh38
rs1031676947 Chr.16: 81264621-81264621 on Build GRCh38
rs577100191 Chr.16: 81264896-81264896 on Build GRCh38
rs531622003 Chr.16: 81264607-81264607 on Build GRCh38
rs1006824357 Chr.16: 81264812-81264812 on Build GRCh38
rs778969138 Chr.16: 81264964-81264964 on Build GRCh38
rs549113902 Chr.16: 81264542-81264542 on Build GRCh38
rs200531385 Chr.16: 81264765-81264765 on Build GRCh38
rs765945610 Chr.16: 81264631-81264632 on Build GRCh38
rs752757562 Chr.16: 81264826-81264826 on Build GRCh38
rs119478057 Chr.16: 81264677-81264677 on Build GRCh38
rs778912655 Chr.16: 81264692-81264692 on Build GRCh38
rs188474473 Chr.16: 81264895-81264895 on Build GRCh38
rs570813648 Chr.16: 81264924-81264924 on Build GRCh38
rs769682221 Chr.16: 81264787-81264787 on Build GRCh38
rs904667001 Chr.16: 81264862-81264862 on Build GRCh38
rs1000751570 Chr.16: 81264864-81264864 on Build GRCh38
rs375537404 Chr.16: 81264687-81264687 on Build GRCh38
rs552746766 Chr.16: 81264822-81264822 on Build GRCh38
rs750755364 Chr.16: 81264730-81264730 on Build GRCh38
rs751586063 Chr.16: 81264685-81264685 on Build GRCh38
rs1029578972 Chr.16: 81264901-81264901 on Build GRCh38
rs954635835 Chr.16: 81264914-81264914 on Build GRCh38
rs963267724 Chr.16: 81264934-81264934 on Build GRCh38
rs372038083 Chr.16: 81264629-81264629 on Build GRCh38
rs371921845 Chr.16: 81264795-81264795 on Build GRCh38
rs531685028 Chr.16: 81264881-81264881 on Build GRCh38
rs149029789 Chr.16: 81264678-81264678 on Build GRCh38
rs779046372 Chr.16: 81264757-81264757 on Build GRCh38
rs776458742 Chr.16: 81264772-81264772 on Build GRCh38
rs771881545 Chr.16: 81264617-81264617 on Build GRCh38
rs776579175 Chr.16: 81264709-81264709 on Build GRCh38
rs973633425 Chr.16: 81264940-81264940 on Build GRCh38
rs371846416 Chr.16: 81264701-81264701 on Build GRCh38
rs917546796 Chr.16: 81264829-81264829 on Build GRCh38
rs372455855 Chr.16: 81264860-81264860 on Build GRCh38
rs145127976 Chr.16: 81264674-81264674 on Build GRCh38
rs145899743 Chr.16: 81264753-81264753 on Build GRCh38
rs755481903 Chr.16: 81264756-81264756 on Build GRCh38
rs770733875 Chr.16: 81264771-81264771 on Build GRCh38
rs147813781 Chr.16: 81264649-81264649 on Build GRCh38
rs151149493 Chr.16: 81264736-81264736 on Build GRCh38
rs199556335 Chr.16: 81264796-81264796 on Build GRCh38
rs369214670 Chr.16: 81264540-81264540 on Build GRCh38
rs917437205 Chr.16: 81264879-81264879 on Build GRCh38
rs200324875 Chr.16: 81264784-81264784 on Build GRCh38
rs568154975 Chr.16: 81264650-81264650 on Build GRCh38
rs763765997 Chr.16: 81264801-81264801 on Build GRCh38
rs780274924 Chr.16: 81264591-81264591 on Build GRCh38
rs762658812 Chr.16: 81264728-81264728 on Build GRCh38
rs375362122 Chr.16: 81264631-81264631 on Build GRCh38
rs201220518 Chr.16: 81264600-81264600 on Build GRCh38
rs536329686 Chr.16: 81264700-81264700 on Build GRCh38
rs770786848 Chr.16: 81264703-81264703 on Build GRCh38
rs764781169 Chr.16: 81264714-81264714 on Build GRCh38
rs762469254 Chr.16: 81264953-81264953 on Build GRCh38
rs748236344 Chr.16: 81264760-81264760 on Build GRCh38

View Full Product Details