Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 85611251-85611747 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GSE1
Entrez Gene ID
23199
Gene Name
Gse1 coiled-coil protein
Gene Aliases
CRHSP24, KIAA0182
UniGene
Hs.461647
Gene Chromosome Location
Chr.16: 85169512-85676206 on Build GRCh38
Transcripts
NM_001134473.2 XM_005255859.4 XM_005255860.3 XM_005255861.4 XM_005255863.4 XM_005255864.4 XM_005255865.4 XM_011522965.2 XM_017023083.1

SNP Detail

SNP ID Chromosome Location
rs1036957414 Chr.16: 85611386-85611386 on Build GRCh38
rs563224376 Chr.16: 85611410-85611410 on Build GRCh38
rs911830529 Chr.16: 85611603-85611603 on Build GRCh38
rs956661155 Chr.16: 85611570-85611570 on Build GRCh38
rs906377103 Chr.16: 85611545-85611545 on Build GRCh38
rs545190676 Chr.16: 85611365-85611365 on Build GRCh38
rs1040687320 Chr.16: 85611401-85611401 on Build GRCh38
rs918970846 Chr.16: 85611547-85611547 on Build GRCh38
rs1022281796 Chr.16: 85611573-85611573 on Build GRCh38
rs982114613 Chr.16: 85611275-85611275 on Build GRCh38
rs970417089 Chr.16: 85611351-85611351 on Build GRCh38
rs888300174 Chr.16: 85611323-85611323 on Build GRCh38
rs556658419 Chr.16: 85611572-85611572 on Build GRCh38
rs941775448 Chr.16: 85611387-85611387 on Build GRCh38
rs888121518 Chr.16: 85611671-85611671 on Build GRCh38
rs549803890 Chr.16: 85611366-85611366 on Build GRCh38
rs948504207 Chr.16: 85611311-85611311 on Build GRCh38
rs912061092 Chr.16: 85611477-85611477 on Build GRCh38
rs538202481 Chr.16: 85611551-85611551 on Build GRCh38
rs532900216 Chr.16: 85611649-85611649 on Build GRCh38
rs998110877 Chr.16: 85611488-85611488 on Build GRCh38
rs868487108 Chr.16: 85611277-85611277 on Build GRCh38
rs936636522 Chr.16: 85611645-85611645 on Build GRCh38
rs912027341 Chr.16: 85611615-85611615 on Build GRCh38
rs980461314 Chr.16: 85611363-85611363 on Build GRCh38
rs888177414 Chr.16: 85611525-85611525 on Build GRCh38
rs963349944 Chr.16: 85611527-85611527 on Build GRCh38
rs1030816552 Chr.16: 85611716-85611716 on Build GRCh38
rs780488438 Chr.16: 85611458-85611458 on Build GRCh38
rs534569279 Chr.16: 85611483-85611483 on Build GRCh38
rs1048076473 Chr.16: 85611664-85611664 on Build GRCh38
rs1023438515 Chr.16: 85611538-85611538 on Build GRCh38
rs536850800 Chr.16: 85611568-85611568 on Build GRCh38
rs866223977 Chr.16: 85611533-85611533 on Build GRCh38
rs919504674 Chr.16: 85611639-85611639 on Build GRCh38
rs890696115 Chr.16: 85611397-85611397 on Build GRCh38
rs1023911106 Chr.16: 85611431-85611431 on Build GRCh38
rs1051072297 Chr.16: 85611341-85611341 on Build GRCh38
rs980874216 Chr.16: 85611516-85611516 on Build GRCh38
rs890363197 Chr.16: 85611417-85611417 on Build GRCh38
rs919109600 Chr.16: 85611398-85611398 on Build GRCh38
rs1053817455 Chr.16: 85611307-85611318 on Build GRCh38
rs932083319 Chr.16: 85611466-85611466 on Build GRCh38
rs956708546 Chr.16: 85611455-85611455 on Build GRCh38
rs941146215 Chr.16: 85611329-85611329 on Build GRCh38
rs867746224 Chr.16: 85611697-85611697 on Build GRCh38
rs375671352 Chr.16: 85611429-85611429 on Build GRCh38
rs918141452 Chr.16: 85611332-85611332 on Build GRCh38
rs897023511 Chr.16: 85611403-85611403 on Build GRCh38
rs953461173 Chr.16: 85611604-85611604 on Build GRCh38
rs943315643 Chr.16: 85611613-85611613 on Build GRCh38
rs982450167 Chr.16: 85611445-85611445 on Build GRCh38
rs915371039 Chr.16: 85611318-85611318 on Build GRCh38
rs547989370 Chr.16: 85611506-85611506 on Build GRCh38
rs929101982 Chr.16: 85611405-85611405 on Build GRCh38
rs926389879 Chr.16: 85611377-85611377 on Build GRCh38
rs1033364524 Chr.16: 85611522-85611522 on Build GRCh38
rs866412678 Chr.16: 85611725-85611725 on Build GRCh38
rs539064127 Chr.16: 85611616-85611616 on Build GRCh38
rs532165113 Chr.16: 85611422-85611422 on Build GRCh38
rs944880244 Chr.16: 85611625-85611625 on Build GRCh38
rs896792834 Chr.16: 85611489-85611489 on Build GRCh38
rs1037827999 Chr.16: 85611395-85611395 on Build GRCh38
rs576443915 Chr.16: 85611614-85611614 on Build GRCh38
rs116818028 Chr.16: 85611699-85611699 on Build GRCh38
rs1045216864 Chr.16: 85611691-85611691 on Build GRCh38
rs970655711 Chr.16: 85611434-85611434 on Build GRCh38
rs972261922 Chr.16: 85611633-85611633 on Build GRCh38
rs944954619 Chr.16: 85611481-85611481 on Build GRCh38
rs932143861 Chr.16: 85611322-85611333 on Build GRCh38
rs565657609 Chr.16: 85611452-85611452 on Build GRCh38
rs749574242 Chr.16: 85611682-85611682 on Build GRCh38
rs929174035 Chr.16: 85611550-85611550 on Build GRCh38
rs1039687433 Chr.16: 85611438-85611438 on Build GRCh38
rs1048127226 Chr.16: 85611515-85611515 on Build GRCh38
rs989473801 Chr.16: 85611457-85611457 on Build GRCh38
rs892647488 Chr.16: 85611476-85611476 on Build GRCh38
rs1044151214 Chr.16: 85611317-85611317 on Build GRCh38
rs1032803434 Chr.16: 85611461-85611461 on Build GRCh38
rs899836243 Chr.16: 85611627-85611627 on Build GRCh38
rs556303097 Chr.16: 85611605-85611605 on Build GRCh38
rs1025270833 Chr.16: 85611496-85611496 on Build GRCh38

View Full Product Details