Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 85613226-85613720 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GSE1
Entrez Gene ID
23199
Gene Name
Gse1 coiled-coil protein
Gene Aliases
CRHSP24, KIAA0182
UniGene
Hs.461647
Gene Chromosome Location
Chr.16: 85169512-85676206 on Build GRCh38
Transcripts
NM_001134473.2 NM_001278184.1 NM_014615.3 XM_005255859.4 XM_005255860.3 XM_005255861.4 XM_005255863.4 XM_005255864.4 XM_005255865.4 XM_011522965.2 XM_011522966.2 XM_017023083.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4619443 c.4A>G p.K2E Chr.16: 85613395-85613395 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs371154038 Chr.16: 85613356-85613356 on Build GRCh38
rs758657285 Chr.16: 85613374-85613374 on Build GRCh38
rs898609867 Chr.16: 85613664-85613665 on Build GRCh38
rs1029746340 Chr.16: 85613378-85613378 on Build GRCh38
rs768228816 Chr.16: 85613436-85613436 on Build GRCh38
rs1041334774 Chr.16: 85613398-85613398 on Build GRCh38
rs899831595 Chr.16: 85613368-85613368 on Build GRCh38
rs528587639 Chr.16: 85613664-85613664 on Build GRCh38
rs548439395 Chr.16: 85613682-85613682 on Build GRCh38
rs544189275 Chr.16: 85613461-85613461 on Build GRCh38
rs891206008 Chr.16: 85613479-85613479 on Build GRCh38
rs373746902 Chr.16: 85613381-85613381 on Build GRCh38
rs761315818 Chr.16: 85613362-85613362 on Build GRCh38
rs999111121 Chr.16: 85613482-85613482 on Build GRCh38
rs995670817 Chr.16: 85613669-85613669 on Build GRCh38
rs915638593 Chr.16: 85613350-85613350 on Build GRCh38
rs1055728913 Chr.16: 85613699-85613699 on Build GRCh38
rs931186503 Chr.16: 85613353-85613353 on Build GRCh38
rs933282372 Chr.16: 85613434-85613434 on Build GRCh38
rs903085409 Chr.16: 85613349-85613349 on Build GRCh38
rs555346742 Chr.16: 85613446-85613446 on Build GRCh38
rs982985233 Chr.16: 85613252-85613252 on Build GRCh38
rs922501339 Chr.16: 85613540-85613540 on Build GRCh38
rs753150412 Chr.16: 85613370-85613370 on Build GRCh38
rs967612625 Chr.16: 85613310-85613310 on Build GRCh38
rs961153096 Chr.16: 85613531-85613531 on Build GRCh38
rs762122898 Chr.16: 85613503-85613503 on Build GRCh38
rs891257975 Chr.16: 85613389-85613389 on Build GRCh38
rs193217765 Chr.16: 85613450-85613450 on Build GRCh38
rs540440833 Chr.16: 85613577-85613577 on Build GRCh38
rs1036318297 Chr.16: 85613490-85613490 on Build GRCh38
rs757400279 Chr.16: 85613386-85613386 on Build GRCh38
rs749139594 Chr.16: 85613435-85613435 on Build GRCh38
rs996957460 Chr.16: 85613248-85613248 on Build GRCh38
rs1050845967 Chr.16: 85613570-85613570 on Build GRCh38
rs917030613 Chr.16: 85613336-85613336 on Build GRCh38
rs572620021 Chr.16: 85613438-85613438 on Build GRCh38
rs867132303 Chr.16: 85613442-85613442 on Build GRCh38
rs949885180 Chr.16: 85613338-85613338 on Build GRCh38
rs947474115 Chr.16: 85613582-85613582 on Build GRCh38
rs774771821 Chr.16: 85613417-85613417 on Build GRCh38
rs903330493 Chr.16: 85613476-85613476 on Build GRCh38
rs937062871 Chr.16: 85613320-85613320 on Build GRCh38
rs1035380658 Chr.16: 85613324-85613324 on Build GRCh38
rs971096338 Chr.16: 85613554-85613554 on Build GRCh38
rs957218533 Chr.16: 85613572-85613572 on Build GRCh38
rs1011782550 Chr.16: 85613457-85613457 on Build GRCh38
rs974370975 Chr.16: 85613425-85613426 on Build GRCh38
rs554405869 Chr.16: 85613550-85613550 on Build GRCh38
rs1043123251 Chr.16: 85613589-85613589 on Build GRCh38
rs759935507 Chr.16: 85613449-85613449 on Build GRCh38
rs908399644 Chr.16: 85613525-85613525 on Build GRCh38
rs924405289 Chr.16: 85613571-85613571 on Build GRCh38
rs903360047 Chr.16: 85613604-85613604 on Build GRCh38
rs940048745 Chr.16: 85613637-85613637 on Build GRCh38
rs1023938393 Chr.16: 85613552-85613552 on Build GRCh38
rs376861316 Chr.16: 85613621-85613621 on Build GRCh38
rs896978758 Chr.16: 85613569-85613569 on Build GRCh38
rs866491880 Chr.16: 85613257-85613257 on Build GRCh38
rs1037128278 Chr.16: 85613648-85613648 on Build GRCh38
rs756126377 Chr.16: 85613420-85613420 on Build GRCh38
rs952451343 Chr.16: 85613515-85613515 on Build GRCh38
rs1037311521 Chr.16: 85613568-85613568 on Build GRCh38
rs545420284 Chr.16: 85613299-85613299 on Build GRCh38
rs1011751375 Chr.16: 85613278-85613278 on Build GRCh38
rs376195898 Chr.16: 85613415-85613415 on Build GRCh38
rs1027695374 Chr.16: 85613427-85613427 on Build GRCh38
rs781401738 Chr.16: 85613392-85613392 on Build GRCh38
rs373660795 Chr.16: 85613345-85613345 on Build GRCh38
rs368496384 Chr.16: 85613341-85613341 on Build GRCh38
rs1004366819 Chr.16: 85613414-85613414 on Build GRCh38
rs932586093 Chr.16: 85613618-85613618 on Build GRCh38
rs896037557 Chr.16: 85613634-85613634 on Build GRCh38
rs977195501 Chr.16: 85613450-85613450 on Build GRCh38
rs773810463 Chr.16: 85613361-85613361 on Build GRCh38
rs983805967 Chr.16: 85613518-85613518 on Build GRCh38
rs977643082 Chr.16: 85613317-85613317 on Build GRCh38
rs766811967 Chr.16: 85613363-85613363 on Build GRCh38
rs867267982 Chr.16: 85613251-85613251 on Build GRCh38
rs551030631 Chr.16: 85613292-85613292 on Build GRCh38
rs889296858 Chr.16: 85613254-85613254 on Build GRCh38
rs779664315 Chr.16: 85613422-85613422 on Build GRCh38
rs748871873 Chr.16: 85613357-85613357 on Build GRCh38
rs370532396 Chr.16: 85613688-85613688 on Build GRCh38
rs935875963 Chr.16: 85613452-85613452 on Build GRCh38
rs776986993 Chr.16: 85613366-85613366 on Build GRCh38
rs977141428 Chr.16: 85613566-85613566 on Build GRCh38
rs912586200 Chr.16: 85613627-85613629 on Build GRCh38
rs1022735336 Chr.16: 85613506-85613506 on Build GRCh38
rs371365008 Chr.16: 85613344-85613344 on Build GRCh38
rs768318801 Chr.16: 85613358-85613358 on Build GRCh38
rs976703355 Chr.16: 85613533-85613533 on Build GRCh38
rs377753359 Chr.16: 85613307-85613307 on Build GRCh38
rs921174711 Chr.16: 85613593-85613593 on Build GRCh38
rs747853083 Chr.16: 85613439-85613439 on Build GRCh38
rs979862932 Chr.16: 85613255-85613255 on Build GRCh38
rs959769166 Chr.16: 85613495-85613495 on Build GRCh38
rs963260742 Chr.16: 85613424-85613424 on Build GRCh38
rs1033932409 Chr.16: 85613528-85613528 on Build GRCh38
rs781295094 Chr.16: 85613343-85613343 on Build GRCh38
rs535163971 Chr.16: 85613445-85613445 on Build GRCh38
rs909597272 Chr.16: 85613268-85613268 on Build GRCh38
rs866581287 Chr.16: 85613642-85613642 on Build GRCh38
rs991558887 Chr.16: 85613505-85613505 on Build GRCh38
rs760153456 Chr.16: 85613367-85613367 on Build GRCh38
rs371557812 Chr.16: 85613377-85613377 on Build GRCh38
rs999422237 Chr.16: 85613607-85613607 on Build GRCh38
rs991202775 Chr.16: 85613331-85613331 on Build GRCh38
rs539673213 Chr.16: 85613332-85613332 on Build GRCh38

View Full Product Details