Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 85633762-85634263 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GSE1
Entrez Gene ID
23199
Gene Name
Gse1 coiled-coil protein
Gene Aliases
CRHSP24, KIAA0182
UniGene
Hs.461647
Gene Chromosome Location
Chr.16: 85169512-85676206 on Build GRCh38
Transcripts
NM_001134473.2 NM_001278184.1 NM_014615.3 XM_005255859.4 XM_005255860.3 XM_005255861.4 XM_005255863.4 XM_005255864.4 XM_005255865.4 XM_011522965.2 XM_011522966.2 XM_017023083.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4695138 c.109G>A p.A37T Chr.16: 85634015-85634015 on Build GRCh38
6345515 c.140G>T p.S47I Chr.16: 85634046-85634046 on Build GRCh38
5625169 c.102C>G p.L34L Chr.16: 85634008-85634008 on Build GRCh38
6345514 c.70G>T p.A24S Chr.16: 85633976-85633976 on Build GRCh38
4695137 c.98C>T p.P33L Chr.16: 85634004-85634004 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759629419 Chr.16: 85634029-85634029 on Build GRCh38
rs549358932 Chr.16: 85634154-85634154 on Build GRCh38
rs925851727 Chr.16: 85634241-85634241 on Build GRCh38
rs753572545 Chr.16: 85633892-85633892 on Build GRCh38
rs766427270 Chr.16: 85633957-85633957 on Build GRCh38
rs562413164 Chr.16: 85633893-85633893 on Build GRCh38
rs760822694 Chr.16: 85633956-85633956 on Build GRCh38
rs776736483 Chr.16: 85633996-85633996 on Build GRCh38
rs772126001 Chr.16: 85633986-85633986 on Build GRCh38
rs769838290 Chr.16: 85634070-85634070 on Build GRCh38
rs571845741 Chr.16: 85634176-85634176 on Build GRCh38
rs766447038 Chr.16: 85633994-85633994 on Build GRCh38
rs36017765 Chr.16: 85633817-85633818 on Build GRCh38
rs751194039 Chr.16: 85634004-85634004 on Build GRCh38
rs758874796 Chr.16: 85634059-85634059 on Build GRCh38
rs999737688 Chr.16: 85634173-85634173 on Build GRCh38
rs756909155 Chr.16: 85634005-85634005 on Build GRCh38
rs188721700 Chr.16: 85634119-85634119 on Build GRCh38
rs771803711 Chr.16: 85633910-85633910 on Build GRCh38
rs1030642537 Chr.16: 85633819-85633819 on Build GRCh38
rs759430740 Chr.16: 85634243-85634243 on Build GRCh38
rs542526858 Chr.16: 85633867-85633867 on Build GRCh38
rs904923484 Chr.16: 85633795-85633795 on Build GRCh38
rs371764645 Chr.16: 85634118-85634118 on Build GRCh38
rs755964711 Chr.16: 85633887-85633887 on Build GRCh38
rs199588508 Chr.16: 85634051-85634051 on Build GRCh38
rs772503252 Chr.16: 85633904-85633904 on Build GRCh38
rs763866369 Chr.16: 85634002-85634002 on Build GRCh38
rs768773627 Chr.16: 85633941-85633941 on Build GRCh38
rs375816658 Chr.16: 85633960-85633960 on Build GRCh38
rs796619057 Chr.16: 85634023-85634023 on Build GRCh38
rs1046597549 Chr.16: 85634170-85634170 on Build GRCh38
rs375060429 Chr.16: 85633924-85633924 on Build GRCh38
rs555784895 Chr.16: 85634205-85634205 on Build GRCh38
rs991131553 Chr.16: 85633826-85633826 on Build GRCh38
rs371951850 Chr.16: 85633921-85633921 on Build GRCh38
rs753767802 Chr.16: 85633958-85633958 on Build GRCh38
rs773470396 Chr.16: 85633988-85633988 on Build GRCh38
rs964095236 Chr.16: 85633856-85633856 on Build GRCh38
rs141419803 Chr.16: 85634050-85634050 on Build GRCh38
rs759417177 Chr.16: 85633998-85633998 on Build GRCh38
rs778146704 Chr.16: 85634062-85634062 on Build GRCh38
rs780045942 Chr.16: 85633888-85633888 on Build GRCh38
rs775406138 Chr.16: 85634033-85634033 on Build GRCh38
rs370450221 Chr.16: 85634069-85634069 on Build GRCh38
rs147016831 Chr.16: 85634052-85634052 on Build GRCh38
rs200168352 Chr.16: 85634077-85634077 on Build GRCh38
rs919053302 Chr.16: 85634199-85634199 on Build GRCh38
rs867309792 Chr.16: 85633797-85633797 on Build GRCh38
rs200241797 Chr.16: 85634038-85634038 on Build GRCh38
rs202054304 Chr.16: 85633952-85633952 on Build GRCh38
rs771961427 Chr.16: 85634147-85634147 on Build GRCh38
rs200574628 Chr.16: 85634071-85634071 on Build GRCh38
rs780477042 Chr.16: 85633868-85633868 on Build GRCh38
rs533511744 Chr.16: 85633967-85633967 on Build GRCh38
rs941881260 Chr.16: 85633796-85633796 on Build GRCh38
rs566890495 Chr.16: 85633978-85633978 on Build GRCh38
rs747763298 Chr.16: 85633907-85633907 on Build GRCh38
rs781580362 Chr.16: 85634107-85634107 on Build GRCh38
rs143102373 Chr.16: 85633937-85633937 on Build GRCh38
rs760507308 Chr.16: 85634208-85634208 on Build GRCh38
rs746536663 Chr.16: 85633916-85633916 on Build GRCh38
rs989974955 Chr.16: 85633863-85633863 on Build GRCh38
rs769310279 Chr.16: 85633979-85633979 on Build GRCh38
rs551766960 Chr.16: 85634171-85634171 on Build GRCh38
rs974486336 Chr.16: 85633834-85633835 on Build GRCh38
rs770872004 Chr.16: 85634213-85634213 on Build GRCh38
rs143822889 Chr.16: 85633962-85633962 on Build GRCh38
rs764870639 Chr.16: 85634183-85634183 on Build GRCh38
rs148537143 Chr.16: 85634010-85634010 on Build GRCh38
rs776056520 Chr.16: 85633935-85633935 on Build GRCh38
rs746071025 Chr.16: 85634108-85634108 on Build GRCh38
rs972664874 Chr.16: 85634217-85634218 on Build GRCh38
rs767810394 Chr.16: 85633943-85633943 on Build GRCh38
rs756791311 Chr.16: 85633969-85633969 on Build GRCh38
rs1031374034 Chr.16: 85633976-85633976 on Build GRCh38
rs891141526 Chr.16: 85633825-85633825 on Build GRCh38
rs748526936 Chr.16: 85633985-85633985 on Build GRCh38
rs776686409 Chr.16: 85634242-85634242 on Build GRCh38
rs757616693 Chr.16: 85634099-85634099 on Build GRCh38
rs560356645 Chr.16: 85634095-85634095 on Build GRCh38
rs758978785 Chr.16: 85634177-85634177 on Build GRCh38
rs547932536 Chr.16: 85634167-85634167 on Build GRCh38
rs770026074 Chr.16: 85634113-85634113 on Build GRCh38
rs371349851 Chr.16: 85633871-85633871 on Build GRCh38
rs765961888 Chr.16: 85634093-85634093 on Build GRCh38
rs553778330 Chr.16: 85633846-85633846 on Build GRCh38
rs950513493 Chr.16: 85634201-85634201 on Build GRCh38
rs376208046 Chr.16: 85633874-85633874 on Build GRCh38
rs376074064 Chr.16: 85634068-85634068 on Build GRCh38
rs538956724 Chr.16: 85634240-85634240 on Build GRCh38
rs989900051 Chr.16: 85633989-85633989 on Build GRCh38
rs866224331 Chr.16: 85633980-85633980 on Build GRCh38
rs776050879 Chr.16: 85634136-85634137 on Build GRCh38
rs773573755 Chr.16: 85634207-85634207 on Build GRCh38
rs780802449 Chr.16: 85634007-85634007 on Build GRCh38
rs1011220536 Chr.16: 85634191-85634191 on Build GRCh38
rs569522691 Chr.16: 85634220-85634220 on Build GRCh38
rs924389807 Chr.16: 85633841-85633841 on Build GRCh38
rs529273255 Chr.16: 85633981-85633981 on Build GRCh38
rs201763364 Chr.16: 85633975-85633975 on Build GRCh38
rs562861824 Chr.16: 85634159-85634159 on Build GRCh38
rs1051714968 Chr.16: 85634009-85634009 on Build GRCh38
rs952680340 Chr.16: 85634225-85634225 on Build GRCh38
rs971145122 Chr.16: 85633836-85633836 on Build GRCh38
rs534103030 Chr.16: 85634184-85634184 on Build GRCh38
rs372516330 Chr.16: 85633963-85633963 on Build GRCh38
rs767473439 Chr.16: 85633881-85633881 on Build GRCh38
rs538416190 Chr.16: 85633799-85633799 on Build GRCh38
rs370372940 Chr.16: 85634017-85634017 on Build GRCh38
rs556048268 Chr.16: 85634058-85634058 on Build GRCh38
rs772479714 Chr.16: 85634016-85634016 on Build GRCh38
rs750095056 Chr.16: 85634090-85634090 on Build GRCh38
rs557891969 Chr.16: 85633800-85633800 on Build GRCh38
rs377684294 Chr.16: 85634039-85634039 on Build GRCh38
rs572129976 Chr.16: 85634089-85634089 on Build GRCh38
rs919867557 Chr.16: 85634218-85634218 on Build GRCh38
rs932561128 Chr.16: 85633899-85633899 on Build GRCh38
rs74737530 Chr.16: 85633973-85633973 on Build GRCh38
rs755468132 Chr.16: 85634011-85634011 on Build GRCh38
rs150719571 Chr.16: 85633922-85633922 on Build GRCh38
rs375526805 Chr.16: 85634053-85634053 on Build GRCh38
rs780143008 Chr.16: 85634117-85634117 on Build GRCh38
rs184424758 Chr.16: 85634030-85634030 on Build GRCh38
rs974659488 Chr.16: 85634229-85634229 on Build GRCh38
rs777095213 Chr.16: 85633915-85633915 on Build GRCh38
rs200247132 Chr.16: 85634001-85634001 on Build GRCh38
rs771071094 Chr.16: 85634067-85634067 on Build GRCh38
rs577627227 Chr.16: 85633837-85633837 on Build GRCh38
rs547471071 Chr.16: 85634195-85634195 on Build GRCh38
rs369127760 Chr.16: 85633906-85633906 on Build GRCh38
rs1039686088 Chr.16: 85634024-85634024 on Build GRCh38
rs752169202 Chr.16: 85634098-85634098 on Build GRCh38
rs761907985 Chr.16: 85634144-85634144 on Build GRCh38
rs540734949 Chr.16: 85634094-85634094 on Build GRCh38
rs912460657 Chr.16: 85634116-85634116 on Build GRCh38
rs776648561 Chr.16: 85634021-85634021 on Build GRCh38
rs763622327 Chr.16: 85633936-85633936 on Build GRCh38
rs1038904102 Chr.16: 85634156-85634156 on Build GRCh38
rs898682632 Chr.16: 85633808-85633808 on Build GRCh38
rs943290853 Chr.16: 85634141-85634141 on Build GRCh38
rs1002476352 Chr.16: 85634188-85634188 on Build GRCh38
rs538453007 Chr.16: 85634014-85634014 on Build GRCh38
rs762079929 Chr.16: 85633942-85633942 on Build GRCh38
rs779574898 Chr.16: 85634012-85634012 on Build GRCh38
rs73271255 Chr.16: 85634198-85634198 on Build GRCh38
rs773189258 Chr.16: 85634157-85634157 on Build GRCh38
rs761712147 Chr.16: 85634081-85634081 on Build GRCh38
rs995697166 Chr.16: 85633816-85633816 on Build GRCh38
rs1033904347 Chr.16: 85634192-85634192 on Build GRCh38
rs748390732 Chr.16: 85634092-85634092 on Build GRCh38
rs762841856 Chr.16: 85634037-85634037 on Build GRCh38
rs539943361 Chr.16: 85633840-85633840 on Build GRCh38
rs747361801 Chr.16: 85634064-85634064 on Build GRCh38
rs895324720 Chr.16: 85634054-85634054 on Build GRCh38
rs1033925266 Chr.16: 85633820-85633820 on Build GRCh38
rs145744466 Chr.16: 85633993-85633993 on Build GRCh38
rs768508867 Chr.16: 85634074-85634074 on Build GRCh38
rs774140695 Chr.16: 85634143-85634143 on Build GRCh38
rs765186908 Chr.16: 85633999-85633999 on Build GRCh38
rs140144665 Chr.16: 85633961-85633961 on Build GRCh38
rs573535667 Chr.16: 85633847-85633847 on Build GRCh38
rs374299844 Chr.16: 85634204-85634204 on Build GRCh38
rs771161124 Chr.16: 85634018-85634018 on Build GRCh38

View Full Product Details