Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 85707996-85708491 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C16orf74
Entrez Gene ID
404550
Gene Name
chromosome 16 open reading frame 74
Gene Aliases
-
UniGene
Hs.461655
Gene Chromosome Location
Chr.16: 85707517-85751083 on Build GRCh38
Transcripts
NM_206967.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6573501 c.361G>A p.G121R Chr.16: 85708031-85708031 on Build GRCh38
6195245 c.380C>T p.A127V Chr.16: 85708012-85708012 on Build GRCh38
7400320 c.364G>A p.E122K Chr.16: 85708028-85708028 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs11864007 Chr.16: 85708338-85708338 on Build GRCh38
rs372115862 Chr.16: 85708023-85708023 on Build GRCh38
rs769728815 Chr.16: 85708028-85708028 on Build GRCh38
rs895467862 Chr.16: 85708386-85708386 on Build GRCh38
rs565282225 Chr.16: 85708221-85708221 on Build GRCh38
rs923314901 Chr.16: 85708124-85708124 on Build GRCh38
rs75034934 Chr.16: 85708130-85708130 on Build GRCh38
rs753821950 Chr.16: 85708312-85708312 on Build GRCh38
rs547618356 Chr.16: 85708271-85708271 on Build GRCh38
rs561811899 Chr.16: 85708212-85708212 on Build GRCh38
rs750401569 Chr.16: 85708048-85708048 on Build GRCh38
rs11864004 Chr.16: 85708299-85708299 on Build GRCh38
rs757401554 Chr.16: 85708110-85708110 on Build GRCh38
rs1013209044 Chr.16: 85708257-85708257 on Build GRCh38
rs561301791 Chr.16: 85708276-85708276 on Build GRCh38
rs568179496 Chr.16: 85708463-85708463 on Build GRCh38
rs369828885 Chr.16: 85708058-85708058 on Build GRCh38
rs888388747 Chr.16: 85708417-85708417 on Build GRCh38
rs544953276 Chr.16: 85708205-85708205 on Build GRCh38
rs998957741 Chr.16: 85708038-85708038 on Build GRCh38
rs1009611191 Chr.16: 85708368-85708368 on Build GRCh38
rs910718573 Chr.16: 85708168-85708168 on Build GRCh38
rs895488500 Chr.16: 85708242-85708242 on Build GRCh38
rs774722781 Chr.16: 85708438-85708438 on Build GRCh38
rs1039377773 Chr.16: 85708096-85708096 on Build GRCh38
rs972185887 Chr.16: 85708328-85708328 on Build GRCh38
rs747440348 Chr.16: 85708293-85708293 on Build GRCh38
rs948399237 Chr.16: 85708392-85708392 on Build GRCh38
rs978878315 Chr.16: 85708361-85708361 on Build GRCh38
rs763000792 Chr.16: 85708037-85708037 on Build GRCh38
rs1044321571 Chr.16: 85708404-85708404 on Build GRCh38
rs910833366 Chr.16: 85708339-85708339 on Build GRCh38
rs960207171 Chr.16: 85708065-85708065 on Build GRCh38
rs749882953 Chr.16: 85708070-85708070 on Build GRCh38
rs868444847 Chr.16: 85708235-85708235 on Build GRCh38
rs369943295 Chr.16: 85708106-85708106 on Build GRCh38
rs547264614 Chr.16: 85708322-85708322 on Build GRCh38
rs755639697 Chr.16: 85708120-85708120 on Build GRCh38
rs201074227 Chr.16: 85708044-85708044 on Build GRCh38
rs771199535 Chr.16: 85708017-85708017 on Build GRCh38
rs758969667 Chr.16: 85708097-85708097 on Build GRCh38
rs781298367 Chr.16: 85708114-85708114 on Build GRCh38
rs763225324 Chr.16: 85708409-85708409 on Build GRCh38
rs754632559 Chr.16: 85708226-85708226 on Build GRCh38
rs1012850580 Chr.16: 85708068-85708068 on Build GRCh38
rs1005491196 Chr.16: 85708419-85708419 on Build GRCh38
rs993987130 Chr.16: 85708439-85708439 on Build GRCh38
rs778185686 Chr.16: 85708290-85708290 on Build GRCh38
rs954773087 Chr.16: 85708335-85708335 on Build GRCh38
rs73255141 Chr.16: 85708109-85708109 on Build GRCh38
rs565844183 Chr.16: 85708372-85708372 on Build GRCh38
rs1015406356 Chr.16: 85708423-85708423 on Build GRCh38
rs11860727 Chr.16: 85708424-85708424 on Build GRCh38
rs973963781 Chr.16: 85708449-85708449 on Build GRCh38
rs903927658 Chr.16: 85708379-85708379 on Build GRCh38
rs968206421 Chr.16: 85708272-85708272 on Build GRCh38
rs150985095 Chr.16: 85708383-85708383 on Build GRCh38
rs200329146 Chr.16: 85708063-85708063 on Build GRCh38
rs947541785 Chr.16: 85708348-85708348 on Build GRCh38
rs914060090 Chr.16: 85708036-85708038 on Build GRCh38
rs1005899399 Chr.16: 85708125-85708125 on Build GRCh38
rs979685258 Chr.16: 85708066-85708066 on Build GRCh38
rs1030449864 Chr.16: 85708446-85708446 on Build GRCh38
rs143484567 Chr.16: 85708362-85708362 on Build GRCh38
rs374154054 Chr.16: 85708228-85708228 on Build GRCh38
rs941168209 Chr.16: 85708344-85708344 on Build GRCh38
rs905102506 Chr.16: 85708144-85708144 on Build GRCh38
rs755611325 Chr.16: 85708072-85708072 on Build GRCh38
rs11865566 Chr.16: 85708331-85708331 on Build GRCh38
rs775308939 Chr.16: 85708029-85708029 on Build GRCh38
rs576366642 Chr.16: 85708204-85708204 on Build GRCh38
rs1029349929 Chr.16: 85708406-85708406 on Build GRCh38
rs531654974 Chr.16: 85708071-85708071 on Build GRCh38
rs11860689 Chr.16: 85708320-85708320 on Build GRCh38
rs765726101 Chr.16: 85708079-85708079 on Build GRCh38
rs539585471 Chr.16: 85708241-85708241 on Build GRCh38
rs905135248 Chr.16: 85708350-85708350 on Build GRCh38
rs763953882 Chr.16: 85708040-85708040 on Build GRCh38
rs902958172 Chr.16: 85708223-85708223 on Build GRCh38
rs140809174 Chr.16: 85708448-85708448 on Build GRCh38
rs769888495 Chr.16: 85708364-85708364 on Build GRCh38
rs543735493 Chr.16: 85708189-85708189 on Build GRCh38
rs753113028 Chr.16: 85708092-85708092 on Build GRCh38
rs775481116 Chr.16: 85708382-85708382 on Build GRCh38
rs367980585 Chr.16: 85708022-85708022 on Build GRCh38
rs778096568 Chr.16: 85708104-85708104 on Build GRCh38
rs1050640807 Chr.16: 85708145-85708145 on Build GRCh38

View Full Product Details