Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 90032313-90032817 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAS8
Entrez Gene ID
2622
Gene Name
growth arrest specific 8
Gene Aliases
CILD33, DRC4, GAS11
UniGene
Hs.431792
Gene Chromosome Location
Chr.16: 90019629-90044971 on Build GRCh38
Transcripts
NM_001286205.1 NM_001286208.1 NM_001286209.1 NM_001481.2 XM_005256304.4 XM_005256309.4 XM_006721175.3 XM_011522990.2 XM_011522991.1 XM_011522992.2 XM_017023122.1 XM_017023123.1 XM_017023124.1 XM_017023125.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1196858 c.322G>A p.E108K Chr.16: 90032751-90032751 on Build GRCh38
6669517 c.368T>A p.V123D Chr.16: 90032797-90032797 on Build GRCh38
4390128 c.376C>G p.L126V Chr.16: 90032805-90032805 on Build GRCh38
6423963 c.316C>A p.L106I Chr.16: 90032745-90032745 on Build GRCh38
6870042 c.289-1G>T p.? Chr.16: 90032717-90032717 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs202207673 Chr.16: 90032701-90032701 on Build GRCh38
rs554014349 Chr.16: 90032444-90032444 on Build GRCh38
rs543090227 Chr.16: 90032538-90032538 on Build GRCh38
rs541353020 Chr.16: 90032580-90032580 on Build GRCh38
rs764466712 Chr.16: 90032668-90032668 on Build GRCh38
rs568324140 Chr.16: 90032383-90032383 on Build GRCh38
rs183146531 Chr.16: 90032458-90032458 on Build GRCh38
rs772149907 Chr.16: 90032720-90032720 on Build GRCh38
rs570899099 Chr.16: 90032661-90032661 on Build GRCh38
rs1030552940 Chr.16: 90032481-90032481 on Build GRCh38
rs561732411 Chr.16: 90032576-90032576 on Build GRCh38
rs375388215 Chr.16: 90032714-90032714 on Build GRCh38
rs750746156 Chr.16: 90032698-90032698 on Build GRCh38
rs200708560 Chr.16: 90032751-90032751 on Build GRCh38
rs376510125 Chr.16: 90032344-90032344 on Build GRCh38
rs1006295928 Chr.16: 90032358-90032358 on Build GRCh38
rs773484225 Chr.16: 90032761-90032761 on Build GRCh38
rs889358165 Chr.16: 90032766-90032766 on Build GRCh38
rs373212112 Chr.16: 90032696-90032696 on Build GRCh38
rs958207667 Chr.16: 90032659-90032659 on Build GRCh38
rs556513441 Chr.16: 90032336-90032336 on Build GRCh38
rs572319267 Chr.16: 90032447-90032447 on Build GRCh38
rs756865971 Chr.16: 90032788-90032788 on Build GRCh38
rs948003943 Chr.16: 90032681-90032681 on Build GRCh38
rs528868051 Chr.16: 90032579-90032579 on Build GRCh38
rs978264882 Chr.16: 90032643-90032643 on Build GRCh38
rs906283725 Chr.16: 90032390-90032390 on Build GRCh38
rs776712411 Chr.16: 90032678-90032678 on Build GRCh38
rs4354938 Chr.16: 90032420-90032420 on Build GRCh38
rs763539642 Chr.16: 90032772-90032772 on Build GRCh38
rs1007310494 Chr.16: 90032597-90032597 on Build GRCh38
rs753348616 Chr.16: 90032781-90032781 on Build GRCh38
rs997933384 Chr.16: 90032431-90032431 on Build GRCh38
rs899070289 Chr.16: 90032449-90032449 on Build GRCh38
rs763242945 Chr.16: 90032764-90032764 on Build GRCh38
rs916839032 Chr.16: 90032662-90032662 on Build GRCh38
rs776961106 Chr.16: 90032739-90032739 on Build GRCh38
rs531844309 Chr.16: 90032693-90032693 on Build GRCh38
rs745964862 Chr.16: 90032768-90032768 on Build GRCh38
rs559670357 Chr.16: 90032587-90032587 on Build GRCh38
rs1021828910 Chr.16: 90032541-90032541 on Build GRCh38
rs199635939 Chr.16: 90032606-90032607 on Build GRCh38
rs116537058 Chr.16: 90032706-90032706 on Build GRCh38
rs867843979 Chr.16: 90032618-90032618 on Build GRCh38
rs780128065 Chr.16: 90032722-90032722 on Build GRCh38
rs747151390 Chr.16: 90032725-90032725 on Build GRCh38
rs778571857 Chr.16: 90032791-90032791 on Build GRCh38
rs376239328 Chr.16: 90032715-90032715 on Build GRCh38
rs889744808 Chr.16: 90032530-90032530 on Build GRCh38
rs576084293 Chr.16: 90032489-90032489 on Build GRCh38
rs539830776 Chr.16: 90032488-90032488 on Build GRCh38
rs766940589 Chr.16: 90032767-90032767 on Build GRCh38
rs770015007 Chr.16: 90032747-90032747 on Build GRCh38
rs934986179 Chr.16: 90032648-90032648 on Build GRCh38
rs140344685 Chr.16: 90032745-90032745 on Build GRCh38
rs760230699 Chr.16: 90032770-90032770 on Build GRCh38
rs940528126 Chr.16: 90032409-90032409 on Build GRCh38
rs533190346 Chr.16: 90032626-90032626 on Build GRCh38
rs1019685354 Chr.16: 90032625-90032625 on Build GRCh38
rs941872201 Chr.16: 90032707-90032707 on Build GRCh38
rs752214524 Chr.16: 90032705-90032705 on Build GRCh38
rs766801714 Chr.16: 90032703-90032703 on Build GRCh38
rs549962625 Chr.16: 90032695-90032695 on Build GRCh38
rs768586092 Chr.16: 90032734-90032734 on Build GRCh38
rs371791728 Chr.16: 90032333-90032333 on Build GRCh38
rs867036289 Chr.16: 90032384-90032384 on Build GRCh38
rs887850603 Chr.16: 90032339-90032339 on Build GRCh38
rs147133929 Chr.16: 90032718-90032718 on Build GRCh38
rs4324129 Chr.16: 90032653-90032653 on Build GRCh38
rs947731768 Chr.16: 90032378-90032378 on Build GRCh38

View Full Product Details