Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 3636306-3636803 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTNS
Entrez Gene ID
1497
Gene Name
cystinosin, lysosomal cystine transporter
Gene Aliases
CTNS-LSB, PQLC4
UniGene
Hs.187667
Gene Chromosome Location
Chr.17: 3636391-3663103 on Build GRCh38
Transcripts
NM_001031681.2 NM_004937.2 XM_005256485.2 XM_006721463.2 XM_006721464.2 XM_011523691.2 XM_011523692.2 XM_017024254.1 XM_017024255.1 XM_017024256.1 XM_017024257.1 XM_017024258.1 NM_013276.2

Gene Symbol
SHPK
Entrez Gene ID
23729
Gene Name
sedoheptulokinase
Gene Aliases
CARKL, SHK
UniGene
Hs.579217
Gene Chromosome Location
Chr.17: 3608262-3636322 on Build GRCh38
Transcripts
NM_001031681.2 NM_004937.2 XM_005256485.2 XM_006721463.2 XM_006721464.2 XM_011523691.2 XM_011523692.2 XM_017024254.1 XM_017024255.1 XM_017024256.1 XM_017024257.1 XM_017024258.1 NM_013276.2

SNP Detail

SNP ID Chromosome Location
rs111541274 Chr.17: 3636333-3636333 on Build GRCh38
rs918689543 Chr.17: 3636777-3636777 on Build GRCh38
rs955726514 Chr.17: 3636486-3636486 on Build GRCh38
rs1044928126 Chr.17: 3636615-3636615 on Build GRCh38
rs1048350297 Chr.17: 3636593-3636593 on Build GRCh38
rs35813652 Chr.17: 3636440-3636440 on Build GRCh38
rs573729122 Chr.17: 3636606-3636606 on Build GRCh38
rs886052860 Chr.17: 3636632-3636632 on Build GRCh38
rs550915875 Chr.17: 3636464-3636464 on Build GRCh38
rs550669452 Chr.17: 3636419-3636419 on Build GRCh38
rs953186893 Chr.17: 3636679-3636679 on Build GRCh38
rs938786620 Chr.17: 3636549-3636549 on Build GRCh38
rs138913261 Chr.17: 3636427-3636427 on Build GRCh38
rs17707869 Chr.17: 3636748-3636748 on Build GRCh38
rs1031417953 Chr.17: 3636622-3636622 on Build GRCh38
rs965934495 Chr.17: 3636706-3636706 on Build GRCh38
rs961719517 Chr.17: 3636521-3636521 on Build GRCh38
rs111977802 Chr.17: 3636541-3636541 on Build GRCh38
rs906377353 Chr.17: 3636623-3636623 on Build GRCh38
rs373260107 Chr.17: 3636585-3636585 on Build GRCh38
rs1011254470 Chr.17: 3636633-3636633 on Build GRCh38
rs986017083 Chr.17: 3636680-3636680 on Build GRCh38
rs911362538 Chr.17: 3636699-3636699 on Build GRCh38
rs920063266 Chr.17: 3636368-3636368 on Build GRCh38
rs902538687 Chr.17: 3636424-3636424 on Build GRCh38
rs1012120282 Chr.17: 3636484-3636484 on Build GRCh38
rs887711612 Chr.17: 3636601-3636601 on Build GRCh38
rs961668468 Chr.17: 3636663-3636663 on Build GRCh38
rs551536848 Chr.17: 3636376-3636376 on Build GRCh38
rs952992768 Chr.17: 3636532-3636532 on Build GRCh38
rs944277850 Chr.17: 3636553-3636553 on Build GRCh38
rs534946329 Chr.17: 3636552-3636552 on Build GRCh38
rs973116962 Chr.17: 3636529-3636529 on Build GRCh38
rs994546048 Chr.17: 3636672-3636672 on Build GRCh38
rs1050394310 Chr.17: 3636387-3636387 on Build GRCh38
rs960225050 Chr.17: 3636762-3636762 on Build GRCh38
rs981834725 Chr.17: 3636332-3636332 on Build GRCh38
rs577929057 Chr.17: 3636575-3636575 on Build GRCh38
rs530371788 Chr.17: 3636564-3636564 on Build GRCh38
rs920347235 Chr.17: 3636530-3636530 on Build GRCh38
rs112140949 Chr.17: 3636426-3636426 on Build GRCh38
rs893599733 Chr.17: 3636449-3636449 on Build GRCh38
rs980329610 Chr.17: 3636739-3636739 on Build GRCh38
rs973167802 Chr.17: 3636358-3636358 on Build GRCh38
rs1012479993 Chr.17: 3636610-3636610 on Build GRCh38
rs567603534 Chr.17: 3636422-3636422 on Build GRCh38
rs1014792738 Chr.17: 3636719-3636719 on Build GRCh38
rs1031783359 Chr.17: 3636480-3636480 on Build GRCh38
rs886052861 Chr.17: 3636697-3636697 on Build GRCh38
rs931318125 Chr.17: 3636589-3636589 on Build GRCh38
rs990441311 Chr.17: 3636492-3636492 on Build GRCh38
rs1014574967 Chr.17: 3636518-3636518 on Build GRCh38
rs886052862 Chr.17: 3636699-3636699 on Build GRCh38
rs893924611 Chr.17: 3636608-3636608 on Build GRCh38
rs931508396 Chr.17: 3636380-3636380 on Build GRCh38
rs777638404 Chr.17: 3636657-3636657 on Build GRCh38
rs924233349 Chr.17: 3636750-3636750 on Build GRCh38
rs970263189 Chr.17: 3636500-3636500 on Build GRCh38
rs537110680 Chr.17: 3636416-3636416 on Build GRCh38
rs970315813 Chr.17: 3636731-3636731 on Build GRCh38
rs1048957677 Chr.17: 3636383-3636383 on Build GRCh38
rs911519085 Chr.17: 3636403-3636403 on Build GRCh38
rs1023965026 Chr.17: 3636488-3636488 on Build GRCh38
rs887657778 Chr.17: 3636418-3636418 on Build GRCh38
rs559794013 Chr.17: 3636331-3636331 on Build GRCh38
rs940445408 Chr.17: 3636355-3636355 on Build GRCh38
rs997257719 Chr.17: 3636438-3636438 on Build GRCh38
rs571526276 Chr.17: 3636383-3636383 on Build GRCh38
rs542251529 Chr.17: 3636678-3636678 on Build GRCh38
rs997709445 Chr.17: 3636616-3636616 on Build GRCh38
rs1053794837 Chr.17: 3636607-3636607 on Build GRCh38
rs1003434728 Chr.17: 3636632-3636632 on Build GRCh38
rs1003602215 Chr.17: 3636514-3636514 on Build GRCh38
rs970697822 Chr.17: 3636496-3636496 on Build GRCh38
rs751822425 Chr.17: 3636768-3636768 on Build GRCh38
rs373781177 Chr.17: 3636444-3636444 on Build GRCh38
rs1014858355 Chr.17: 3636641-3636641 on Build GRCh38
rs35387702 Chr.17: 3636458-3636459 on Build GRCh38
rs11657606 Chr.17: 3636603-3636603 on Build GRCh38

View Full Product Details