Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 3656569-3657062 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTNS
Entrez Gene ID
1497
Gene Name
cystinosin, lysosomal cystine transporter
Gene Aliases
CTNS-LSB, PQLC4
UniGene
Hs.187667
Gene Chromosome Location
Chr.17: 3636391-3663103 on Build GRCh38
Transcripts
NM_001031681.2 NM_004937.2 XM_005256485.2 XM_006721463.2 XM_006721464.2 XM_011523691.2 XM_011523692.2 XM_017024254.1 XM_017024255.1 XM_017024256.1 XM_017024257.1 XM_017024258.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4518316 c.599_600CC>TT p.P200L Chr.17: 3656713-3656714 on Build GRCh38
1202619 c.635C>T p.A212V Chr.17: 3656749-3656749 on Build GRCh38
6637661 c.615C>T p.D205D Chr.17: 3656729-3656729 on Build GRCh38
6880662 c.633C>T p.H211H Chr.17: 3656747-3656747 on Build GRCh38
4675433 c.589G>A p.G197R Chr.17: 3656703-3656703 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373745850 Chr.17: 3656790-3656790 on Build GRCh38
rs759165887 Chr.17: 3656840-3656840 on Build GRCh38
rs770507590 Chr.17: 3656838-3656838 on Build GRCh38
rs201042876 Chr.17: 3656607-3656607 on Build GRCh38
rs1015164055 Chr.17: 3656976-3656976 on Build GRCh38
rs919667517 Chr.17: 3656655-3656655 on Build GRCh38
rs772467714 Chr.17: 3656812-3656812 on Build GRCh38
rs764063960 Chr.17: 3656656-3656656 on Build GRCh38
rs749317721 Chr.17: 3656796-3656796 on Build GRCh38
rs757360314 Chr.17: 3656802-3656802 on Build GRCh38
rs755297309 Chr.17: 3656782-3656782 on Build GRCh38
rs749999133 Chr.17: 3656684-3656684 on Build GRCh38
rs752833776 Chr.17: 3656809-3656810 on Build GRCh38
rs1023162876 Chr.17: 3656608-3656608 on Build GRCh38
rs140572605 Chr.17: 3656730-3656730 on Build GRCh38
rs756137690 Chr.17: 3656729-3656729 on Build GRCh38
rs769144872 Chr.17: 3656621-3656621 on Build GRCh38
rs764453407 Chr.17: 3656847-3656847 on Build GRCh38
rs940644999 Chr.17: 3656743-3656743 on Build GRCh38
rs762581003 Chr.17: 3656765-3656765 on Build GRCh38
rs746112494 Chr.17: 3656808-3656808 on Build GRCh38
rs111256750 Chr.17: 3656590-3656590 on Build GRCh38
rs973369089 Chr.17: 3657041-3657041 on Build GRCh38
rs893207601 Chr.17: 3656759-3656760 on Build GRCh38
rs761088992 Chr.17: 3656846-3656846 on Build GRCh38
rs764861416 Chr.17: 3656676-3656676 on Build GRCh38
rs761366500 Chr.17: 3656672-3656672 on Build GRCh38
rs369084366 Chr.17: 3656871-3656871 on Build GRCh38
rs753058855 Chr.17: 3656845-3656845 on Build GRCh38
rs772591288 Chr.17: 3656762-3656762 on Build GRCh38
rs547710556 Chr.17: 3656954-3656954 on Build GRCh38
rs372250201 Chr.17: 3656650-3656650 on Build GRCh38
rs373806413 Chr.17: 3656974-3656974 on Build GRCh38
rs912410408 Chr.17: 3656717-3656717 on Build GRCh38
rs113994208 Chr.17: 3656727-3656727 on Build GRCh38
rs771057130 Chr.17: 3656738-3656738 on Build GRCh38
rs760843675 Chr.17: 3656756-3656756 on Build GRCh38
rs770145986 Chr.17: 3656782-3656782 on Build GRCh38
rs762304420 Chr.17: 3656837-3656837 on Build GRCh38
rs201759090 Chr.17: 3656761-3656761 on Build GRCh38
rs200273011 Chr.17: 3656723-3656723 on Build GRCh38
rs773212644 Chr.17: 3656836-3656836 on Build GRCh38
rs201287562 Chr.17: 3656625-3656625 on Build GRCh38
rs1800531 Chr.17: 3656804-3656804 on Build GRCh38
rs970949314 Chr.17: 3656967-3656967 on Build GRCh38
rs113994207 Chr.17: 3656703-3656703 on Build GRCh38
rs912356958 Chr.17: 3656940-3656940 on Build GRCh38
rs138270433 Chr.17: 3656775-3656775 on Build GRCh38
rs372542386 Chr.17: 3656624-3656624 on Build GRCh38
rs765259334 Chr.17: 3656750-3656750 on Build GRCh38
rs767202021 Chr.17: 3656841-3656841 on Build GRCh38
rs369227591 Chr.17: 3656774-3656774 on Build GRCh38
rs1057516993 Chr.17: 3656760-3656761 on Build GRCh38
rs1057516723 Chr.17: 3656797-3656797 on Build GRCh38
rs577044481 Chr.17: 3656714-3656714 on Build GRCh38
rs531995684 Chr.17: 3656742-3656742 on Build GRCh38
rs775142361 Chr.17: 3656621-3656622 on Build GRCh38
rs775118383 Chr.17: 3656638-3656638 on Build GRCh38
rs894209737 Chr.17: 3656892-3656892 on Build GRCh38
rs962293523 Chr.17: 3656980-3656980 on Build GRCh38
rs1020349553 Chr.17: 3656628-3656628 on Build GRCh38
rs786204423 Chr.17: 3656728-3656730 on Build GRCh38
rs780255290 Chr.17: 3656610-3656610 on Build GRCh38
rs773945181 Chr.17: 3656839-3656839 on Build GRCh38
rs749428707 Chr.17: 3656735-3656735 on Build GRCh38
rs201858846 Chr.17: 3656634-3656634 on Build GRCh38
rs201788681 Chr.17: 3656816-3656816 on Build GRCh38
rs527864684 Chr.17: 3656952-3656952 on Build GRCh38
rs878879922 Chr.17: 3657009-3657009 on Build GRCh38
rs757955245 Chr.17: 3656702-3656702 on Build GRCh38
rs760256854 Chr.17: 3656725-3656727 on Build GRCh38
rs374751105 Chr.17: 3656591-3656591 on Build GRCh38
rs776712980 Chr.17: 3656660-3656660 on Build GRCh38
rs778414542 Chr.17: 3656795-3656795 on Build GRCh38
rs370832972 Chr.17: 3656817-3656817 on Build GRCh38
rs768600603 Chr.17: 3656748-3656748 on Build GRCh38
rs150428029 Chr.17: 3656747-3656747 on Build GRCh38
rs767891544 Chr.17: 3656788-3656788 on Build GRCh38
rs1003086406 Chr.17: 3656798-3656798 on Build GRCh38
rs1054489127 Chr.17: 3656753-3656753 on Build GRCh38
rs763462965 Chr.17: 3656812-3656812 on Build GRCh38
rs776658046 Chr.17: 3656749-3656749 on Build GRCh38
rs149915021 Chr.17: 3656715-3656715 on Build GRCh38
rs200170842 Chr.17: 3656726-3656726 on Build GRCh38
rs773158063 Chr.17: 3656872-3656872 on Build GRCh38
rs142860397 Chr.17: 3656791-3656791 on Build GRCh38
rs977986376 Chr.17: 3657022-3657022 on Build GRCh38
rs369956858 Chr.17: 3656635-3656635 on Build GRCh38
rs961335018 Chr.17: 3656647-3656647 on Build GRCh38
rs1041585970 Chr.17: 3656831-3656831 on Build GRCh38
rs769870975 Chr.17: 3656632-3656632 on Build GRCh38

View Full Product Details