Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 4163841-4164389 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKFY1
Entrez Gene ID
51479
Gene Name
ankyrin repeat and FYVE domain containing 1
Gene Aliases
ANKHZN, BTBD23, ZFYVE14
UniGene
Hs.696087
Gene Chromosome Location
Chr.17: 4163371-4264026 on Build GRCh38
Transcripts
NM_001257999.1 NM_016376.3 NR_047571.1 XM_005256679.3 XM_011523926.1 XM_011523928.2 XM_011523933.1 XM_017024733.1

SNP Detail

SNP ID Chromosome Location
rs928674615 Chr.17: 4164015-4164015 on Build GRCh38
rs1027403276 Chr.17: 4164115-4164115 on Build GRCh38
rs966331832 Chr.17: 4163876-4163876 on Build GRCh38
rs141700516 Chr.17: 4164306-4164306 on Build GRCh38
rs924871840 Chr.17: 4163905-4163905 on Build GRCh38
rs975955839 Chr.17: 4164173-4164173 on Build GRCh38
rs761896669 Chr.17: 4164016-4164016 on Build GRCh38
rs755234838 Chr.17: 4164212-4164212 on Build GRCh38
rs1043592 Chr.17: 4164021-4164021 on Build GRCh38
rs999188872 Chr.17: 4164176-4164176 on Build GRCh38
rs763983279 Chr.17: 4164019-4164019 on Build GRCh38
rs1007730667 Chr.17: 4164130-4164130 on Build GRCh38
rs551848121 Chr.17: 4164201-4164201 on Build GRCh38
rs536408605 Chr.17: 4164209-4164213 on Build GRCh38
rs767896244 Chr.17: 4164247-4164247 on Build GRCh38
rs897976464 Chr.17: 4164103-4164103 on Build GRCh38
rs940044444 Chr.17: 4164262-4164262 on Build GRCh38
rs186934018 Chr.17: 4164254-4164254 on Build GRCh38
rs970193837 Chr.17: 4164240-4164240 on Build GRCh38
rs955863945 Chr.17: 4164192-4164192 on Build GRCh38
rs916087790 Chr.17: 4164227-4164227 on Build GRCh38
rs957663152 Chr.17: 4164190-4164190 on Build GRCh38
rs889583072 Chr.17: 4164106-4164106 on Build GRCh38
rs1032397752 Chr.17: 4164181-4164181 on Build GRCh38
rs1043587 Chr.17: 4164258-4164258 on Build GRCh38
rs529541785 Chr.17: 4163995-4163995 on Build GRCh38
rs560755729 Chr.17: 4163937-4163937 on Build GRCh38
rs975986750 Chr.17: 4163931-4163931 on Build GRCh38
rs571591643 Chr.17: 4164202-4164202 on Build GRCh38
rs1056750894 Chr.17: 4164092-4164092 on Build GRCh38
rs916056472 Chr.17: 4163943-4163943 on Build GRCh38
rs966391422 Chr.17: 4164169-4164169 on Build GRCh38
rs1019364714 Chr.17: 4164153-4164153 on Build GRCh38
rs761460096 Chr.17: 4164051-4164051 on Build GRCh38
rs779400643 Chr.17: 4164322-4164322 on Build GRCh38
rs1037048155 Chr.17: 4164297-4164297 on Build GRCh38
rs990769048 Chr.17: 4164199-4164199 on Build GRCh38
rs750546498 Chr.17: 4163891-4163891 on Build GRCh38
rs1045760107 Chr.17: 4164001-4164001 on Build GRCh38
rs760222511 Chr.17: 4163879-4163879 on Build GRCh38
rs147959179 Chr.17: 4164062-4164062 on Build GRCh38
rs563958086 Chr.17: 4163941-4163941 on Build GRCh38
rs547829472 Chr.17: 4163901-4163904 on Build GRCh38
rs995801606 Chr.17: 4164091-4164091 on Build GRCh38
rs8261 Chr.17: 4164121-4164121 on Build GRCh38
rs767483828 Chr.17: 4163875-4163875 on Build GRCh38
rs753089391 Chr.17: 4163988-4163988 on Build GRCh38
rs936197478 Chr.17: 4163916-4163916 on Build GRCh38
rs939357522 Chr.17: 4164287-4164287 on Build GRCh38
rs375536743 Chr.17: 4163965-4163965 on Build GRCh38
rs113367578 Chr.17: 4164018-4164018 on Build GRCh38
rs1049597299 Chr.17: 4164100-4164100 on Build GRCh38
rs929548069 Chr.17: 4164358-4164358 on Build GRCh38

View Full Product Details