Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 4166985-4167494 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKFY1
Entrez Gene ID
51479
Gene Name
ankyrin repeat and FYVE domain containing 1
Gene Aliases
ANKHZN, BTBD23, ZFYVE14
UniGene
Hs.696087
Gene Chromosome Location
Chr.17: 4163371-4264026 on Build GRCh38
Transcripts
NM_001257999.1 NM_016376.3 NR_047571.1 XM_005256679.3 XM_011523926.1 XM_011523928.2 XM_011523933.1 XM_017024733.1

SNP Detail

SNP ID Chromosome Location
rs1027946989 Chr.17: 4167099-4167099 on Build GRCh38
rs959452661 Chr.17: 4167089-4167089 on Build GRCh38
rs544477349 Chr.17: 4167316-4167316 on Build GRCh38
rs957392290 Chr.17: 4167291-4167291 on Build GRCh38
rs1021606801 Chr.17: 4167025-4167025 on Build GRCh38
rs996559023 Chr.17: 4167450-4167450 on Build GRCh38
rs1003948029 Chr.17: 4167078-4167078 on Build GRCh38
rs992620144 Chr.17: 4167090-4167090 on Build GRCh38
rs904133959 Chr.17: 4167389-4167389 on Build GRCh38
rs755326378 Chr.17: 4167387-4167387 on Build GRCh38
rs575614227 Chr.17: 4167293-4167293 on Build GRCh38
rs534352977 Chr.17: 4167135-4167135 on Build GRCh38
rs1038119796 Chr.17: 4167385-4167385 on Build GRCh38
rs898199170 Chr.17: 4167429-4167429 on Build GRCh38
rs140175390 Chr.17: 4167193-4167193 on Build GRCh38
rs546237408 Chr.17: 4167259-4167259 on Build GRCh38
rs556571274 Chr.17: 4167283-4167283 on Build GRCh38
rs890807442 Chr.17: 4167452-4167452 on Build GRCh38
rs996955072 Chr.17: 4167093-4167093 on Build GRCh38
rs145691891 Chr.17: 4167368-4167368 on Build GRCh38
rs1038555791 Chr.17: 4167080-4167080 on Build GRCh38
rs950252519 Chr.17: 4167356-4167356 on Build GRCh38
rs929293107 Chr.17: 4167101-4167101 on Build GRCh38
rs887286479 Chr.17: 4167070-4167070 on Build GRCh38
rs540350640 Chr.17: 4167464-4167464 on Build GRCh38
rs538119594 Chr.17: 4167076-4167076 on Build GRCh38
rs1008078735 Chr.17: 4167459-4167459 on Build GRCh38
rs917957565 Chr.17: 4167097-4167097 on Build GRCh38
rs558195368 Chr.17: 4167126-4167126 on Build GRCh38
rs1038902303 Chr.17: 4167213-4167213 on Build GRCh38
rs553956043 Chr.17: 4167138-4167138 on Build GRCh38
rs766918026 Chr.17: 4167374-4167374 on Build GRCh38
rs977522377 Chr.17: 4167269-4167269 on Build GRCh38
rs750571988 Chr.17: 4167251-4167251 on Build GRCh38
rs991449350 Chr.17: 4167307-4167307 on Build GRCh38
rs915923468 Chr.17: 4167309-4167309 on Build GRCh38
rs1001034554 Chr.17: 4167451-4167451 on Build GRCh38
rs983409105 Chr.17: 4167167-4167167 on Build GRCh38
rs752435701 Chr.17: 4167064-4167065 on Build GRCh38
rs761898955 Chr.17: 4167057-4167057 on Build GRCh38
rs900296684 Chr.17: 4167223-4167223 on Build GRCh38
rs950283253 Chr.17: 4167009-4167009 on Build GRCh38
rs568518043 Chr.17: 4167102-4167102 on Build GRCh38
rs572270195 Chr.17: 4167317-4167317 on Build GRCh38
rs184642263 Chr.17: 4167081-4167081 on Build GRCh38
rs968217384 Chr.17: 4167036-4167036 on Build GRCh38
rs926066390 Chr.17: 4167286-4167286 on Build GRCh38
rs751886765 Chr.17: 4167375-4167375 on Build GRCh38
rs941897683 Chr.17: 4167194-4167194 on Build GRCh38
rs781539963 Chr.17: 4167153-4167153 on Build GRCh38

View Full Product Details