Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 4172361-4172856 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKFY1
Entrez Gene ID
51479
Gene Name
ankyrin repeat and FYVE domain containing 1
Gene Aliases
ANKHZN, BTBD23, ZFYVE14
UniGene
Hs.696087
Gene Chromosome Location
Chr.17: 4163371-4264026 on Build GRCh38
Transcripts
NM_001257999.1 NM_016376.3 NR_047571.1 XM_005256679.3 XM_011523926.1 XM_011523928.2 XM_011523933.1 XM_017024733.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5426072 c.3018-3C>T p.? Chr.17: 4172683-4172683 on Build GRCh38
5426073 c.3141-3C>T p.? Chr.17: 4172683-4172683 on Build GRCh38
3958392 c.3142+10A>T p.? Chr.17: 4172546-4172546 on Build GRCh38
6190919 c.3207C>T p.L1069L Chr.17: 4172614-4172614 on Build GRCh38
4066600 c.3140C>T p.T1047M Chr.17: 4172558-4172558 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs114884685 Chr.17: 4172710-4172710 on Build GRCh38
rs760506347 Chr.17: 4172541-4172541 on Build GRCh38
rs544244653 Chr.17: 4172797-4172797 on Build GRCh38
rs781738083 Chr.17: 4172524-4172524 on Build GRCh38
rs766174304 Chr.17: 4172596-4172596 on Build GRCh38
rs374496100 Chr.17: 4172604-4172604 on Build GRCh38
rs945597066 Chr.17: 4172481-4172481 on Build GRCh38
rs201949200 Chr.17: 4172528-4172528 on Build GRCh38
rs755265255 Chr.17: 4172523-4172523 on Build GRCh38
rs912891530 Chr.17: 4172792-4172792 on Build GRCh38
rs749362861 Chr.17: 4172533-4172533 on Build GRCh38
rs371345403 Chr.17: 4172535-4172535 on Build GRCh38
rs761599068 Chr.17: 4172569-4172569 on Build GRCh38
rs532708045 Chr.17: 4172613-4172613 on Build GRCh38
rs34771708 Chr.17: 4172656-4172657 on Build GRCh38
rs765148229 Chr.17: 4172685-4172685 on Build GRCh38
rs201982886 Chr.17: 4172557-4172557 on Build GRCh38
rs1038007300 Chr.17: 4172454-4172454 on Build GRCh38
rs191996603 Chr.17: 4172464-4172464 on Build GRCh38
rs750113523 Chr.17: 4172510-4172510 on Build GRCh38
rs756475807 Chr.17: 4172732-4172732 on Build GRCh38
rs778237541 Chr.17: 4172625-4172625 on Build GRCh38
rs534261049 Chr.17: 4172737-4172737 on Build GRCh38
rs932975665 Chr.17: 4172463-4172463 on Build GRCh38
rs1019067560 Chr.17: 4172503-4172503 on Build GRCh38
rs146538109 Chr.17: 4172730-4172730 on Build GRCh38
rs1037974647 Chr.17: 4172776-4172776 on Build GRCh38
rs368929290 Chr.17: 4172518-4172518 on Build GRCh38
rs746032914 Chr.17: 4172656-4172656 on Build GRCh38
rs377072239 Chr.17: 4172629-4172629 on Build GRCh38
rs904047020 Chr.17: 4172815-4172815 on Build GRCh38
rs750023237 Chr.17: 4172593-4172593 on Build GRCh38
rs983287729 Chr.17: 4172405-4172405 on Build GRCh38
rs919615286 Chr.17: 4172787-4172787 on Build GRCh38
rs1009387669 Chr.17: 4172477-4172477 on Build GRCh38
rs909014320 Chr.17: 4172422-4172422 on Build GRCh38
rs751215327 Chr.17: 4172598-4172598 on Build GRCh38
rs941810542 Chr.17: 4172427-4172427 on Build GRCh38
rs758057045 Chr.17: 4172513-4172513 on Build GRCh38
rs773261749 Chr.17: 4172694-4172694 on Build GRCh38
rs921617885 Chr.17: 4172451-4172451 on Build GRCh38
rs778240425 Chr.17: 4172508-4172508 on Build GRCh38
rs987105522 Chr.17: 4172789-4172789 on Build GRCh38
rs759620070 Chr.17: 4172576-4172576 on Build GRCh38
rs779849601 Chr.17: 4172516-4172516 on Build GRCh38
rs752775855 Chr.17: 4172796-4172796 on Build GRCh38
rs774590085 Chr.17: 4172653-4172653 on Build GRCh38
rs757970755 Chr.17: 4172594-4172594 on Build GRCh38
rs1013612898 Chr.17: 4172574-4172574 on Build GRCh38
rs767190788 Chr.17: 4172728-4172728 on Build GRCh38
rs1042624738 Chr.17: 4172810-4172810 on Build GRCh38
rs781353549 Chr.17: 4172606-4172606 on Build GRCh38
rs950621343 Chr.17: 4172607-4172607 on Build GRCh38
rs769927329 Chr.17: 4172529-4172529 on Build GRCh38
rs2305504 Chr.17: 4172415-4172415 on Build GRCh38
rs774537193 Chr.17: 4172538-4172538 on Build GRCh38
rs577310324 Chr.17: 4172814-4172814 on Build GRCh38
rs771197727 Chr.17: 4172537-4172537 on Build GRCh38
rs568481716 Chr.17: 4172734-4172734 on Build GRCh38
rs933031348 Chr.17: 4172757-4172757 on Build GRCh38
rs376122707 Chr.17: 4172566-4172566 on Build GRCh38
rs895193667 Chr.17: 4172571-4172571 on Build GRCh38
rs200094920 Chr.17: 4172671-4172671 on Build GRCh38
rs368077855 Chr.17: 4172723-4172723 on Build GRCh38
rs776460432 Chr.17: 4172455-4172455 on Build GRCh38
rs891381394 Chr.17: 4172467-4172467 on Build GRCh38
rs890989420 Chr.17: 4172474-4172474 on Build GRCh38
rs770970574 Chr.17: 4172630-4172630 on Build GRCh38
rs939784845 Chr.17: 4172745-4172745 on Build GRCh38
rs754516258 Chr.17: 4172727-4172727 on Build GRCh38
rs762582126 Chr.17: 4172702-4172702 on Build GRCh38
rs773095412 Chr.17: 4172530-4172530 on Build GRCh38
rs552448221 Chr.17: 4172616-4172616 on Build GRCh38
rs974518028 Chr.17: 4172439-4172439 on Build GRCh38
rs569134707 Chr.17: 4172679-4172679 on Build GRCh38
rs886769276 Chr.17: 4172443-4172443 on Build GRCh38
rs577136299 Chr.17: 4172805-4172810 on Build GRCh38
rs72829328 Chr.17: 4172403-4172403 on Build GRCh38

View Full Product Details