Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 4733428-4733931 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CXCL16
Entrez Gene ID
58191
Gene Name
C-X-C motif chemokine ligand 16
Gene Aliases
CXCLG16, SR-PSOX, SRPSOX
UniGene
Hs.745037
Gene Chromosome Location
Chr.17: 4733529-4739928 on Build GRCh38
Transcripts
NM_022059.3 NM_001001683.3 NM_001305000.1

Gene Symbol
MED11
Entrez Gene ID
400569
Gene Name
mediator complex subunit 11
Gene Aliases
HSPC296
UniGene
Hs.513885
Gene Chromosome Location
Chr.17: 4731408-4733610 on Build GRCh38
Transcripts
NM_022059.3 NM_001001683.3 NM_001305000.1

SNP Detail

SNP ID Chromosome Location
rs954286116 Chr.17: 4733643-4733643 on Build GRCh38
rs924075433 Chr.17: 4733731-4733731 on Build GRCh38
rs1042368968 Chr.17: 4733495-4733495 on Build GRCh38
rs947974457 Chr.17: 4733786-4733786 on Build GRCh38
rs936667723 Chr.17: 4733736-4733736 on Build GRCh38
rs148734426 Chr.17: 4733498-4733498 on Build GRCh38
rs893480088 Chr.17: 4733879-4733879 on Build GRCh38
rs537802107 Chr.17: 4733903-4733903 on Build GRCh38
rs527815974 Chr.17: 4733568-4733582 on Build GRCh38
rs546360023 Chr.17: 4733470-4733470 on Build GRCh38
rs1004749670 Chr.17: 4733866-4733866 on Build GRCh38
rs909888528 Chr.17: 4733621-4733621 on Build GRCh38
rs993623102 Chr.17: 4733459-4733459 on Build GRCh38
rs944077010 Chr.17: 4733679-4733679 on Build GRCh38
rs35185827 Chr.17: 4733696-4733697 on Build GRCh38
rs748509715 Chr.17: 4733673-4733673 on Build GRCh38
rs898953612 Chr.17: 4733885-4733885 on Build GRCh38
rs568018909 Chr.17: 4733535-4733535 on Build GRCh38
rs181252549 Chr.17: 4733846-4733846 on Build GRCh38
rs941561280 Chr.17: 4733628-4733628 on Build GRCh38
rs973115898 Chr.17: 4733633-4733633 on Build GRCh38
rs556147721 Chr.17: 4733843-4733843 on Build GRCh38
rs966588546 Chr.17: 4733537-4733537 on Build GRCh38
rs773451698 Chr.17: 4733775-4733775 on Build GRCh38
rs546158705 Chr.17: 4733488-4733488 on Build GRCh38
rs773889649 Chr.17: 4733847-4733847 on Build GRCh38
rs4790696 Chr.17: 4733721-4733721 on Build GRCh38
rs963360000 Chr.17: 4733622-4733622 on Build GRCh38
rs895108847 Chr.17: 4733551-4733551 on Build GRCh38
rs764905312 Chr.17: 4733722-4733722 on Build GRCh38
rs1014446640 Chr.17: 4733758-4733758 on Build GRCh38
rs759136074 Chr.17: 4733627-4733627 on Build GRCh38
rs912755037 Chr.17: 4733672-4733672 on Build GRCh38
rs1012025070 Chr.17: 4733573-4733575 on Build GRCh38
rs1051007 Chr.17: 4733518-4733518 on Build GRCh38
rs769130534 Chr.17: 4733620-4733620 on Build GRCh38
rs781694318 Chr.17: 4733661-4733661 on Build GRCh38
rs977223818 Chr.17: 4733564-4733564 on Build GRCh38
rs762625909 Chr.17: 4733797-4733797 on Build GRCh38
rs547656320 Chr.17: 4733577-4733577 on Build GRCh38
rs562539799 Chr.17: 4733497-4733497 on Build GRCh38
rs35936412 Chr.17: 4733751-4733752 on Build GRCh38
rs750603467 Chr.17: 4733486-4733486 on Build GRCh38
rs906316349 Chr.17: 4733845-4733845 on Build GRCh38
rs567842184 Chr.17: 4733465-4733465 on Build GRCh38
rs568672682 Chr.17: 4733887-4733887 on Build GRCh38
rs895030356 Chr.17: 4733743-4733743 on Build GRCh38
rs932638269 Chr.17: 4733608-4733608 on Build GRCh38
rs867681259 Chr.17: 4733543-4733543 on Build GRCh38
rs550119454 Chr.17: 4733680-4733680 on Build GRCh38
rs897289747 Chr.17: 4733724-4733724 on Build GRCh38
rs117048273 Chr.17: 4733536-4733536 on Build GRCh38
rs1053823268 Chr.17: 4733741-4733741 on Build GRCh38
rs1001903939 Chr.17: 4733481-4733481 on Build GRCh38
rs36056309 Chr.17: 4733761-4733762 on Build GRCh38
rs570660311 Chr.17: 4733701-4733701 on Build GRCh38
rs545585833 Chr.17: 4733643-4733644 on Build GRCh38
rs190833337 Chr.17: 4733469-4733469 on Build GRCh38
rs906251550 Chr.17: 4733595-4733595 on Build GRCh38
rs868644468 Chr.17: 4733464-4733464 on Build GRCh38
rs752154348 Chr.17: 4733784-4733784 on Build GRCh38
rs1004606929 Chr.17: 4733600-4733601 on Build GRCh38

View Full Product Details