Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 4739173-4739665 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CXCL16
Entrez Gene ID
58191
Gene Name
C-X-C motif chemokine ligand 16
Gene Aliases
CXCLG16, SR-PSOX, SRPSOX
UniGene
Hs.745037
Gene Chromosome Location
Chr.17: 4733529-4739928 on Build GRCh38
Transcripts
NM_001100812.1 NM_022059.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3680498 c.99C>G p.L33L Chr.17: 4739298-4739298 on Build GRCh38
1384206 c.63A>G p.G21G Chr.17: 4739334-4739334 on Build GRCh38
4832468 c.5C>G p.S2C Chr.17: 4739392-4739392 on Build GRCh38
4941713 c.125T>C p.L42P Chr.17: 4739272-4739272 on Build GRCh38
6406303 c.73C>T p.R25W Chr.17: 4739324-4739324 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201106268 Chr.17: 4739424-4739424 on Build GRCh38
rs1032075230 Chr.17: 4739571-4739571 on Build GRCh38
rs997275109 Chr.17: 4739610-4739610 on Build GRCh38
rs758513537 Chr.17: 4739238-4739238 on Build GRCh38
rs777653971 Chr.17: 4739265-4739265 on Build GRCh38
rs773694740 Chr.17: 4739295-4739295 on Build GRCh38
rs143565057 Chr.17: 4739272-4739272 on Build GRCh38
rs953383792 Chr.17: 4739407-4739407 on Build GRCh38
rs769785444 Chr.17: 4739247-4739247 on Build GRCh38
rs1011946952 Chr.17: 4739623-4739623 on Build GRCh38
rs777494621 Chr.17: 4739365-4739365 on Build GRCh38
rs112611943 Chr.17: 4739351-4739351 on Build GRCh38
rs75334328 Chr.17: 4739388-4739388 on Build GRCh38
rs752370 Chr.17: 4739519-4739519 on Build GRCh38
rs965747670 Chr.17: 4739606-4739606 on Build GRCh38
rs2304970 Chr.17: 4739492-4739492 on Build GRCh38
rs770707432 Chr.17: 4739379-4739379 on Build GRCh38
rs781048899 Chr.17: 4739239-4739239 on Build GRCh38
rs1008101337 Chr.17: 4739315-4739315 on Build GRCh38
rs769537190 Chr.17: 4739211-4739230 on Build GRCh38
rs759137776 Chr.17: 4739292-4739293 on Build GRCh38
rs996781096 Chr.17: 4739209-4739209 on Build GRCh38
rs570618745 Chr.17: 4739591-4739591 on Build GRCh38
rs773010780 Chr.17: 4739235-4739237 on Build GRCh38
rs887625969 Chr.17: 4739274-4739274 on Build GRCh38
rs759230148 Chr.17: 4739308-4739308 on Build GRCh38
rs749252832 Chr.17: 4739371-4739371 on Build GRCh38
rs985098815 Chr.17: 4739628-4739628 on Build GRCh38
rs773725607 Chr.17: 4739287-4739292 on Build GRCh38
rs766669724 Chr.17: 4739432-4739432 on Build GRCh38
rs796343274 Chr.17: 4739519-4739520 on Build GRCh38
rs1023602052 Chr.17: 4739640-4739640 on Build GRCh38
rs1036472288 Chr.17: 4739325-4739325 on Build GRCh38
rs1049330268 Chr.17: 4739211-4739211 on Build GRCh38
rs949472414 Chr.17: 4739477-4739477 on Build GRCh38
rs775981222 Chr.17: 4739317-4739317 on Build GRCh38
rs538625577 Chr.17: 4739334-4739334 on Build GRCh38
rs755152192 Chr.17: 4739440-4739440 on Build GRCh38
rs140340376 Chr.17: 4739441-4739441 on Build GRCh38
rs140866155 Chr.17: 4739335-4739335 on Build GRCh38
rs771824284 Chr.17: 4739394-4739394 on Build GRCh38
rs762907110 Chr.17: 4739429-4739429 on Build GRCh38
rs767249912 Chr.17: 4739422-4739422 on Build GRCh38
rs770902327 Chr.17: 4739280-4739281 on Build GRCh38
rs921599066 Chr.17: 4739405-4739405 on Build GRCh38
rs138561221 Chr.17: 4739352-4739352 on Build GRCh38
rs752371 Chr.17: 4739520-4739520 on Build GRCh38
rs189927576 Chr.17: 4739473-4739473 on Build GRCh38
rs892617474 Chr.17: 4739557-4739557 on Build GRCh38
rs898488721 Chr.17: 4739462-4739462 on Build GRCh38
rs953091282 Chr.17: 4739619-4739619 on Build GRCh38
rs890935163 Chr.17: 4739307-4739307 on Build GRCh38
rs1009772937 Chr.17: 4739559-4739559 on Build GRCh38
rs371823055 Chr.17: 4739423-4739423 on Build GRCh38
rs762738913 Chr.17: 4739243-4739244 on Build GRCh38
rs760688882 Chr.17: 4739580-4739580 on Build GRCh38
rs372669540 Chr.17: 4739243-4739243 on Build GRCh38
rs563812362 Chr.17: 4739607-4739607 on Build GRCh38
rs1045008914 Chr.17: 4739485-4739485 on Build GRCh38
rs778498335 Chr.17: 4739380-4739380 on Build GRCh38
rs547733615 Chr.17: 4739575-4739575 on Build GRCh38
rs770447699 Chr.17: 4739277-4739277 on Build GRCh38
rs771567992 Chr.17: 4739313-4739313 on Build GRCh38
rs753572649 Chr.17: 4739443-4739443 on Build GRCh38
rs761119055 Chr.17: 4739324-4739324 on Build GRCh38
rs757382005 Chr.17: 4739212-4739212 on Build GRCh38
rs574652255 Chr.17: 4739398-4739398 on Build GRCh38
rs770654013 Chr.17: 4739596-4739596 on Build GRCh38
rs751801027 Chr.17: 4739439-4739439 on Build GRCh38
rs561871654 Chr.17: 4739534-4739534 on Build GRCh38
rs527434288 Chr.17: 4739541-4739541 on Build GRCh38
rs1028635370 Chr.17: 4739616-4739616 on Build GRCh38
rs750406656 Chr.17: 4739340-4739340 on Build GRCh38
rs750682680 Chr.17: 4739236-4739236 on Build GRCh38
rs755877977 Chr.17: 4739364-4739364 on Build GRCh38
rs959026590 Chr.17: 4739601-4739601 on Build GRCh38
rs151229852 Chr.17: 4739332-4739332 on Build GRCh38
rs959060280 Chr.17: 4739354-4739354 on Build GRCh38
rs908589945 Chr.17: 4739420-4739420 on Build GRCh38
rs150143436 Chr.17: 4739342-4739342 on Build GRCh38
rs540529925 Chr.17: 4739402-4739402 on Build GRCh38
rs769168839 Chr.17: 4739412-4739412 on Build GRCh38
rs757278306 Chr.17: 4739448-4739448 on Build GRCh38
rs1020073048 Chr.17: 4739395-4739395 on Build GRCh38
rs752527033 Chr.17: 4739362-4739362 on Build GRCh38
rs779055969 Chr.17: 4739234-4739234 on Build GRCh38
rs2304971 Chr.17: 4739338-4739338 on Build GRCh38
rs940742897 Chr.17: 4739322-4739322 on Build GRCh38

View Full Product Details