Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 11636554-11637054 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAH9
Entrez Gene ID
1770
Gene Name
dynein axonemal heavy chain 9
Gene Aliases
DNAH17L, DNEL1, DYH9, Dnahc9, HL-20, HL20
UniGene
Hs.567259
Gene Chromosome Location
Chr.17: 11598431-11970168 on Build GRCh38
Transcripts
NM_001372.3 XM_011523703.1 XM_017024292.1 XM_017024294.1 XM_017024295.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1204163 c.1709T>G p.L570R Chr.17: 11636707-11636707 on Build GRCh38
6647014 c.1757G>A p.S586N Chr.17: 11636755-11636755 on Build GRCh38
4382850 c.1735G>C p.D579H Chr.17: 11636733-11636733 on Build GRCh38
6883130 c.1637T>G p.L546R Chr.17: 11636635-11636635 on Build GRCh38
4596448 c.1747A>G p.M583V Chr.17: 11636745-11636745 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753371504 Chr.17: 11636817-11636817 on Build GRCh38
rs187311754 Chr.17: 11636900-11636900 on Build GRCh38
rs1006486234 Chr.17: 11636731-11636731 on Build GRCh38
rs184410301 Chr.17: 11636956-11636956 on Build GRCh38
rs151215889 Chr.17: 11636599-11636600 on Build GRCh38
rs756984342 Chr.17: 11636660-11636660 on Build GRCh38
rs150122579 Chr.17: 11636702-11636702 on Build GRCh38
rs936073478 Chr.17: 11636717-11636717 on Build GRCh38
rs375961188 Chr.17: 11636774-11636774 on Build GRCh38
rs895395192 Chr.17: 11636984-11636984 on Build GRCh38
rs763145751 Chr.17: 11636691-11636691 on Build GRCh38
rs544899049 Chr.17: 11636740-11636740 on Build GRCh38
rs760648273 Chr.17: 11636813-11636813 on Build GRCh38
rs534432763 Chr.17: 11636745-11636745 on Build GRCh38
rs537727364 Chr.17: 11636719-11636719 on Build GRCh38
rs748628607 Chr.17: 11636672-11636672 on Build GRCh38
rs933074201 Chr.17: 11636929-11636929 on Build GRCh38
rs759374376 Chr.17: 11636708-11636708 on Build GRCh38
rs771834613 Chr.17: 11636801-11636801 on Build GRCh38
rs751953599 Chr.17: 11636751-11636751 on Build GRCh38
rs1034581753 Chr.17: 11636967-11636967 on Build GRCh38
rs972637786 Chr.17: 11636631-11636631 on Build GRCh38
rs575487404 Chr.17: 11636837-11636837 on Build GRCh38
rs192209809 Chr.17: 11636934-11636934 on Build GRCh38
rs767236602 Chr.17: 11636714-11636714 on Build GRCh38
rs775163392 Chr.17: 11636812-11636812 on Build GRCh38
rs766799921 Chr.17: 11636731-11636731 on Build GRCh38
rs1015922701 Chr.17: 11636965-11636965 on Build GRCh38
rs886080216 Chr.17: 11636707-11636707 on Build GRCh38
rs556895264 Chr.17: 11636819-11636819 on Build GRCh38
rs764005259 Chr.17: 11636814-11636814 on Build GRCh38
rs896834522 Chr.17: 11636904-11636904 on Build GRCh38
rs1007827000 Chr.17: 11636598-11636598 on Build GRCh38
rs371695921 Chr.17: 11636739-11636739 on Build GRCh38
rs372282496 Chr.17: 11636668-11636668 on Build GRCh38
rs201461518 Chr.17: 11636681-11636681 on Build GRCh38
rs913278372 Chr.17: 11637001-11637001 on Build GRCh38
rs752125479 Chr.17: 11636632-11636632 on Build GRCh38
rs35822016 Chr.17: 11636951-11636951 on Build GRCh38
rs151314858 Chr.17: 11636783-11636783 on Build GRCh38
rs61701166 Chr.17: 11636578-11636578 on Build GRCh38
rs990646114 Chr.17: 11636993-11636993 on Build GRCh38
rs8065057 Chr.17: 11636762-11636762 on Build GRCh38
rs1005731245 Chr.17: 11636866-11636866 on Build GRCh38
rs781749122 Chr.17: 11636671-11636671 on Build GRCh38
rs184123769 Chr.17: 11636869-11636869 on Build GRCh38
rs774101993 Chr.17: 11636706-11636706 on Build GRCh38
rs530072737 Chr.17: 11636653-11636653 on Build GRCh38
rs879234347 Chr.17: 11636865-11636865 on Build GRCh38
rs777181049 Chr.17: 11636642-11636642 on Build GRCh38
rs773437773 Chr.17: 11636586-11636586 on Build GRCh38
rs771917153 Chr.17: 11636673-11636673 on Build GRCh38
rs771119765 Chr.17: 11636793-11636793 on Build GRCh38
rs377051669 Chr.17: 11636677-11636677 on Build GRCh38
rs767530140 Chr.17: 11636622-11636622 on Build GRCh38
rs549055800 Chr.17: 11636964-11636964 on Build GRCh38
rs751410103 Chr.17: 11636611-11636611 on Build GRCh38
rs192901923 Chr.17: 11636800-11636800 on Build GRCh38
rs755599891 Chr.17: 11636633-11636633 on Build GRCh38
rs766101465 Chr.17: 11636600-11636600 on Build GRCh38
rs781443832 Chr.17: 11636763-11636763 on Build GRCh38
rs541885531 Chr.17: 11636958-11636958 on Build GRCh38
rs570676495 Chr.17: 11636730-11636730 on Build GRCh38
rs763602808 Chr.17: 11636722-11636722 on Build GRCh38
rs957195987 Chr.17: 11636991-11636991 on Build GRCh38
rs901636127 Chr.17: 11636918-11636918 on Build GRCh38
rs764953459 Chr.17: 11636909-11636909 on Build GRCh38
rs774506379 Chr.17: 11636796-11636796 on Build GRCh38
rs1038881753 Chr.17: 11636599-11636599 on Build GRCh38
rs761563082 Chr.17: 11636669-11636669 on Build GRCh38
rs754905788 Chr.17: 11636620-11636620 on Build GRCh38
rs563453410 Chr.17: 11636916-11636916 on Build GRCh38
rs140815859 Chr.17: 11636685-11636685 on Build GRCh38
rs190478356 Chr.17: 11636980-11636980 on Build GRCh38
rs866795534 Chr.17: 11636870-11636870 on Build GRCh38
rs552531254 Chr.17: 11636721-11636721 on Build GRCh38
rs773005835 Chr.17: 11636678-11636678 on Build GRCh38
rs146918340 Chr.17: 11636769-11636769 on Build GRCh38
rs374886127 Chr.17: 11636587-11636587 on Build GRCh38

View Full Product Details