Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 29929749-29930242 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EFCAB5
Entrez Gene ID
374786
Gene Name
EF-hand calcium binding domain 5
Gene Aliases
-
UniGene
Hs.662411
Gene Chromosome Location
Chr.17: 29929856-30108452 on Build GRCh38
Transcripts
NM_001145053.1 NM_001282129.1 NM_001282130.1 NM_033389.3 XM_005258058.3 XM_017025251.1 XM_017025252.1 XM_017025253.1 XM_017025254.1 XM_017025255.1

Gene Symbol
SSH2
Entrez Gene ID
85464
Gene Name
slingshot protein phosphatase 2
Gene Aliases
SSH-2, SSH-2L
UniGene
Hs.654754
Gene Chromosome Location
Chr.17: 29625938-29930239 on Build GRCh38
Transcripts
NM_001145053.1 NM_001282129.1 NM_001282130.1 NM_033389.3 XM_005258058.3 XM_017025251.1 XM_017025252.1 XM_017025253.1 XM_017025254.1 XM_017025255.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6202116 c.35C>T p.P12L Chr.17: 29929966-29929966 on Build GRCh38
6790779 c.2T>C p.M1T Chr.17: 29929999-29929999 on Build GRCh38
5573585 c.63+2T>C p.? Chr.17: 29929936-29929936 on Build GRCh38
6258628 c.62C>A p.S21* Chr.17: 29929939-29929939 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1000538162 Chr.17: 29929923-29929923 on Build GRCh38
rs989021435 Chr.17: 29929920-29929920 on Build GRCh38
rs901171311 Chr.17: 29929875-29929875 on Build GRCh38
rs760096513 Chr.17: 29929902-29929902 on Build GRCh38
rs770410251 Chr.17: 29930028-29930028 on Build GRCh38
rs1009890551 Chr.17: 29930222-29930222 on Build GRCh38
rs1051766180 Chr.17: 29930096-29930096 on Build GRCh38
rs889984391 Chr.17: 29929893-29929893 on Build GRCh38
rs867530617 Chr.17: 29929780-29929780 on Build GRCh38
rs927180794 Chr.17: 29929804-29929804 on Build GRCh38
rs901287928 Chr.17: 29930197-29930197 on Build GRCh38
rs564979271 Chr.17: 29929837-29929838 on Build GRCh38
rs546088909 Chr.17: 29929844-29929844 on Build GRCh38
rs930382144 Chr.17: 29930072-29930072 on Build GRCh38
rs982327400 Chr.17: 29930053-29930053 on Build GRCh38
rs1032862001 Chr.17: 29929949-29929949 on Build GRCh38
rs938245780 Chr.17: 29929813-29929813 on Build GRCh38
rs889631003 Chr.17: 29930209-29930209 on Build GRCh38
rs938166055 Chr.17: 29930151-29930151 on Build GRCh38
rs778736605 Chr.17: 29929801-29929801 on Build GRCh38
rs541848058 Chr.17: 29929793-29929793 on Build GRCh38
rs566495468 Chr.17: 29930186-29930186 on Build GRCh38
rs369553394 Chr.17: 29929917-29929917 on Build GRCh38
rs775938040 Chr.17: 29930045-29930045 on Build GRCh38
rs998658148 Chr.17: 29930204-29930204 on Build GRCh38
rs763161164 Chr.17: 29930046-29930046 on Build GRCh38
rs369609551 Chr.17: 29929966-29929966 on Build GRCh38
rs534243488 Chr.17: 29930194-29930194 on Build GRCh38
rs940043855 Chr.17: 29930188-29930188 on Build GRCh38
rs901256383 Chr.17: 29929862-29929862 on Build GRCh38
rs572385388 Chr.17: 29929818-29929818 on Build GRCh38
rs949844511 Chr.17: 29929800-29929800 on Build GRCh38
rs761586064 Chr.17: 29929913-29929913 on Build GRCh38
rs1032891475 Chr.17: 29930213-29930213 on Build GRCh38
rs907670613 Chr.17: 29930117-29930117 on Build GRCh38
rs758647306 Chr.17: 29929973-29929973 on Build GRCh38
rs766328918 Chr.17: 29929944-29929944 on Build GRCh38
rs753825036 Chr.17: 29929948-29929948 on Build GRCh38
rs930456943 Chr.17: 29929806-29929806 on Build GRCh38
rs746444236 Chr.17: 29930027-29930027 on Build GRCh38
rs891958573 Chr.17: 29930206-29930206 on Build GRCh38
rs1049811387 Chr.17: 29930205-29930205 on Build GRCh38
rs955951442 Chr.17: 29930216-29930216 on Build GRCh38
rs111605484 Chr.17: 29929826-29929826 on Build GRCh38
rs771182861 Chr.17: 29930007-29930007 on Build GRCh38
rs778750653 Chr.17: 29929965-29929965 on Build GRCh38
rs777228281 Chr.17: 29929890-29929890 on Build GRCh38
rs781388814 Chr.17: 29930022-29930022 on Build GRCh38
rs955711923 Chr.17: 29929952-29929952 on Build GRCh38
rs768354117 Chr.17: 29929903-29929904 on Build GRCh38
rs8071113 Chr.17: 29930210-29930210 on Build GRCh38
rs777816061 Chr.17: 29929977-29929977 on Build GRCh38
rs145397094 Chr.17: 29929782-29929782 on Build GRCh38
rs147061646 Chr.17: 29929953-29929953 on Build GRCh38
rs370191442 Chr.17: 29930049-29930049 on Build GRCh38
rs998064955 Chr.17: 29929876-29929876 on Build GRCh38
rs1040306613 Chr.17: 29930152-29930155 on Build GRCh38
rs7209622 Chr.17: 29929789-29929789 on Build GRCh38
rs1015689500 Chr.17: 29929845-29929845 on Build GRCh38
rs770322310 Chr.17: 29929912-29929912 on Build GRCh38
rs1048656068 Chr.17: 29929816-29929816 on Build GRCh38
rs905474180 Chr.17: 29929805-29929805 on Build GRCh38
rs760064584 Chr.17: 29929926-29929926 on Build GRCh38
rs1001621669 Chr.17: 29930223-29930223 on Build GRCh38
rs548190330 Chr.17: 29930133-29930133 on Build GRCh38
rs551093286 Chr.17: 29930056-29930056 on Build GRCh38
rs374291688 Chr.17: 29929986-29929986 on Build GRCh38
rs1048688258 Chr.17: 29930185-29930185 on Build GRCh38
rs562840834 Chr.17: 29930079-29930079 on Build GRCh38
rs530091563 Chr.17: 29930086-29930086 on Build GRCh38
rs879942292 Chr.17: 29930173-29930173 on Build GRCh38
rs971652239 Chr.17: 29930218-29930218 on Build GRCh38
rs560149491 Chr.17: 29929800-29929800 on Build GRCh38

View Full Product Details