Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 19021580-19022075 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRAP
Entrez Gene ID
10750
Gene Name
GRB2-related adaptor protein
Gene Aliases
-
UniGene
Hs.567416
Gene Chromosome Location
Chr.17: 19020656-19051352 on Build GRCh38
Transcripts
NM_006613.3 XM_017024029.1 XM_017024030.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5669045 c.574G>T p.D192Y Chr.17: 19022039-19022039 on Build GRCh38
7349347 c.583T>C p.W195R Chr.17: 19022030-19022030 on Build GRCh38
4688397 c.569G>A p.R190H Chr.17: 19022044-19022044 on Build GRCh38
4554669 c.627G>A p.R209R Chr.17: 19021986-19021986 on Build GRCh38
5385834 c.626G>A p.R209Q Chr.17: 19021987-19021987 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs993486015 Chr.17: 19021612-19021612 on Build GRCh38
rs766540399 Chr.17: 19022041-19022041 on Build GRCh38
rs879545958 Chr.17: 19021761-19021761 on Build GRCh38
rs1033840439 Chr.17: 19021651-19021651 on Build GRCh38
rs769503688 Chr.17: 19021866-19021866 on Build GRCh38
rs766873552 Chr.17: 19021951-19021951 on Build GRCh38
rs528150785 Chr.17: 19021848-19021848 on Build GRCh38
rs775265448 Chr.17: 19022005-19022005 on Build GRCh38
rs946870235 Chr.17: 19021758-19021770 on Build GRCh38
rs756757755 Chr.17: 19021983-19021983 on Build GRCh38
rs749150633 Chr.17: 19022009-19022009 on Build GRCh38
rs180846047 Chr.17: 19021867-19021867 on Build GRCh38
rs377220975 Chr.17: 19021980-19021980 on Build GRCh38
rs985298031 Chr.17: 19021788-19021788 on Build GRCh38
rs375764703 Chr.17: 19021989-19021989 on Build GRCh38
rs372985295 Chr.17: 19022024-19022024 on Build GRCh38
rs565957769 Chr.17: 19021602-19021602 on Build GRCh38
rs547412312 Chr.17: 19021984-19021984 on Build GRCh38
rs763368287 Chr.17: 19021954-19021954 on Build GRCh38
rs539058799 Chr.17: 19021930-19021930 on Build GRCh38
rs1057399 Chr.17: 19021835-19021835 on Build GRCh38
rs766784776 Chr.17: 19022034-19022034 on Build GRCh38
rs550499409 Chr.17: 19021896-19021896 on Build GRCh38
rs755548763 Chr.17: 19021962-19021962 on Build GRCh38
rs774325073 Chr.17: 19022018-19022018 on Build GRCh38
rs746504231 Chr.17: 19021975-19021975 on Build GRCh38
rs761858852 Chr.17: 19021899-19021899 on Build GRCh38
rs545470060 Chr.17: 19021814-19021814 on Build GRCh38
rs760821883 Chr.17: 19022028-19022028 on Build GRCh38
rs779619850 Chr.17: 19021972-19021972 on Build GRCh38
rs368972418 Chr.17: 19021838-19021838 on Build GRCh38
rs140507873 Chr.17: 19021852-19021852 on Build GRCh38
rs767524784 Chr.17: 19022023-19022023 on Build GRCh38
rs929322524 Chr.17: 19021857-19021857 on Build GRCh38
rs887851098 Chr.17: 19021631-19021631 on Build GRCh38
rs188114963 Chr.17: 19021673-19021673 on Build GRCh38
rs769570810 Chr.17: 19022003-19022003 on Build GRCh38
rs185335436 Chr.17: 19021626-19021626 on Build GRCh38
rs925051936 Chr.17: 19021724-19021724 on Build GRCh38
rs1006422817 Chr.17: 19021648-19021648 on Build GRCh38
rs1041373889 Chr.17: 19021834-19021834 on Build GRCh38
rs755441479 Chr.17: 19022044-19022044 on Build GRCh38
rs147181892 Chr.17: 19021686-19021686 on Build GRCh38
rs992418762 Chr.17: 19021674-19021675 on Build GRCh38
rs552267271 Chr.17: 19021932-19021932 on Build GRCh38
rs767761158 Chr.17: 19021907-19021907 on Build GRCh38
rs576818023 Chr.17: 19021805-19021805 on Build GRCh38
rs543748807 Chr.17: 19021679-19021679 on Build GRCh38
rs17847612 Chr.17: 19021996-19021996 on Build GRCh38
rs895328754 Chr.17: 19021967-19021967 on Build GRCh38
rs530187066 Chr.17: 19021876-19021876 on Build GRCh38
rs765807135 Chr.17: 19022045-19022045 on Build GRCh38
rs966796233 Chr.17: 19021681-19021681 on Build GRCh38
rs767188423 Chr.17: 19021862-19021862 on Build GRCh38
rs762967703 Chr.17: 19021905-19021905 on Build GRCh38
rs369064348 Chr.17: 19022010-19022010 on Build GRCh38
rs754240410 Chr.17: 19022042-19022042 on Build GRCh38
rs756461457 Chr.17: 19021945-19021945 on Build GRCh38
rs1057400 Chr.17: 19021812-19021812 on Build GRCh38
rs759344407 Chr.17: 19021853-19021853 on Build GRCh38
rs113703856 Chr.17: 19021791-19021791 on Build GRCh38

View Full Product Details