Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 43780348-43780846 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C17orf105
Entrez Gene ID
284067
Gene Name
chromosome 17 open reading frame 105
Gene Aliases
-
UniGene
Hs.252707
Gene Chromosome Location
Chr.17: 43778391-43784842 on Build GRCh38
Transcripts
NM_001136483.1 XM_011524633.1 XM_011524634.2 XM_011524635.1 XM_011524636.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5822106 c.98T>C p.V33A Chr.17: 43780560-43780560 on Build GRCh38
1198222 c.115A>G p.I39V Chr.17: 43780577-43780577 on Build GRCh38
4976059 c.44C>T p.S15F Chr.17: 43780506-43780506 on Build GRCh38
6587616 c.75G>A p.K25K Chr.17: 43780537-43780537 on Build GRCh38
5464050 c.81G>A p.L27L Chr.17: 43780543-43780543 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs570579350 Chr.17: 43780385-43780385 on Build GRCh38
rs958699367 Chr.17: 43780508-43780508 on Build GRCh38
rs991527474 Chr.17: 43780554-43780554 on Build GRCh38
rs550596386 Chr.17: 43780640-43780640 on Build GRCh38
rs777725073 Chr.17: 43780558-43780558 on Build GRCh38
rs557494278 Chr.17: 43780697-43780697 on Build GRCh38
rs780545436 Chr.17: 43780587-43780587 on Build GRCh38
rs1048081600 Chr.17: 43780722-43780722 on Build GRCh38
rs981564893 Chr.17: 43780487-43780487 on Build GRCh38
rs539230907 Chr.17: 43780688-43780688 on Build GRCh38
rs563814513 Chr.17: 43780807-43780807 on Build GRCh38
rs964728057 Chr.17: 43780592-43780592 on Build GRCh38
rs576943947 Chr.17: 43780733-43780733 on Build GRCh38
rs769217591 Chr.17: 43780629-43780629 on Build GRCh38
rs754012548 Chr.17: 43780442-43780442 on Build GRCh38
rs372343752 Chr.17: 43780517-43780517 on Build GRCh38
rs771048192 Chr.17: 43780564-43780564 on Build GRCh38
rs758220443 Chr.17: 43780555-43780555 on Build GRCh38
rs1004443983 Chr.17: 43780412-43780412 on Build GRCh38
rs770348938 Chr.17: 43780701-43780701 on Build GRCh38
rs567922770 Chr.17: 43780542-43780542 on Build GRCh38
rs745421466 Chr.17: 43780593-43780593 on Build GRCh38
rs868354304 Chr.17: 43780375-43780375 on Build GRCh38
rs9908083 Chr.17: 43780636-43780636 on Build GRCh38
rs755136313 Chr.17: 43780458-43780458 on Build GRCh38
rs920888927 Chr.17: 43780675-43780675 on Build GRCh38
rs938273361 Chr.17: 43780766-43780766 on Build GRCh38
rs962417170 Chr.17: 43780457-43780457 on Build GRCh38
rs924822535 Chr.17: 43780655-43780655 on Build GRCh38
rs187854750 Chr.17: 43780768-43780768 on Build GRCh38
rs73987086 Chr.17: 43780501-43780501 on Build GRCh38
rs1040778709 Chr.17: 43780660-43780660 on Build GRCh38
rs1037246058 Chr.17: 43780743-43780743 on Build GRCh38
rs1009588414 Chr.17: 43780818-43780818 on Build GRCh38
rs746877537 Chr.17: 43780559-43780559 on Build GRCh38
rs528165628 Chr.17: 43780448-43780448 on Build GRCh38
rs752896043 Chr.17: 43780516-43780516 on Build GRCh38
rs2301682 Chr.17: 43780658-43780658 on Build GRCh38
rs775854585 Chr.17: 43780724-43780724 on Build GRCh38
rs958813642 Chr.17: 43780800-43780800 on Build GRCh38
rs566694253 Chr.17: 43780739-43780739 on Build GRCh38
rs1015454420 Chr.17: 43780440-43780440 on Build GRCh38
rs747064503 Chr.17: 43780380-43780380 on Build GRCh38
rs553375966 Chr.17: 43780745-43780745 on Build GRCh38
rs574740653 Chr.17: 43780752-43780752 on Build GRCh38
rs760601570 Chr.17: 43780799-43780799 on Build GRCh38

View Full Product Details