Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 44758968-44759501 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAM11
Entrez Gene ID
4185
Gene Name
ADAM metallopeptidase domain 11
Gene Aliases
MDC
UniGene
Hs.6088
Gene Chromosome Location
Chr.17: 44759037-44781846 on Build GRCh38
Transcripts
NM_001318933.1 NM_002390.5 XM_005257373.3

SNP Detail

SNP ID Chromosome Location
rs919164280 Chr.17: 44759425-44759425 on Build GRCh38
rs541960716 Chr.17: 44759455-44759455 on Build GRCh38
rs1023140747 Chr.17: 44759131-44759131 on Build GRCh38
rs556160832 Chr.17: 44759314-44759314 on Build GRCh38
rs1004270063 Chr.17: 44759085-44759085 on Build GRCh38
rs971931036 Chr.17: 44759193-44759193 on Build GRCh38
rs1037813623 Chr.17: 44759404-44759404 on Build GRCh38
rs1031855159 Chr.17: 44759215-44759215 on Build GRCh38
rs910495011 Chr.17: 44759035-44759035 on Build GRCh38
rs914133844 Chr.17: 44759321-44759321 on Build GRCh38
rs1013533902 Chr.17: 44759099-44759099 on Build GRCh38
rs993471479 Chr.17: 44759070-44759070 on Build GRCh38
rs970175598 Chr.17: 44759247-44759247 on Build GRCh38
rs998193121 Chr.17: 44759419-44759419 on Build GRCh38
rs944516332 Chr.17: 44759395-44759395 on Build GRCh38
rs887864032 Chr.17: 44759083-44759083 on Build GRCh38
rs910755869 Chr.17: 44759373-44759373 on Build GRCh38
rs1047777363 Chr.17: 44759474-44759474 on Build GRCh38
rs888310235 Chr.17: 44759074-44759074 on Build GRCh38
rs956402183 Chr.17: 44759220-44759220 on Build GRCh38
rs986250926 Chr.17: 44759338-44759338 on Build GRCh38
rs752441017 Chr.17: 44759269-44759269 on Build GRCh38
rs925593329 Chr.17: 44759289-44759289 on Build GRCh38
rs965330983 Chr.17: 44759148-44759148 on Build GRCh38
rs897848687 Chr.17: 44759063-44759064 on Build GRCh38
rs1049204904 Chr.17: 44759077-44759077 on Build GRCh38
rs987749271 Chr.17: 44759231-44759231 on Build GRCh38
rs577597816 Chr.17: 44759457-44759457 on Build GRCh38
rs774361323 Chr.17: 44759298-44759303 on Build GRCh38
rs933578500 Chr.17: 44759348-44759348 on Build GRCh38
rs759181330 Chr.17: 44759217-44759217 on Build GRCh38
rs1051847419 Chr.17: 44759369-44759369 on Build GRCh38
rs993466941 Chr.17: 44759459-44759459 on Build GRCh38
rs1040277385 Chr.17: 44759047-44759047 on Build GRCh38
rs943445448 Chr.17: 44759386-44759386 on Build GRCh38
rs544457368 Chr.17: 44759006-44759006 on Build GRCh38
rs1015704190 Chr.17: 44759098-44759098 on Build GRCh38
rs1048207222 Chr.17: 44759063-44759063 on Build GRCh38
rs988451832 Chr.17: 44759318-44759318 on Build GRCh38
rs978566468 Chr.17: 44759281-44759281 on Build GRCh38
rs764824719 Chr.17: 44759240-44759240 on Build GRCh38
rs929328681 Chr.17: 44759440-44759440 on Build GRCh38
rs866870166 Chr.17: 44759020-44759020 on Build GRCh38
rs762481952 Chr.17: 44759284-44759284 on Build GRCh38
rs776641756 Chr.17: 44759195-44759195 on Build GRCh38
rs944648655 Chr.17: 44759044-44759044 on Build GRCh38

View Full Product Details