Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 34795511-34796016 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GJA4
Entrez Gene ID
2701
Gene Name
gap junction protein alpha 4
Gene Aliases
CX37
UniGene
Hs.296310
Gene Chromosome Location
Chr.1: 34792958-34795747 on Build GRCh38
Transcripts
NM_002060.2 XM_005270750.2 XM_017001043.1

SNP Detail

SNP ID Chromosome Location
rs28473760 Chr.1: 34795674-34795674 on Build GRCh38
rs77051837 Chr.1: 34795985-34795985 on Build GRCh38
rs1052183795 Chr.1: 34795841-34795841 on Build GRCh38
rs768965399 Chr.1: 34795912-34795912 on Build GRCh38
rs996154546 Chr.1: 34795602-34795602 on Build GRCh38
rs994110828 Chr.1: 34795920-34795920 on Build GRCh38
rs529298730 Chr.1: 34795888-34795888 on Build GRCh38
rs952689797 Chr.1: 34795978-34795978 on Build GRCh38
rs697372 Chr.1: 34795565-34795565 on Build GRCh38
rs705193 Chr.1: 34795631-34795631 on Build GRCh38
rs1044760521 Chr.1: 34795789-34795789 on Build GRCh38
rs966465968 Chr.1: 34795599-34795599 on Build GRCh38
rs1008045483 Chr.1: 34795867-34795867 on Build GRCh38
rs746372730 Chr.1: 34795651-34795651 on Build GRCh38
rs557392830 Chr.1: 34795661-34795661 on Build GRCh38
rs1028951426 Chr.1: 34795944-34795944 on Build GRCh38
rs892323720 Chr.1: 34795862-34795862 on Build GRCh38
rs534459371 Chr.1: 34795542-34795542 on Build GRCh38
rs1041254856 Chr.1: 34795871-34795871 on Build GRCh38
rs984798897 Chr.1: 34795632-34795632 on Build GRCh38
rs952042505 Chr.1: 34795957-34795957 on Build GRCh38
rs1006556859 Chr.1: 34795533-34795533 on Build GRCh38
rs575861763 Chr.1: 34795737-34795737 on Build GRCh38
rs1025918989 Chr.1: 34795622-34795622 on Build GRCh38
rs1019250375 Chr.1: 34795546-34795546 on Build GRCh38
rs562196340 Chr.1: 34795812-34795812 on Build GRCh38
rs1019516474 Chr.1: 34795597-34795597 on Build GRCh38
rs568000664 Chr.1: 34795589-34795589 on Build GRCh38
rs373276545 Chr.1: 34795586-34795586 on Build GRCh38
rs1025548910 Chr.1: 34795963-34795963 on Build GRCh38
rs918156135 Chr.1: 34795770-34795770 on Build GRCh38
rs573468876 Chr.1: 34795801-34795801 on Build GRCh38
rs567356042 Chr.1: 34795652-34795652 on Build GRCh38
rs546427527 Chr.1: 34795747-34795747 on Build GRCh38
rs917215635 Chr.1: 34795759-34795759 on Build GRCh38
rs374142184 Chr.1: 34795930-34795930 on Build GRCh38
rs946316277 Chr.1: 34795780-34795780 on Build GRCh38
rs774869708 Chr.1: 34795784-34795784 on Build GRCh38
rs1040745128 Chr.1: 34795847-34795847 on Build GRCh38
rs766139655 Chr.1: 34795794-34795795 on Build GRCh38
rs544564730 Chr.1: 34795927-34795927 on Build GRCh38
rs889543671 Chr.1: 34795815-34795815 on Build GRCh38
rs3738353 Chr.1: 34795804-34795804 on Build GRCh38
rs1028923943 Chr.1: 34795606-34795606 on Build GRCh38
rs558335199 Chr.1: 34795777-34795777 on Build GRCh38
rs958739005 Chr.1: 34795690-34795690 on Build GRCh38
rs769506256 Chr.1: 34795718-34795718 on Build GRCh38
rs1006608620 Chr.1: 34795830-34795830 on Build GRCh38

View Full Product Details