Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 35714571-35715078 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf216
Entrez Gene ID
127703
Gene Name
chromosome 1 open reading frame 216
Gene Aliases
-
UniGene
Hs.112023
Gene Chromosome Location
Chr.1: 35713876-35719189 on Build GRCh38
Transcripts
NM_152374.1

SNP Detail

SNP ID Chromosome Location
rs577476483 Chr.1: 35714814-35714814 on Build GRCh38
rs146146969 Chr.1: 35714671-35714671 on Build GRCh38
rs116504001 Chr.1: 35714720-35714720 on Build GRCh38
rs143908579 Chr.1: 35714675-35714675 on Build GRCh38
rs753977504 Chr.1: 35714749-35714749 on Build GRCh38
rs28578485 Chr.1: 35714952-35714952 on Build GRCh38
rs386630304 Chr.1: 35714672-35714675 on Build GRCh38
rs918108583 Chr.1: 35714601-35714601 on Build GRCh38
rs772975336 Chr.1: 35714762-35714762 on Build GRCh38
rs534296921 Chr.1: 35714869-35714869 on Build GRCh38
rs1019693409 Chr.1: 35714959-35714961 on Build GRCh38
rs959669473 Chr.1: 35715024-35715024 on Build GRCh38
rs1052389496 Chr.1: 35714673-35714673 on Build GRCh38
rs548751576 Chr.1: 35714654-35714654 on Build GRCh38
rs1007335273 Chr.1: 35714937-35714937 on Build GRCh38
rs41267255 Chr.1: 35715050-35715050 on Build GRCh38
rs1025868857 Chr.1: 35714650-35714650 on Build GRCh38
rs1045350574 Chr.1: 35714622-35714622 on Build GRCh38
rs148520098 Chr.1: 35715021-35715021 on Build GRCh38
rs1015280306 Chr.1: 35714930-35714930 on Build GRCh38
rs952739067 Chr.1: 35714889-35714889 on Build GRCh38
rs904097777 Chr.1: 35714637-35714637 on Build GRCh38
rs965565753 Chr.1: 35714601-35714602 on Build GRCh38
rs140168275 Chr.1: 35714674-35714674 on Build GRCh38
rs889780240 Chr.1: 35714936-35714936 on Build GRCh38
rs146772607 Chr.1: 35714993-35714993 on Build GRCh38
rs948202519 Chr.1: 35714603-35714603 on Build GRCh38
rs546221504 Chr.1: 35714903-35714903 on Build GRCh38
rs1026943067 Chr.1: 35714734-35714734 on Build GRCh38
rs369474144 Chr.1: 35714685-35714685 on Build GRCh38
rs1025836229 Chr.1: 35715045-35715045 on Build GRCh38
rs576803457 Chr.1: 35714836-35714836 on Build GRCh38
rs899692829 Chr.1: 35714713-35714713 on Build GRCh38
rs1041537903 Chr.1: 35714734-35714735 on Build GRCh38
rs952954613 Chr.1: 35714670-35714672 on Build GRCh38
rs1050325209 Chr.1: 35714854-35714854 on Build GRCh38
rs917026473 Chr.1: 35714707-35714707 on Build GRCh38

View Full Product Details