Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 45236052-45236559 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FMNL1
Entrez Gene ID
752
Gene Name
formin like 1
Gene Aliases
C17orf1, C17orf1B, FHOD4, FMNL, KW-13
UniGene
Hs.100217
Gene Chromosome Location
Chr.17: 45221728-45247322 on Build GRCh38
Transcripts
NM_005892.3 XM_006722062.3 XM_006722063.3 XM_006722064.3 XM_006722065.3 XM_006722066.3 XM_006722069.2 XM_006722070.3 XM_011525179.2 XM_011525180.2 XM_011525182.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4554906 c.635G>A p.R212K Chr.17: 45236156-45236156 on Build GRCh38
1383787 c.685G>A p.V229I Chr.17: 45236206-45236206 on Build GRCh38
6081166 c.684C>T p.H228H Chr.17: 45236205-45236205 on Build GRCh38
7202645 c.654C>T p.S218S Chr.17: 45236175-45236175 on Build GRCh38
4637761 c.625G>T p.A209S Chr.17: 45236146-45236146 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746166316 Chr.17: 45236284-45236284 on Build GRCh38
rs144714204 Chr.17: 45236185-45236185 on Build GRCh38
rs757671058 Chr.17: 45236280-45236280 on Build GRCh38
rs924245059 Chr.17: 45236423-45236423 on Build GRCh38
rs914319970 Chr.17: 45236457-45236457 on Build GRCh38
rs200547847 Chr.17: 45236168-45236168 on Build GRCh38
rs374960639 Chr.17: 45236144-45236144 on Build GRCh38
rs995515360 Chr.17: 45236520-45236520 on Build GRCh38
rs563802546 Chr.17: 45236526-45236526 on Build GRCh38
rs778551863 Chr.17: 45236271-45236271 on Build GRCh38
rs187296602 Chr.17: 45236424-45236424 on Build GRCh38
rs780760900 Chr.17: 45236096-45236096 on Build GRCh38
rs897426487 Chr.17: 45236519-45236519 on Build GRCh38
rs535461614 Chr.17: 45236189-45236189 on Build GRCh38
rs201153167 Chr.17: 45236230-45236230 on Build GRCh38
rs1037235345 Chr.17: 45236508-45236508 on Build GRCh38
rs147244126 Chr.17: 45236411-45236411 on Build GRCh38
rs374834363 Chr.17: 45236281-45236281 on Build GRCh38
rs182196971 Chr.17: 45236252-45236252 on Build GRCh38
rs148536209 Chr.17: 45236199-45236199 on Build GRCh38
rs73984349 Chr.17: 45236277-45236277 on Build GRCh38
rs1049845534 Chr.17: 45236482-45236482 on Build GRCh38
rs776852929 Chr.17: 45236155-45236155 on Build GRCh38
rs775397083 Chr.17: 45236169-45236169 on Build GRCh38
rs774475483 Chr.17: 45236227-45236227 on Build GRCh38
rs747529739 Chr.17: 45236202-45236202 on Build GRCh38
rs753545097 Chr.17: 45236263-45236263 on Build GRCh38
rs767205834 Chr.17: 45236182-45236182 on Build GRCh38
rs748871175 Chr.17: 45236130-45236130 on Build GRCh38
rs915569611 Chr.17: 45236370-45236370 on Build GRCh38
rs386364984 Chr.17: 45236474-45236475 on Build GRCh38
rs1022811185 Chr.17: 45236301-45236301 on Build GRCh38
rs763983223 Chr.17: 45236179-45236179 on Build GRCh38
rs757079078 Chr.17: 45236094-45236094 on Build GRCh38
rs371416228 Chr.17: 45236184-45236184 on Build GRCh38
rs951747273 Chr.17: 45236336-45236336 on Build GRCh38
rs112471450 Chr.17: 45236358-45236358 on Build GRCh38
rs769880915 Chr.17: 45236216-45236216 on Build GRCh38
rs998047335 Chr.17: 45236417-45236417 on Build GRCh38
rs759663688 Chr.17: 45236156-45236156 on Build GRCh38
rs755856437 Chr.17: 45236129-45236129 on Build GRCh38
rs778357711 Chr.17: 45236200-45236200 on Build GRCh38
rs771426176 Chr.17: 45236149-45236149 on Build GRCh38
rs199632499 Chr.17: 45236226-45236226 on Build GRCh38
rs934463141 Chr.17: 45236514-45236514 on Build GRCh38
rs528855994 Chr.17: 45236428-45236428 on Build GRCh38
rs762957389 Chr.17: 45236225-45236225 on Build GRCh38
rs374827847 Chr.17: 45236269-45236269 on Build GRCh38
rs990262289 Chr.17: 45236438-45236438 on Build GRCh38
rs566668495 Chr.17: 45236408-45236408 on Build GRCh38
rs113422375 Chr.17: 45236471-45236472 on Build GRCh38
rs971263795 Chr.17: 45236381-45236381 on Build GRCh38
rs778447690 Chr.17: 45236143-45236143 on Build GRCh38
rs767532251 Chr.17: 45236233-45236233 on Build GRCh38
rs760541784 Chr.17: 45236251-45236251 on Build GRCh38
rs781402859 Chr.17: 45236206-45236206 on Build GRCh38
rs745522670 Chr.17: 45236119-45236119 on Build GRCh38
rs893095656 Chr.17: 45236527-45236527 on Build GRCh38
rs561768313 Chr.17: 45236318-45236318 on Build GRCh38
rs368489127 Chr.17: 45236250-45236250 on Build GRCh38
rs149409789 Chr.17: 45236093-45236093 on Build GRCh38
rs1054224479 Chr.17: 45236106-45236106 on Build GRCh38
rs981374184 Chr.17: 45236389-45236389 on Build GRCh38
rs924463367 Chr.17: 45236476-45236476 on Build GRCh38
rs112328895 Chr.17: 45236488-45236488 on Build GRCh38
rs991599860 Chr.17: 45236368-45236368 on Build GRCh38
rs912750873 Chr.17: 45236492-45236492 on Build GRCh38
rs763065736 Chr.17: 45236176-45236176 on Build GRCh38
rs1027204425 Chr.17: 45236316-45236316 on Build GRCh38
rs367859403 Chr.17: 45236092-45236092 on Build GRCh38
rs892904881 Chr.17: 45236124-45236124 on Build GRCh38
rs146916852 Chr.17: 45236137-45236137 on Build GRCh38
rs974334627 Chr.17: 45236299-45236299 on Build GRCh38
rs757427674 Chr.17: 45236205-45236205 on Build GRCh38
rs374169714 Chr.17: 45236208-45236208 on Build GRCh38
rs775801071 Chr.17: 45236224-45236224 on Build GRCh38
rs751506418 Chr.17: 45236181-45236181 on Build GRCh38
rs944266246 Chr.17: 45236495-45236495 on Build GRCh38
rs767012221 Chr.17: 45236196-45236196 on Build GRCh38
rs369812651 Chr.17: 45236283-45236283 on Build GRCh38
rs930266349 Chr.17: 45236083-45236083 on Build GRCh38

View Full Product Details