Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 45237139-45237633 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FMNL1
Entrez Gene ID
752
Gene Name
formin like 1
Gene Aliases
C17orf1, C17orf1B, FHOD4, FMNL, KW-13
UniGene
Hs.100217
Gene Chromosome Location
Chr.17: 45221728-45247322 on Build GRCh38
Transcripts
NM_005892.3 XM_006722062.3 XM_006722063.3 XM_006722064.3 XM_006722065.3 XM_006722066.3 XM_006722069.2 XM_006722070.3 XM_011525179.2 XM_011525180.2 XM_011525182.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5565379 c.847C>T p.R283W Chr.17: 45237592-45237592 on Build GRCh38
5226034 c.850G>A p.G284R Chr.17: 45237595-45237595 on Build GRCh38
5419855 c.741C>T p.V247V Chr.17: 45237298-45237298 on Build GRCh38
436728 c.846G>A p.V282V Chr.17: 45237591-45237591 on Build GRCh38
3518235 c.864C>T p.I288I Chr.17: 45237609-45237609 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs114245235 Chr.17: 45237487-45237487 on Build GRCh38
rs544206078 Chr.17: 45237227-45237227 on Build GRCh38
rs1006761929 Chr.17: 45237570-45237570 on Build GRCh38
rs766209725 Chr.17: 45237309-45237309 on Build GRCh38
rs749905758 Chr.17: 45237500-45237500 on Build GRCh38
rs952977489 Chr.17: 45237437-45237437 on Build GRCh38
rs35909374 Chr.17: 45237577-45237577 on Build GRCh38
rs1021945690 Chr.17: 45237228-45237228 on Build GRCh38
rs935406211 Chr.17: 45237209-45237209 on Build GRCh38
rs746447740 Chr.17: 45237361-45237362 on Build GRCh38
rs528624251 Chr.17: 45237462-45237462 on Build GRCh38
rs1002534416 Chr.17: 45237194-45237194 on Build GRCh38
rs764928976 Chr.17: 45237556-45237556 on Build GRCh38
rs955819836 Chr.17: 45237214-45237214 on Build GRCh38
rs914408808 Chr.17: 45237219-45237219 on Build GRCh38
rs1031290754 Chr.17: 45237195-45237195 on Build GRCh38
rs753423557 Chr.17: 45237237-45237238 on Build GRCh38
rs748278646 Chr.17: 45237282-45237282 on Build GRCh38
rs771856191 Chr.17: 45237294-45237294 on Build GRCh38
rs773671320 Chr.17: 45237400-45237402 on Build GRCh38
rs138651184 Chr.17: 45237592-45237592 on Build GRCh38
rs182570413 Chr.17: 45237499-45237499 on Build GRCh38
rs780397132 Chr.17: 45237406-45237406 on Build GRCh38
rs756639917 Chr.17: 45237403-45237403 on Build GRCh38
rs775311487 Chr.17: 45237567-45237567 on Build GRCh38
rs369408882 Chr.17: 45237516-45237516 on Build GRCh38
rs569280215 Chr.17: 45237553-45237553 on Build GRCh38
rs369841052 Chr.17: 45237386-45237386 on Build GRCh38
rs200464299 Chr.17: 45237580-45237580 on Build GRCh38
rs749697608 Chr.17: 45237497-45237497 on Build GRCh38
rs768936426 Chr.17: 45237251-45237251 on Build GRCh38
rs912440359 Chr.17: 45237267-45237267 on Build GRCh38
rs377527873 Chr.17: 45237514-45237514 on Build GRCh38
rs955682584 Chr.17: 45237481-45237481 on Build GRCh38
rs77496285 Chr.17: 45237273-45237273 on Build GRCh38
rs182815218 Chr.17: 45237220-45237220 on Build GRCh38
rs780195441 Chr.17: 45237238-45237238 on Build GRCh38
rs186056963 Chr.17: 45237390-45237390 on Build GRCh38
rs768217664 Chr.17: 45237386-45237387 on Build GRCh38
rs765018636 Chr.17: 45237372-45237372 on Build GRCh38
rs77766814 Chr.17: 45237177-45237177 on Build GRCh38
rs372821100 Chr.17: 45237235-45237235 on Build GRCh38
rs868773576 Chr.17: 45237578-45237578 on Build GRCh38
rs1037067048 Chr.17: 45237296-45237296 on Build GRCh38
rs906820887 Chr.17: 45237458-45237458 on Build GRCh38
rs772962912 Chr.17: 45237511-45237511 on Build GRCh38
rs776535465 Chr.17: 45237526-45237526 on Build GRCh38
rs1012379027 Chr.17: 45237431-45237431 on Build GRCh38
rs547963485 Chr.17: 45237579-45237579 on Build GRCh38
rs531104370 Chr.17: 45237345-45237345 on Build GRCh38
rs879249761 Chr.17: 45237353-45237353 on Build GRCh38
rs546524920 Chr.17: 45237377-45237377 on Build GRCh38
rs1042931325 Chr.17: 45237541-45237541 on Build GRCh38
rs373201342 Chr.17: 45237576-45237576 on Build GRCh38
rs190079247 Chr.17: 45237474-45237474 on Build GRCh38
rs760319880 Chr.17: 45237306-45237306 on Build GRCh38
rs751785973 Chr.17: 45237457-45237457 on Build GRCh38
rs918577044 Chr.17: 45237312-45237312 on Build GRCh38
rs774223435 Chr.17: 45237215-45237215 on Build GRCh38
rs372763331 Chr.17: 45237498-45237498 on Build GRCh38
rs200827907 Chr.17: 45237303-45237303 on Build GRCh38
rs1046471966 Chr.17: 45237169-45237169 on Build GRCh38
rs759256335 Chr.17: 45237361-45237361 on Build GRCh38
rs757945544 Chr.17: 45237385-45237385 on Build GRCh38
rs982070059 Chr.17: 45237482-45237482 on Build GRCh38

View Full Product Details