Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 48722223-48722716 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRAC1
Entrez Gene ID
84366
Gene Name
prostate cancer susceptibility candidate 1
Gene Aliases
C17orf92, PRAC
UniGene
Hs.116467
Gene Chromosome Location
Chr.17: 48721719-48722520 on Build GRCh38
Transcripts
NM_032391.2 XM_011524747.2

Gene Symbol
PRAC2
Entrez Gene ID
360205
Gene Name
prostate cancer susceptibility candidate 2
Gene Aliases
C17orf93, HOXB-AS5, HOXB13-AS1, NCRNA00253
UniGene
Hs.236557
Gene Chromosome Location
Chr.17: 48720802-48724758 on Build GRCh38
Transcripts
NM_032391.2 XM_011524747.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6147384 c.9C>A p.C3* Chr.17: 48722384-48722384 on Build GRCh38
3958490 c.32C>A p.P11Q Chr.17: 48722361-48722361 on Build GRCh38
707049 c.71A>G p.N24S Chr.17: 48722322-48722322 on Build GRCh38
1221850 c.56G>T p.S19I Chr.17: 48722337-48722337 on Build GRCh38
707048 c.64C>T p.L22F Chr.17: 48722329-48722329 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752510016 Chr.17: 48722351-48722351 on Build GRCh38
rs143323056 Chr.17: 48722388-48722388 on Build GRCh38
rs766533722 Chr.17: 48722352-48722352 on Build GRCh38
rs368233157 Chr.17: 48722379-48722379 on Build GRCh38
rs756253190 Chr.17: 48722278-48722278 on Build GRCh38
rs764728534 Chr.17: 48722318-48722318 on Build GRCh38
rs753077142 Chr.17: 48722603-48722603 on Build GRCh38
rs757521578 Chr.17: 48722274-48722274 on Build GRCh38
rs377467326 Chr.17: 48722330-48722330 on Build GRCh38
rs760966201 Chr.17: 48722427-48722427 on Build GRCh38
rs369643028 Chr.17: 48722277-48722277 on Build GRCh38
rs1040515666 Chr.17: 48722667-48722667 on Build GRCh38
rs777427014 Chr.17: 48722372-48722372 on Build GRCh38
rs546750533 Chr.17: 48722411-48722411 on Build GRCh38
rs373185199 Chr.17: 48722321-48722321 on Build GRCh38
rs750721449 Chr.17: 48722343-48722343 on Build GRCh38
rs917319262 Chr.17: 48722541-48722541 on Build GRCh38
rs1054072 Chr.17: 48722450-48722450 on Build GRCh38
rs759048451 Chr.17: 48722317-48722317 on Build GRCh38
rs767737425 Chr.17: 48722342-48722342 on Build GRCh38
rs554097506 Chr.17: 48722458-48722458 on Build GRCh38
rs995902775 Chr.17: 48722397-48722397 on Build GRCh38
rs200185376 Chr.17: 48722281-48722281 on Build GRCh38
rs936234229 Chr.17: 48722624-48722624 on Build GRCh38
rs778758751 Chr.17: 48722286-48722286 on Build GRCh38
rs754014231 Chr.17: 48722434-48722434 on Build GRCh38
rs955934103 Chr.17: 48722587-48722587 on Build GRCh38
rs889260055 Chr.17: 48722449-48722449 on Build GRCh38
rs756273867 Chr.17: 48722346-48722346 on Build GRCh38
rs188219832 Chr.17: 48722582-48722582 on Build GRCh38
rs1031633464 Chr.17: 48722586-48722586 on Build GRCh38
rs377501304 Chr.17: 48722499-48722499 on Build GRCh38
rs774958586 Chr.17: 48722395-48722395 on Build GRCh38
rs993496704 Chr.17: 48722476-48722476 on Build GRCh38
rs371130765 Chr.17: 48722359-48722359 on Build GRCh38
rs765270447 Chr.17: 48722437-48722437 on Build GRCh38
rs758256018 Chr.17: 48722367-48722367 on Build GRCh38
rs971605374 Chr.17: 48722564-48722564 on Build GRCh38
rs370875142 Chr.17: 48722394-48722394 on Build GRCh38
rs1014866357 Chr.17: 48722445-48722445 on Build GRCh38
rs952188985 Chr.17: 48722500-48722500 on Build GRCh38
rs771717210 Chr.17: 48722297-48722297 on Build GRCh38
rs981361116 Chr.17: 48722580-48722580 on Build GRCh38
rs762282947 Chr.17: 48722335-48722335 on Build GRCh38
rs952222984 Chr.17: 48722263-48722263 on Build GRCh38
rs746613352 Chr.17: 48722309-48722309 on Build GRCh38
rs201787181 Chr.17: 48722358-48722358 on Build GRCh38
rs778248143 Chr.17: 48722666-48722666 on Build GRCh38
rs1055036868 Chr.17: 48722647-48722647 on Build GRCh38
rs898114956 Chr.17: 48722683-48722683 on Build GRCh38
rs758822093 Chr.17: 48722611-48722611 on Build GRCh38
rs565248694 Chr.17: 48722658-48722658 on Build GRCh38
rs897830848 Chr.17: 48722455-48722455 on Build GRCh38
rs776293281 Chr.17: 48722316-48722316 on Build GRCh38
rs756091486 Chr.17: 48722368-48722374 on Build GRCh38
rs574171864 Chr.17: 48722529-48722529 on Build GRCh38
rs117391237 Chr.17: 48722413-48722413 on Build GRCh38
rs769402207 Chr.17: 48722393-48722393 on Build GRCh38
rs751610338 Chr.17: 48722256-48722256 on Build GRCh38
rs973650072 Chr.17: 48722508-48722508 on Build GRCh38
rs745393086 Chr.17: 48722392-48722392 on Build GRCh38
rs368016773 Chr.17: 48722415-48722415 on Build GRCh38
rs766623644 Chr.17: 48722428-48722428 on Build GRCh38
rs34734055 Chr.17: 48722382-48722382 on Build GRCh38
rs772969164 Chr.17: 48722306-48722306 on Build GRCh38
rs576280801 Chr.17: 48722577-48722577 on Build GRCh38
rs971395128 Chr.17: 48722554-48722554 on Build GRCh38
rs754925601 Chr.17: 48722283-48722283 on Build GRCh38
rs778844899 Chr.17: 48722361-48722361 on Build GRCh38
rs748013660 Chr.17: 48722365-48722365 on Build GRCh38
rs932436503 Chr.17: 48722619-48722619 on Build GRCh38
rs1040568191 Chr.17: 48722451-48722451 on Build GRCh38
rs867011313 Chr.17: 48722329-48722329 on Build GRCh38
rs556448054 Chr.17: 48722540-48722540 on Build GRCh38
rs748010346 Chr.17: 48722292-48722292 on Build GRCh38
rs755099370 Chr.17: 48722436-48722436 on Build GRCh38
rs183892842 Chr.17: 48722538-48722538 on Build GRCh38
rs752749334 Chr.17: 48722438-48722438 on Build GRCh38
rs985584245 Chr.17: 48722626-48722626 on Build GRCh38
rs373273135 Chr.17: 48722311-48722311 on Build GRCh38

View Full Product Details