Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 58301122-58301613 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TSPOAP1
Entrez Gene ID
9256
Gene Name
TSPO associated protein 1
Gene Aliases
BZRAP1, PBR-IP, PRAX-1, PRAX1, RIM-BP1, RIMBP1
UniGene
Hs.112499
Gene Chromosome Location
Chr.17: 58301227-58329030 on Build GRCh38
Transcripts
NM_001261835.1 NM_004758.3 NM_024418.2 XM_005257807.2 XM_006722173.2 XM_006722174.2 XM_006722175.2 XM_006722176.2 XM_006722177.3 XM_006722178.3 XM_011525471.2 XM_011525472.1 XM_017025327.1 XM_017025328.1 XM_017025329.1

SNP Detail

SNP ID Chromosome Location
rs757703651 Chr.17: 58301417-58301417 on Build GRCh38
rs1004827713 Chr.17: 58301341-58301341 on Build GRCh38
rs923339660 Chr.17: 58301440-58301440 on Build GRCh38
rs767217233 Chr.17: 58301188-58301188 on Build GRCh38
rs527300838 Chr.17: 58301409-58301409 on Build GRCh38
rs62084577 Chr.17: 58301203-58301203 on Build GRCh38
rs1039451156 Chr.17: 58301432-58301432 on Build GRCh38
rs758916552 Chr.17: 58301169-58301169 on Build GRCh38
rs1009427861 Chr.17: 58301282-58301282 on Build GRCh38
rs1055630000 Chr.17: 58301192-58301192 on Build GRCh38
rs529967637 Chr.17: 58301345-58301345 on Build GRCh38
rs192611285 Chr.17: 58301461-58301461 on Build GRCh38
rs940508639 Chr.17: 58301538-58301538 on Build GRCh38
rs1051391001 Chr.17: 58301261-58301261 on Build GRCh38
rs1053810 Chr.17: 58301400-58301400 on Build GRCh38
rs945253245 Chr.17: 58301519-58301519 on Build GRCh38
rs1053809 Chr.17: 58301420-58301420 on Build GRCh38
rs537440371 Chr.17: 58301416-58301416 on Build GRCh38
rs1011271256 Chr.17: 58301406-58301406 on Build GRCh38
rs532347032 Chr.17: 58301154-58301154 on Build GRCh38
rs569370460 Chr.17: 58301580-58301580 on Build GRCh38
rs551851948 Chr.17: 58301414-58301414 on Build GRCh38
rs1026092523 Chr.17: 58301577-58301577 on Build GRCh38
rs537265988 Chr.17: 58301203-58301203 on Build GRCh38
rs916581972 Chr.17: 58301462-58301462 on Build GRCh38
rs901955842 Chr.17: 58301223-58301223 on Build GRCh38
rs182352353 Chr.17: 58301230-58301230 on Build GRCh38
rs950858716 Chr.17: 58301579-58301579 on Build GRCh38
rs916633898 Chr.17: 58301198-58301198 on Build GRCh38
rs1053811 Chr.17: 58301384-58301384 on Build GRCh38
rs1053239453 Chr.17: 58301475-58301475 on Build GRCh38
rs901723194 Chr.17: 58301569-58301569 on Build GRCh38
rs115986231 Chr.17: 58301360-58301360 on Build GRCh38
rs879282427 Chr.17: 58301500-58301500 on Build GRCh38
rs892788712 Chr.17: 58301588-58301588 on Build GRCh38
rs930744094 Chr.17: 58301473-58301473 on Build GRCh38
rs943373730 Chr.17: 58301170-58301170 on Build GRCh38
rs919433154 Chr.17: 58301407-58301407 on Build GRCh38
rs1050084092 Chr.17: 58301581-58301581 on Build GRCh38
rs901520741 Chr.17: 58301355-58301355 on Build GRCh38
rs779333889 Chr.17: 58301296-58301296 on Build GRCh38
rs1048959398 Chr.17: 58301229-58301229 on Build GRCh38
rs1033782408 Chr.17: 58301546-58301546 on Build GRCh38
rs984990164 Chr.17: 58301587-58301587 on Build GRCh38
rs1022194764 Chr.17: 58301310-58301310 on Build GRCh38
rs907729177 Chr.17: 58301531-58301531 on Build GRCh38
rs950887990 Chr.17: 58301407-58301407 on Build GRCh38
rs931891675 Chr.17: 58301575-58301575 on Build GRCh38
rs1043617468 Chr.17: 58301522-58301522 on Build GRCh38
rs892358121 Chr.17: 58301279-58301279 on Build GRCh38
rs990796165 Chr.17: 58301427-58301427 on Build GRCh38
rs779456190 Chr.17: 58301510-58301510 on Build GRCh38
rs1001838601 Chr.17: 58301544-58301544 on Build GRCh38
rs966830855 Chr.17: 58301413-58301413 on Build GRCh38
rs187836091 Chr.17: 58301350-58301350 on Build GRCh38
rs370410439 Chr.17: 58301589-58301589 on Build GRCh38
rs995824209 Chr.17: 58301359-58301359 on Build GRCh38
rs886261189 Chr.17: 58301322-58301322 on Build GRCh38
rs892929607 Chr.17: 58301390-58301390 on Build GRCh38
rs984856196 Chr.17: 58301528-58301528 on Build GRCh38
rs778012388 Chr.17: 58301562-58301562 on Build GRCh38
rs557495198 Chr.17: 58301574-58301574 on Build GRCh38
rs939342648 Chr.17: 58301185-58301185 on Build GRCh38

View Full Product Details