Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 76137311-76137806 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXJ1
Entrez Gene ID
2302
Gene Name
forkhead box J1
Gene Aliases
FKHL13, HFH-4, HFH4
UniGene
Hs.651204
Gene Chromosome Location
Chr.17: 76136333-76141299 on Build GRCh38
Transcripts
NM_001454.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5581119 c.887C>T p.P296L Chr.17: 76137732-76137732 on Build GRCh38
4069825 c.880C>T p.P294S Chr.17: 76137739-76137739 on Build GRCh38
7368649 c.871A>T p.T291S Chr.17: 76137748-76137748 on Build GRCh38
7327899 c.825A>G p.K275K Chr.17: 76137794-76137794 on Build GRCh38
5364473 c.844G>T p.V282L Chr.17: 76137775-76137775 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs144580544 Chr.17: 76137583-76137583 on Build GRCh38
rs150003378 Chr.17: 76137757-76137757 on Build GRCh38
rs748017781 Chr.17: 76137653-76137653 on Build GRCh38
rs767267314 Chr.17: 76137732-76137732 on Build GRCh38
rs200315041 Chr.17: 76137785-76137786 on Build GRCh38
rs922368569 Chr.17: 76137387-76137387 on Build GRCh38
rs140719410 Chr.17: 76137534-76137534 on Build GRCh38
rs140026938 Chr.17: 76137748-76137748 on Build GRCh38
rs754977794 Chr.17: 76137564-76137564 on Build GRCh38
rs367916894 Chr.17: 76137670-76137670 on Build GRCh38
rs1029058474 Chr.17: 76137482-76137482 on Build GRCh38
rs779517512 Chr.17: 76137559-76137559 on Build GRCh38
rs776458467 Chr.17: 76137514-76137514 on Build GRCh38
rs147874681 Chr.17: 76137668-76137668 on Build GRCh38
rs758017606 Chr.17: 76137618-76137618 on Build GRCh38
rs199916934 Chr.17: 76137777-76137777 on Build GRCh38
rs757962482 Chr.17: 76137557-76137557 on Build GRCh38
rs535872472 Chr.17: 76137699-76137699 on Build GRCh38
rs749050370 Chr.17: 76137667-76137667 on Build GRCh38
rs373797631 Chr.17: 76137392-76137392 on Build GRCh38
rs768139287 Chr.17: 76137396-76137396 on Build GRCh38
rs756148428 Chr.17: 76137759-76137759 on Build GRCh38
rs939411767 Chr.17: 76137523-76137523 on Build GRCh38
rs374989646 Chr.17: 76137760-76137760 on Build GRCh38
rs202087312 Chr.17: 76137604-76137604 on Build GRCh38
rs751724814 Chr.17: 76137391-76137391 on Build GRCh38
rs777253568 Chr.17: 76137673-76137673 on Build GRCh38
rs756963248 Chr.17: 76137467-76137467 on Build GRCh38
rs981384883 Chr.17: 76137656-76137656 on Build GRCh38
rs763257288 Chr.17: 76137731-76137731 on Build GRCh38
rs996737465 Chr.17: 76137772-76137772 on Build GRCh38
rs774138609 Chr.17: 76137475-76137475 on Build GRCh38
rs751106371 Chr.17: 76137620-76137620 on Build GRCh38
rs753247265 Chr.17: 76137416-76137416 on Build GRCh38
rs754391541 Chr.17: 76137645-76137645 on Build GRCh38
rs530057825 Chr.17: 76137389-76137389 on Build GRCh38
rs3192447 Chr.17: 76137788-76137788 on Build GRCh38
rs759753578 Chr.17: 76137524-76137524 on Build GRCh38
rs756020281 Chr.17: 76137655-76137655 on Build GRCh38
rs908179650 Chr.17: 76137458-76137458 on Build GRCh38
rs1010068633 Chr.17: 76137430-76137430 on Build GRCh38
rs940729815 Chr.17: 76137585-76137585 on Build GRCh38
rs764465961 Chr.17: 76137538-76137538 on Build GRCh38
rs550111178 Chr.17: 76137431-76137431 on Build GRCh38
rs761478708 Chr.17: 76137516-76137516 on Build GRCh38
rs1052436042 Chr.17: 76137527-76137527 on Build GRCh38
rs551252596 Chr.17: 76137537-76137537 on Build GRCh38
rs778173571 Chr.17: 76137451-76137451 on Build GRCh38
rs368509818 Chr.17: 76137573-76137573 on Build GRCh38
rs771305310 Chr.17: 76137517-76137517 on Build GRCh38
rs768830919 Chr.17: 76137778-76137778 on Build GRCh38
rs956646426 Chr.17: 76137345-76137345 on Build GRCh38
rs769704897 Chr.17: 76137569-76137569 on Build GRCh38
rs975207311 Chr.17: 76137380-76137380 on Build GRCh38
rs746963003 Chr.17: 76137561-76137561 on Build GRCh38
rs768387796 Chr.17: 76137508-76137508 on Build GRCh38
rs561343952 Chr.17: 76137356-76137356 on Build GRCh38
rs973747724 Chr.17: 76137640-76137640 on Build GRCh38
rs372348253 Chr.17: 76137596-76137596 on Build GRCh38
rs757505002 Chr.17: 76137540-76137540 on Build GRCh38
rs751110542 Chr.17: 76137738-76137738 on Build GRCh38
rs773365878 Chr.17: 76137365-76137365 on Build GRCh38
rs765575443 Chr.17: 76137617-76137617 on Build GRCh38
rs375395677 Chr.17: 76137572-76137572 on Build GRCh38
rs764875027 Chr.17: 76137545-76137545 on Build GRCh38
rs780390414 Chr.17: 76137773-76137773 on Build GRCh38
rs371952924 Chr.17: 76137723-76137723 on Build GRCh38
rs575937592 Chr.17: 76137761-76137761 on Build GRCh38
rs754251901 Chr.17: 76137539-76137539 on Build GRCh38
rs780034970 Chr.17: 76137490-76137490 on Build GRCh38
rs545716107 Chr.17: 76137769-76137769 on Build GRCh38
rs147424553 Chr.17: 76137503-76137503 on Build GRCh38
rs761785819 Chr.17: 76137546-76137546 on Build GRCh38
rs777676324 Chr.17: 76137661-76137661 on Build GRCh38
rs760391614 Chr.17: 76137334-76137334 on Build GRCh38
rs767689240 Chr.17: 76137533-76137533 on Build GRCh38
rs962332266 Chr.17: 76137619-76137619 on Build GRCh38
rs760620567 Chr.17: 76137595-76137595 on Build GRCh38
rs571143288 Chr.17: 76137568-76137568 on Build GRCh38
rs770185869 Chr.17: 76137681-76137681 on Build GRCh38
rs569060949 Chr.17: 76137518-76137518 on Build GRCh38
rs200569506 Chr.17: 76137571-76137571 on Build GRCh38
rs771504245 Chr.17: 76137485-76137485 on Build GRCh38
rs762377959 Chr.17: 76137370-76137370 on Build GRCh38
rs745397559 Chr.17: 76137480-76137480 on Build GRCh38
rs762232230 Chr.17: 76137605-76137605 on Build GRCh38
rs367598262 Chr.17: 76137452-76137452 on Build GRCh38
rs376918102 Chr.17: 76137646-76137646 on Build GRCh38
rs750071975 Chr.17: 76137552-76137552 on Build GRCh38
rs768614679 Chr.17: 76137671-76137671 on Build GRCh38

View Full Product Details