Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 78224126-78224629 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BIRC5
Entrez Gene ID
332
Gene Name
baculoviral IAP repeat containing 5
Gene Aliases
API4, EPR-1
UniGene
Hs.744872
Gene Chromosome Location
Chr.17: 78214196-78225635 on Build GRCh38
Transcripts
NM_001012270.1 NM_001012271.1 NM_001168.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5794977 c.213C>T p.S71S Chr.17: 78224443-78224443 on Build GRCh38
5783275 c.158C>T p.P53L Chr.17: 78224498-78224498 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs2856269 Chr.17: 78224556-78224556 on Build GRCh38
rs963096671 Chr.17: 78224426-78224426 on Build GRCh38
rs540147173 Chr.17: 78224527-78224527 on Build GRCh38
rs950701086 Chr.17: 78224542-78224542 on Build GRCh38
rs895013059 Chr.17: 78224498-78224498 on Build GRCh38
rs1053538596 Chr.17: 78224308-78224308 on Build GRCh38
rs181761491 Chr.17: 78224487-78224487 on Build GRCh38
rs1026250789 Chr.17: 78224540-78224540 on Build GRCh38
rs150652785 Chr.17: 78224528-78224528 on Build GRCh38
rs950616764 Chr.17: 78224415-78224415 on Build GRCh38
rs191250920 Chr.17: 78224363-78224363 on Build GRCh38
rs900576494 Chr.17: 78224348-78224348 on Build GRCh38
rs777306171 Chr.17: 78224246-78224246 on Build GRCh38
rs990560274 Chr.17: 78224184-78224184 on Build GRCh38
rs1012093533 Chr.17: 78224338-78224338 on Build GRCh38
rs758812176 Chr.17: 78224251-78224251 on Build GRCh38
rs902447228 Chr.17: 78224444-78224444 on Build GRCh38
rs907503797 Chr.17: 78224275-78224275 on Build GRCh38
rs963449326 Chr.17: 78224270-78224270 on Build GRCh38
rs946408509 Chr.17: 78224425-78224425 on Build GRCh38
rs1003483061 Chr.17: 78224476-78224476 on Build GRCh38
rs947440223 Chr.17: 78224538-78224538 on Build GRCh38
rs915007427 Chr.17: 78224607-78224607 on Build GRCh38
rs886248212 Chr.17: 78224581-78224581 on Build GRCh38
rs748880179 Chr.17: 78224242-78224242 on Build GRCh38
rs997564367 Chr.17: 78224571-78224571 on Build GRCh38
rs949137157 Chr.17: 78224502-78224502 on Build GRCh38
rs981576831 Chr.17: 78224508-78224508 on Build GRCh38
rs914973884 Chr.17: 78224420-78224420 on Build GRCh38
rs990876839 Chr.17: 78224486-78224486 on Build GRCh38
rs762447826 Chr.17: 78224336-78224336 on Build GRCh38
rs1045868477 Chr.17: 78224254-78224254 on Build GRCh38
rs919517164 Chr.17: 78224345-78224345 on Build GRCh38
rs1030180514 Chr.17: 78224579-78224579 on Build GRCh38
rs533200033 Chr.17: 78224497-78224497 on Build GRCh38
rs542205905 Chr.17: 78224306-78224306 on Build GRCh38
rs987713248 Chr.17: 78224546-78224546 on Build GRCh38
rs1005104267 Chr.17: 78224592-78224592 on Build GRCh38
rs1013030310 Chr.17: 78224243-78224243 on Build GRCh38
rs771381579 Chr.17: 78224516-78224516 on Build GRCh38
rs377708686 Chr.17: 78224533-78224533 on Build GRCh38
rs1039441081 Chr.17: 78224548-78224548 on Build GRCh38
rs912086476 Chr.17: 78224403-78224403 on Build GRCh38
rs912140353 Chr.17: 78224550-78224550 on Build GRCh38
rs997387693 Chr.17: 78224351-78224351 on Build GRCh38
rs983356888 Chr.17: 78224417-78224417 on Build GRCh38
rs928772680 Chr.17: 78224509-78224509 on Build GRCh38
rs138703176 Chr.17: 78224290-78224290 on Build GRCh38
rs893108510 Chr.17: 78224314-78224314 on Build GRCh38
rs779690467 Chr.17: 78224230-78224230 on Build GRCh38
rs17878624 Chr.17: 78224418-78224418 on Build GRCh38
rs548543992 Chr.17: 78224582-78224582 on Build GRCh38
rs79824097 Chr.17: 78224347-78224347 on Build GRCh38
rs776591534 Chr.17: 78224555-78224555 on Build GRCh38

View Full Product Details