Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 79779256-79779761 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CBX2
Entrez Gene ID
84733
Gene Name
chromobox 2
Gene Aliases
CDCA6, M33, SRXY5
UniGene
Hs.368410
Gene Chromosome Location
Chr.17: 79776254-79787650 on Build GRCh38
Transcripts
NM_005189.2 NM_032647.3 XM_011525382.1 XM_011525383.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6476590 c.156G>A p.P52P Chr.17: 79779401-79779401 on Build GRCh38
6476589 c.156G>A p.P52P Chr.17: 79779401-79779401 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs937349915 Chr.17: 79779317-79779317 on Build GRCh38
rs371788511 Chr.17: 79779326-79779326 on Build GRCh38
rs781974938 Chr.17: 79779720-79779720 on Build GRCh38
rs905918981 Chr.17: 79779301-79779301 on Build GRCh38
rs143293175 Chr.17: 79779446-79779447 on Build GRCh38
rs782720069 Chr.17: 79779693-79779693 on Build GRCh38
rs536388623 Chr.17: 79779673-79779673 on Build GRCh38
rs981542428 Chr.17: 79779532-79779532 on Build GRCh38
rs767112450 Chr.17: 79779449-79779449 on Build GRCh38
rs782567355 Chr.17: 79779471-79779471 on Build GRCh38
rs557052010 Chr.17: 79779404-79779404 on Build GRCh38
rs953811416 Chr.17: 79779453-79779453 on Build GRCh38
rs531112487 Chr.17: 79779721-79779722 on Build GRCh38
rs782001998 Chr.17: 79779415-79779415 on Build GRCh38
rs913429691 Chr.17: 79779277-79779277 on Build GRCh38
rs144741480 Chr.17: 79779641-79779641 on Build GRCh38
rs950191804 Chr.17: 79779284-79779284 on Build GRCh38
rs377624331 Chr.17: 79779676-79779676 on Build GRCh38
rs762839701 Chr.17: 79779543-79779543 on Build GRCh38
rs559224039 Chr.17: 79779732-79779732 on Build GRCh38
rs782154021 Chr.17: 79779435-79779435 on Build GRCh38
rs957078540 Chr.17: 79779617-79779617 on Build GRCh38
rs112582183 Chr.17: 79779689-79779689 on Build GRCh38
rs35124694 Chr.17: 79779392-79779392 on Build GRCh38
rs544649796 Chr.17: 79779584-79779584 on Build GRCh38
rs889607774 Chr.17: 79779605-79779605 on Build GRCh38
rs139072089 Chr.17: 79779411-79779411 on Build GRCh38
rs782348829 Chr.17: 79779363-79779363 on Build GRCh38
rs577663744 Chr.17: 79779508-79779508 on Build GRCh38
rs940656657 Chr.17: 79779583-79779583 on Build GRCh38
rs782458687 Chr.17: 79779446-79779446 on Build GRCh38
rs998095946 Chr.17: 79779588-79779588 on Build GRCh38
rs563374366 Chr.17: 79779586-79779586 on Build GRCh38
rs541210925 Chr.17: 79779469-79779470 on Build GRCh38
rs781804768 Chr.17: 79779298-79779298 on Build GRCh38
rs782549730 Chr.17: 79779470-79779470 on Build GRCh38
rs782524512 Chr.17: 79779478-79779478 on Build GRCh38
rs373265960 Chr.17: 79779348-79779348 on Build GRCh38
rs887810342 Chr.17: 79779559-79779559 on Build GRCh38
rs782705919 Chr.17: 79779316-79779316 on Build GRCh38
rs782164371 Chr.17: 79779401-79779401 on Build GRCh38
rs114269401 Chr.17: 79779371-79779371 on Build GRCh38
rs1049594124 Chr.17: 79779548-79779548 on Build GRCh38
rs3833851 Chr.17: 79779448-79779449 on Build GRCh38
rs554705254 Chr.17: 79779468-79779468 on Build GRCh38
rs558965663 Chr.17: 79779533-79779533 on Build GRCh38
rs782630951 Chr.17: 79779490-79779490 on Build GRCh38
rs3751953 Chr.17: 79779643-79779643 on Build GRCh38
rs3751954 Chr.17: 79779686-79779686 on Build GRCh38
rs782540087 Chr.17: 79779503-79779503 on Build GRCh38
rs782626585 Chr.17: 79779342-79779342 on Build GRCh38
rs8064780 Chr.17: 79779497-79779497 on Build GRCh38
rs782017451 Chr.17: 79779393-79779393 on Build GRCh38
rs118059543 Chr.17: 79779670-79779670 on Build GRCh38
rs782488786 Chr.17: 79779452-79779452 on Build GRCh38
rs782269541 Chr.17: 79779464-79779464 on Build GRCh38
rs781895024 Chr.17: 79779346-79779346 on Build GRCh38
rs187683130 Chr.17: 79779632-79779632 on Build GRCh38
rs782345389 Chr.17: 79779479-79779479 on Build GRCh38
rs782023555 Chr.17: 79779677-79779677 on Build GRCh38
rs79637552 Chr.17: 79779613-79779613 on Build GRCh38
rs546905963 Chr.17: 79779636-79779636 on Build GRCh38
rs782568075 Chr.17: 79779509-79779509 on Build GRCh38
rs528423707 Chr.17: 79779313-79779313 on Build GRCh38
rs782198883 Chr.17: 79779351-79779351 on Build GRCh38
rs781831874 Chr.17: 79779451-79779451 on Build GRCh38
rs957250016 Chr.17: 79779495-79779495 on Build GRCh38
rs577602621 Chr.17: 79779560-79779560 on Build GRCh38
rs137941885 Chr.17: 79779407-79779407 on Build GRCh38
rs369726383 Chr.17: 79779469-79779469 on Build GRCh38
rs377482051 Chr.17: 79779349-79779349 on Build GRCh38
rs569021153 Chr.17: 79779671-79779671 on Build GRCh38
rs369460211 Chr.17: 79779339-79779339 on Build GRCh38
rs370977173 Chr.17: 79779462-79779462 on Build GRCh38
rs573093068 Chr.17: 79779486-79779486 on Build GRCh38
rs782814466 Chr.17: 79779436-79779436 on Build GRCh38
rs530580805 Chr.17: 79779604-79779604 on Build GRCh38
rs782236889 Chr.17: 79779387-79779387 on Build GRCh38
rs182204523 Chr.17: 79779616-79779616 on Build GRCh38
rs528248433 Chr.17: 79779631-79779631 on Build GRCh38

View Full Product Details