Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 80182517-80183013 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CARD14
Entrez Gene ID
79092
Gene Name
caspase recruitment domain family member 14
Gene Aliases
BIMP2, CARMA2, PRP, PSORS2, PSS1
UniGene
Hs.675480
Gene Chromosome Location
Chr.17: 80169992-80209331 on Build GRCh38
Transcripts
NM_001257970.1 NM_024110.4 NR_047566.1 XM_011525212.1 XM_011525213.1 XM_011525215.1 XM_011525216.1 XM_011525217.1 XM_011525218.2 XM_017025057.1 XM_017025058.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6620111 c.250G>A p.G84R Chr.17: 80182691-80182691 on Build GRCh38
6025990 c.228G>A p.L76L Chr.17: 80182669-80182669 on Build GRCh38
2803238 c.295G>A p.V99I Chr.17: 80182736-80182736 on Build GRCh38
3937641 c.212-2A>G p.? Chr.17: 80182651-80182651 on Build GRCh38
6620112 c.307G>A p.V103I Chr.17: 80182748-80182748 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768362011 Chr.17: 80182753-80182753 on Build GRCh38
rs753451217 Chr.17: 80182609-80182609 on Build GRCh38
rs1052382077 Chr.17: 80182954-80182954 on Build GRCh38
rs920737868 Chr.17: 80182577-80182577 on Build GRCh38
rs138139140 Chr.17: 80182770-80182770 on Build GRCh38
rs1050484901 Chr.17: 80182592-80182592 on Build GRCh38
rs868437671 Chr.17: 80182978-80182978 on Build GRCh38
rs757364434 Chr.17: 80182735-80182735 on Build GRCh38
rs143747620 Chr.17: 80182675-80182675 on Build GRCh38
rs776318698 Chr.17: 80182810-80182810 on Build GRCh38
rs200725256 Chr.17: 80182634-80182634 on Build GRCh38
rs760057303 Chr.17: 80182782-80182782 on Build GRCh38
rs112948236 Chr.17: 80182598-80182598 on Build GRCh38
rs769368124 Chr.17: 80182766-80182766 on Build GRCh38
rs776425179 Chr.17: 80182754-80182754 on Build GRCh38
rs281875215 Chr.17: 80182790-80182790 on Build GRCh38
rs547248874 Chr.17: 80182580-80182580 on Build GRCh38
rs903653101 Chr.17: 80182694-80182694 on Build GRCh38
rs1014252436 Chr.17: 80182767-80182767 on Build GRCh38
rs975850621 Chr.17: 80182579-80182580 on Build GRCh38
rs375520842 Chr.17: 80182706-80182706 on Build GRCh38
rs778679607 Chr.17: 80182736-80182736 on Build GRCh38
rs746855162 Chr.17: 80182750-80182750 on Build GRCh38
rs552779505 Chr.17: 80182740-80182740 on Build GRCh38
rs569560613 Chr.17: 80182663-80182663 on Build GRCh38
rs377316099 Chr.17: 80182679-80182679 on Build GRCh38
rs528692199 Chr.17: 80182547-80182547 on Build GRCh38
rs373005620 Chr.17: 80182708-80182708 on Build GRCh38
rs370538076 Chr.17: 80182620-80182620 on Build GRCh38
rs193262780 Chr.17: 80182691-80182691 on Build GRCh38
rs751810450 Chr.17: 80182631-80182631 on Build GRCh38
rs745585891 Chr.17: 80182667-80182667 on Build GRCh38
rs760384971 Chr.17: 80182677-80182677 on Build GRCh38
rs9902358 Chr.17: 80182963-80182963 on Build GRCh38
rs368821179 Chr.17: 80182758-80182758 on Build GRCh38
rs949671749 Chr.17: 80182875-80182875 on Build GRCh38
rs753859758 Chr.17: 80182729-80182729 on Build GRCh38
rs561501269 Chr.17: 80182543-80182543 on Build GRCh38
rs188854006 Chr.17: 80182874-80182874 on Build GRCh38
rs769219418 Chr.17: 80182934-80182934 on Build GRCh38
rs773795280 Chr.17: 80182864-80182864 on Build GRCh38
rs771934781 Chr.17: 80182671-80182671 on Build GRCh38
rs1052713843 Chr.17: 80182619-80182619 on Build GRCh38
rs879084943 Chr.17: 80182566-80182566 on Build GRCh38
rs543268620 Chr.17: 80182863-80182863 on Build GRCh38
rs886041402 Chr.17: 80182791-80182791 on Build GRCh38
rs777801068 Chr.17: 80182637-80182637 on Build GRCh38
rs763724128 Chr.17: 80182607-80182607 on Build GRCh38
rs551385923 Chr.17: 80182657-80182657 on Build GRCh38
rs1001996592 Chr.17: 80182798-80182798 on Build GRCh38
rs778027398 Chr.17: 80182718-80182718 on Build GRCh38
rs905380899 Chr.17: 80182918-80182918 on Build GRCh38
rs761418397 Chr.17: 80182684-80182684 on Build GRCh38
rs770762780 Chr.17: 80182652-80182652 on Build GRCh38
rs965706147 Chr.17: 80182860-80182860 on Build GRCh38
rs553260220 Chr.17: 80182915-80182915 on Build GRCh38
rs148170776 Chr.17: 80182690-80182690 on Build GRCh38
rs903539440 Chr.17: 80182597-80182601 on Build GRCh38
rs372257600 Chr.17: 80182786-80182786 on Build GRCh38
rs767900787 Chr.17: 80182692-80182692 on Build GRCh38
rs760225367 Chr.17: 80182604-80182604 on Build GRCh38
rs587777763 Chr.17: 80182795-80182795 on Build GRCh38
rs766963929 Chr.17: 80182628-80182628 on Build GRCh38
rs1043829364 Chr.17: 80182887-80182887 on Build GRCh38
rs191713350 Chr.17: 80182943-80182943 on Build GRCh38
rs538754535 Chr.17: 80182801-80182801 on Build GRCh38
rs771956573 Chr.17: 80182743-80182743 on Build GRCh38
rs750512409 Chr.17: 80182811-80182811 on Build GRCh38
rs371578752 Chr.17: 80182809-80182809 on Build GRCh38
rs185132539 Chr.17: 80182856-80182856 on Build GRCh38
rs757238045 Chr.17: 80182807-80182807 on Build GRCh38
rs189845028 Chr.17: 80182583-80182583 on Build GRCh38
rs758339735 Chr.17: 80182816-80182816 on Build GRCh38
rs775819149 Chr.17: 80182789-80182789 on Build GRCh38
rs749438022 Chr.17: 80182638-80182638 on Build GRCh38
rs367774865 Chr.17: 80182806-80182806 on Build GRCh38
rs370897817 Chr.17: 80182700-80182700 on Build GRCh38
rs182274963 Chr.17: 80182683-80182683 on Build GRCh38
rs147122239 Chr.17: 80182699-80182699 on Build GRCh38
rs755372322 Chr.17: 80182633-80182633 on Build GRCh38
rs888440146 Chr.17: 80182982-80182982 on Build GRCh38
rs983701162 Chr.17: 80182847-80182847 on Build GRCh38
rs373598742 Chr.17: 80182802-80182802 on Build GRCh38
rs779967952 Chr.17: 80182747-80182747 on Build GRCh38
rs752936293 Chr.17: 80182636-80182636 on Build GRCh38

View Full Product Details