Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 80195093-80195585 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CARD14
Entrez Gene ID
79092
Gene Name
caspase recruitment domain family member 14
Gene Aliases
BIMP2, CARMA2, PRP, PSORS2, PSS1
UniGene
Hs.675480
Gene Chromosome Location
Chr.17: 80169992-80209331 on Build GRCh38
Transcripts
NM_001257970.1 NM_024110.4 NM_052819.2 NR_047566.1 XM_011525212.1 XM_011525213.1 XM_011525215.1 XM_011525216.1 XM_011525217.1 XM_011525218.2 XM_017025057.1 XM_017025058.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3970422 c.668G>A p.S223N Chr.17: 80195213-80195213 on Build GRCh38
1303462 c.1430C>T p.A477V Chr.17: 80195264-80195264 on Build GRCh38
3388273 c.1386G>A p.T462T Chr.17: 80195220-80195220 on Build GRCh38
985560 c.1470G>T p.E490D Chr.17: 80195304-80195304 on Build GRCh38
6620099 c.807G>A p.P269P Chr.17: 80195576-80195576 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs896282788 Chr.17: 80195361-80195361 on Build GRCh38
rs368797054 Chr.17: 80195483-80195483 on Build GRCh38
rs368239982 Chr.17: 80195219-80195219 on Build GRCh38
rs373552852 Chr.17: 80195157-80195157 on Build GRCh38
rs771831115 Chr.17: 80195354-80195354 on Build GRCh38
rs751443194 Chr.17: 80195217-80195217 on Build GRCh38
rs775971131 Chr.17: 80195184-80195184 on Build GRCh38
rs974693576 Chr.17: 80195444-80195444 on Build GRCh38
rs757704864 Chr.17: 80195162-80195162 on Build GRCh38
rs773256259 Chr.17: 80195211-80195211 on Build GRCh38
rs760316201 Chr.17: 80195512-80195512 on Build GRCh38
rs775320526 Chr.17: 80195363-80195363 on Build GRCh38
rs556359326 Chr.17: 80195529-80195529 on Build GRCh38
rs758112012 Chr.17: 80195446-80195446 on Build GRCh38
rs746504878 Chr.17: 80195531-80195531 on Build GRCh38
rs759000751 Chr.17: 80195229-80195229 on Build GRCh38
rs566366503 Chr.17: 80195128-80195128 on Build GRCh38
rs374170794 Chr.17: 80195281-80195281 on Build GRCh38
rs367763902 Chr.17: 80195159-80195159 on Build GRCh38
rs199751229 Chr.17: 80195158-80195158 on Build GRCh38
rs780621356 Chr.17: 80195288-80195288 on Build GRCh38
rs758760584 Chr.17: 80195279-80195279 on Build GRCh38
rs768698549 Chr.17: 80195244-80195244 on Build GRCh38
rs776782295 Chr.17: 80195204-80195204 on Build GRCh38
rs377069416 Chr.17: 80195563-80195563 on Build GRCh38
rs62074378 Chr.17: 80195205-80195205 on Build GRCh38
rs201580571 Chr.17: 80195153-80195153 on Build GRCh38
rs140734867 Chr.17: 80195230-80195230 on Build GRCh38
rs766077998 Chr.17: 80195214-80195214 on Build GRCh38
rs199793195 Chr.17: 80195192-80195192 on Build GRCh38
rs777303119 Chr.17: 80195227-80195227 on Build GRCh38
rs746351148 Chr.17: 80195228-80195228 on Build GRCh38
rs994010684 Chr.17: 80195277-80195277 on Build GRCh38
rs1046703469 Chr.17: 80195458-80195458 on Build GRCh38
rs201906803 Chr.17: 80195339-80195339 on Build GRCh38
rs114086314 Chr.17: 80195346-80195346 on Build GRCh38
rs374847486 Chr.17: 80195311-80195311 on Build GRCh38
rs758784694 Chr.17: 80195200-80195201 on Build GRCh38
rs372805579 Chr.17: 80195302-80195302 on Build GRCh38
rs138388900 Chr.17: 80195387-80195387 on Build GRCh38
rs755654559 Chr.17: 80195222-80195222 on Build GRCh38
rs547954709 Chr.17: 80195408-80195408 on Build GRCh38
rs753454750 Chr.17: 80195276-80195276 on Build GRCh38
rs141045409 Chr.17: 80195199-80195199 on Build GRCh38
rs763000467 Chr.17: 80195212-80195212 on Build GRCh38
rs546212263 Chr.17: 80195264-80195264 on Build GRCh38
rs145876317 Chr.17: 80195248-80195248 on Build GRCh38
rs146882682 Chr.17: 80195220-80195220 on Build GRCh38
rs747400015 Chr.17: 80195235-80195235 on Build GRCh38
rs200592374 Chr.17: 80195196-80195196 on Build GRCh38
rs368432283 Chr.17: 80195262-80195262 on Build GRCh38
rs142981600 Chr.17: 80195434-80195434 on Build GRCh38
rs144710573 Chr.17: 80195310-80195310 on Build GRCh38
rs992126850 Chr.17: 80195548-80195548 on Build GRCh38
rs780143594 Chr.17: 80195265-80195266 on Build GRCh38
rs975397775 Chr.17: 80195131-80195131 on Build GRCh38
rs377226624 Chr.17: 80195249-80195249 on Build GRCh38
rs200960057 Chr.17: 80195564-80195564 on Build GRCh38
rs987526333 Chr.17: 80195115-80195115 on Build GRCh38
rs1008444664 Chr.17: 80195466-80195466 on Build GRCh38
rs968069724 Chr.17: 80195454-80195454 on Build GRCh38
rs777937743 Chr.17: 80195313-80195313 on Build GRCh38
rs759434721 Chr.17: 80195350-80195350 on Build GRCh38
rs947718596 Chr.17: 80195494-80195494 on Build GRCh38
rs758287781 Chr.17: 80195154-80195154 on Build GRCh38
rs914539876 Chr.17: 80195142-80195142 on Build GRCh38
rs370393239 Chr.17: 80195178-80195178 on Build GRCh38
rs564427787 Chr.17: 80195265-80195265 on Build GRCh38
rs145139255 Chr.17: 80195492-80195492 on Build GRCh38
rs939643406 Chr.17: 80195480-80195480 on Build GRCh38
rs747661632 Chr.17: 80195161-80195161 on Build GRCh38
rs768649469 Chr.17: 80195354-80195355 on Build GRCh38
rs752314467 Chr.17: 80195270-80195270 on Build GRCh38
rs149030007 Chr.17: 80195305-80195305 on Build GRCh38
rs978261519 Chr.17: 80195461-80195461 on Build GRCh38
rs533545806 Chr.17: 80195455-80195455 on Build GRCh38
rs118144449 Chr.17: 80195468-80195468 on Build GRCh38
rs574706437 Chr.17: 80195547-80195547 on Build GRCh38
rs372422292 Chr.17: 80195498-80195498 on Build GRCh38
rs572325893 Chr.17: 80195263-80195263 on Build GRCh38
rs762798473 Chr.17: 80195260-80195260 on Build GRCh38
rs763580623 Chr.17: 80195272-80195272 on Build GRCh38
rs924603303 Chr.17: 80195516-80195516 on Build GRCh38
rs747205873 Chr.17: 80195339-80195367 on Build GRCh38
rs868816517 Chr.17: 80195378-80195378 on Build GRCh38
rs772291852 Chr.17: 80195181-80195181 on Build GRCh38
rs947864785 Chr.17: 80195402-80195402 on Build GRCh38
rs1038009652 Chr.17: 80195497-80195497 on Build GRCh38
rs369200145 Chr.17: 80195333-80195333 on Build GRCh38
rs112639680 Chr.17: 80195190-80195190 on Build GRCh38
rs964215715 Chr.17: 80195474-80195474 on Build GRCh38
rs1044869470 Chr.17: 80195421-80195421 on Build GRCh38
rs531888447 Chr.17: 80195294-80195294 on Build GRCh38
rs137954126 Chr.17: 80195256-80195256 on Build GRCh38
rs761404555 Chr.17: 80195534-80195534 on Build GRCh38
rs779419388 Chr.17: 80195174-80195174 on Build GRCh38

View Full Product Details