Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 82035790-82036286 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCXR
Entrez Gene ID
51181
Gene Name
dicarbonyl and L-xylulose reductase
Gene Aliases
DCR, HCR2, HCRII, KIDCR, P34H, PNTSU, SDR20C1, XR
UniGene
Hs.9857
Gene Chromosome Location
Chr.17: 82035881-82037697 on Build GRCh38
Transcripts
NM_001195218.1 NM_016286.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4611566 c.583delC p.H195fs*7 Chr.17: 82036239-82036239 on Build GRCh38
4793571 c.726G>A p.W242* Chr.17: 82035969-82035969 on Build GRCh38
473555 c.683G>A p.G228D Chr.17: 82036012-82036012 on Build GRCh38
1162295 c.539C>G p.T180R Chr.17: 82036283-82036283 on Build GRCh38
5966507 c.718G>A p.G240S Chr.17: 82035977-82035977 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762256289 Chr.17: 82035914-82035914 on Build GRCh38
rs565838532 Chr.17: 82035926-82035926 on Build GRCh38
rs367666668 Chr.17: 82035959-82035959 on Build GRCh38
rs557857160 Chr.17: 82035930-82035930 on Build GRCh38
rs760528025 Chr.17: 82036239-82036239 on Build GRCh38
rs776988993 Chr.17: 82036176-82036176 on Build GRCh38
rs940396241 Chr.17: 82035878-82035878 on Build GRCh38
rs35000932 Chr.17: 82036147-82036148 on Build GRCh38
rs771377752 Chr.17: 82036109-82036109 on Build GRCh38
rs549839866 Chr.17: 82035873-82035873 on Build GRCh38
rs899163272 Chr.17: 82035895-82035895 on Build GRCh38
rs1009552678 Chr.17: 82036235-82036235 on Build GRCh38
rs745666336 Chr.17: 82036053-82036053 on Build GRCh38
rs1057378543 Chr.17: 82035838-82035838 on Build GRCh38
rs746741996 Chr.17: 82035954-82035954 on Build GRCh38
rs762048891 Chr.17: 82036001-82036001 on Build GRCh38
rs780166795 Chr.17: 82036064-82036064 on Build GRCh38
rs372716031 Chr.17: 82036164-82036164 on Build GRCh38
rs376667940 Chr.17: 82036165-82036165 on Build GRCh38
rs766875180 Chr.17: 82036007-82036007 on Build GRCh38
rs753717407 Chr.17: 82036242-82036242 on Build GRCh38
rs1025044689 Chr.17: 82036041-82036041 on Build GRCh38
rs768816595 Chr.17: 82036077-82036077 on Build GRCh38
rs770796308 Chr.17: 82036040-82036040 on Build GRCh38
rs761236094 Chr.17: 82036003-82036003 on Build GRCh38
rs747937505 Chr.17: 82036238-82036239 on Build GRCh38
rs758555409 Chr.17: 82036208-82036208 on Build GRCh38
rs1127805 Chr.17: 82036221-82036221 on Build GRCh38
rs769301193 Chr.17: 82036255-82036255 on Build GRCh38
rs754870898 Chr.17: 82036084-82036085 on Build GRCh38
rs761487390 Chr.17: 82036125-82036125 on Build GRCh38
rs1009697867 Chr.17: 82036243-82036243 on Build GRCh38
rs543002005 Chr.17: 82036233-82036233 on Build GRCh38
rs552483270 Chr.17: 82035994-82036002 on Build GRCh38
rs563366492 Chr.17: 82035851-82035851 on Build GRCh38
rs749263479 Chr.17: 82035908-82035908 on Build GRCh38
rs575588182 Chr.17: 82036178-82036178 on Build GRCh38
rs781201690 Chr.17: 82036047-82036047 on Build GRCh38
rs530910289 Chr.17: 82035852-82035852 on Build GRCh38
rs529120944 Chr.17: 82035896-82035896 on Build GRCh38
rs1001417646 Chr.17: 82036052-82036052 on Build GRCh38
rs1127822 Chr.17: 82036031-82036031 on Build GRCh38
rs751428650 Chr.17: 82036071-82036072 on Build GRCh38
rs923418925 Chr.17: 82035894-82035894 on Build GRCh38
rs995249397 Chr.17: 82035907-82035907 on Build GRCh38
rs112488192 Chr.17: 82036065-82036065 on Build GRCh38
rs372628326 Chr.17: 82036028-82036028 on Build GRCh38
rs368210662 Chr.17: 82036044-82036044 on Build GRCh38
rs372313006 Chr.17: 82036148-82036148 on Build GRCh38
rs779977716 Chr.17: 82036032-82036033 on Build GRCh38
rs778978014 Chr.17: 82036086-82036086 on Build GRCh38
rs778730818 Chr.17: 82036256-82036256 on Build GRCh38
rs766258659 Chr.17: 82036240-82036240 on Build GRCh38
rs747887427 Chr.17: 82036239-82036239 on Build GRCh38
rs780839835 Chr.17: 82036185-82036186 on Build GRCh38
rs767915882 Chr.17: 82036002-82036002 on Build GRCh38
rs761040124 Chr.17: 82036104-82036104 on Build GRCh38
rs547617107 Chr.17: 82035919-82035919 on Build GRCh38
rs771332809 Chr.17: 82036175-82036175 on Build GRCh38
rs768716624 Chr.17: 82035989-82035989 on Build GRCh38
rs927269306 Chr.17: 82035831-82035831 on Build GRCh38
rs750190857 Chr.17: 82036138-82036138 on Build GRCh38
rs748458085 Chr.17: 82036095-82036095 on Build GRCh38
rs147305616 Chr.17: 82035987-82035987 on Build GRCh38
rs767968976 Chr.17: 82035915-82035915 on Build GRCh38
rs200507677 Chr.17: 82036100-82036100 on Build GRCh38
rs201250675 Chr.17: 82036214-82036214 on Build GRCh38
rs745860696 Chr.17: 82035963-82035963 on Build GRCh38
rs6502047 Chr.17: 82036150-82036150 on Build GRCh38
rs147968086 Chr.17: 82036215-82036215 on Build GRCh38
rs752090622 Chr.17: 82036018-82036018 on Build GRCh38
rs561078316 Chr.17: 82036253-82036253 on Build GRCh38
rs769992223 Chr.17: 82035969-82035969 on Build GRCh38
rs568333462 Chr.17: 82035876-82035876 on Build GRCh38
rs749283015 Chr.17: 82036076-82036076 on Build GRCh38
rs757116817 Chr.17: 82035958-82035958 on Build GRCh38
rs770204795 Chr.17: 82036202-82036202 on Build GRCh38
rs895675550 Chr.17: 82035839-82035839 on Build GRCh38
rs111359621 Chr.17: 82035934-82035934 on Build GRCh38
rs892487009 Chr.17: 82036015-82036015 on Build GRCh38
rs75500599 Chr.17: 82035929-82035929 on Build GRCh38
rs777235187 Chr.17: 82036111-82036111 on Build GRCh38
rs371796283 Chr.17: 82036035-82036035 on Build GRCh38
rs767288563 Chr.17: 82036129-82036129 on Build GRCh38
rs1040168134 Chr.17: 82035881-82035881 on Build GRCh38
rs867427353 Chr.17: 82036259-82036259 on Build GRCh38
rs201802500 Chr.17: 82036019-82036019 on Build GRCh38
rs377245458 Chr.17: 82036149-82036149 on Build GRCh38
rs1127830 Chr.17: 82035972-82035972 on Build GRCh38
rs756000092 Chr.17: 82036141-82036141 on Build GRCh38
rs980415892 Chr.17: 82035823-82035823 on Build GRCh38
rs28755469 Chr.17: 82035820-82035820 on Build GRCh38
rs774660365 Chr.17: 82035994-82035994 on Build GRCh38
rs565143449 Chr.17: 82036120-82036120 on Build GRCh38
rs8067668 Chr.17: 82036143-82036143 on Build GRCh38
rs753866546 Chr.17: 82036144-82036144 on Build GRCh38
rs533262853 Chr.17: 82035818-82035818 on Build GRCh38
rs61746217 Chr.17: 82036043-82036043 on Build GRCh38
rs1050876228 Chr.17: 82036169-82036169 on Build GRCh38
rs754459850 Chr.17: 82035922-82035922 on Build GRCh38
rs772225756 Chr.17: 82036163-82036163 on Build GRCh38
rs758420462 Chr.17: 82036000-82036000 on Build GRCh38
rs937160940 Chr.17: 82035833-82035833 on Build GRCh38
rs1052358657 Chr.17: 82036004-82036004 on Build GRCh38
rs1004958485 Chr.17: 82035858-82035858 on Build GRCh38
rs992216927 Chr.17: 82035817-82035817 on Build GRCh38
rs772419492 Chr.17: 82036096-82036096 on Build GRCh38
rs777527142 Chr.17: 82036262-82036262 on Build GRCh38
rs113465430 Chr.17: 82035945-82035945 on Build GRCh38
rs144807548 Chr.17: 82036061-82036061 on Build GRCh38
rs749403860 Chr.17: 82035988-82035988 on Build GRCh38

View Full Product Details