Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 82058259-82058760 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUS1L
Entrez Gene ID
64118
Gene Name
dihydrouridine synthase 1 like
Gene Aliases
DUS1, PP3111
UniGene
Hs.514599
Gene Chromosome Location
Chr.17: 82057872-82065821 on Build GRCh38
Transcripts
NM_022156.4 XM_005256393.2 XM_005256394.2 XM_006722288.2 XM_006722289.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6650254 c.1252C>T p.R418* Chr.17: 82058371-82058371 on Build GRCh38
6398531 c.1235G>T p.R412L Chr.17: 82058388-82058388 on Build GRCh38
4070813 c.1253G>A p.R418Q Chr.17: 82058370-82058370 on Build GRCh38
7105478 c.1252C>G p.R418G Chr.17: 82058371-82058371 on Build GRCh38
1318421 c.1236C>T p.R412R Chr.17: 82058387-82058387 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs777832090 Chr.17: 82058362-82058362 on Build GRCh38
rs758835398 Chr.17: 82058690-82058690 on Build GRCh38
rs890468655 Chr.17: 82058662-82058662 on Build GRCh38
rs142897071 Chr.17: 82058632-82058632 on Build GRCh38
rs879867340 Chr.17: 82058498-82058498 on Build GRCh38
rs752467409 Chr.17: 82058420-82058420 on Build GRCh38
rs914379628 Chr.17: 82058488-82058488 on Build GRCh38
rs766491905 Chr.17: 82058680-82058680 on Build GRCh38
rs202067934 Chr.17: 82058417-82058417 on Build GRCh38
rs372300965 Chr.17: 82058313-82058313 on Build GRCh38
rs552765571 Chr.17: 82058520-82058520 on Build GRCh38
rs201246098 Chr.17: 82058316-82058316 on Build GRCh38
rs555049539 Chr.17: 82058566-82058566 on Build GRCh38
rs769380507 Chr.17: 82058650-82058650 on Build GRCh38
rs569743283 Chr.17: 82058435-82058435 on Build GRCh38
rs150635996 Chr.17: 82058656-82058656 on Build GRCh38
rs757650467 Chr.17: 82058693-82058693 on Build GRCh38
rs894661705 Chr.17: 82058399-82058399 on Build GRCh38
rs868729663 Chr.17: 82058622-82058622 on Build GRCh38
rs772822388 Chr.17: 82058733-82058733 on Build GRCh38
rs779423835 Chr.17: 82058604-82058604 on Build GRCh38
rs771922140 Chr.17: 82058732-82058732 on Build GRCh38
rs771072775 Chr.17: 82058304-82058304 on Build GRCh38
rs776692588 Chr.17: 82058739-82058739 on Build GRCh38
rs1056890591 Chr.17: 82058393-82058393 on Build GRCh38
rs774989848 Chr.17: 82058494-82058494 on Build GRCh38
rs776772338 Chr.17: 82058654-82058663 on Build GRCh38
rs752132431 Chr.17: 82058692-82058692 on Build GRCh38
rs370082561 Chr.17: 82058317-82058317 on Build GRCh38
rs772369745 Chr.17: 82058570-82058579 on Build GRCh38
rs773990146 Chr.17: 82058668-82058668 on Build GRCh38
rs781445339 Chr.17: 82058372-82058372 on Build GRCh38
rs748623442 Chr.17: 82058473-82058473 on Build GRCh38
rs757631906 Chr.17: 82058302-82058303 on Build GRCh38
rs751531064 Chr.17: 82058506-82058506 on Build GRCh38
rs771743965 Chr.17: 82058670-82058670 on Build GRCh38
rs774077853 Chr.17: 82058583-82058583 on Build GRCh38
rs752597705 Chr.17: 82058345-82058345 on Build GRCh38
rs754769541 Chr.17: 82058334-82058334 on Build GRCh38
rs772640481 Chr.17: 82058568-82058568 on Build GRCh38
rs376659580 Chr.17: 82058370-82058370 on Build GRCh38
rs545364780 Chr.17: 82058687-82058687 on Build GRCh38
rs760666251 Chr.17: 82058736-82058736 on Build GRCh38
rs750451136 Chr.17: 82058521-82058521 on Build GRCh38
rs370390336 Chr.17: 82058460-82058460 on Build GRCh38
rs900631126 Chr.17: 82058615-82058615 on Build GRCh38
rs767558688 Chr.17: 82058585-82058585 on Build GRCh38
rs770981128 Chr.17: 82058371-82058371 on Build GRCh38
rs112560415 Chr.17: 82058577-82058578 on Build GRCh38
rs751755062 Chr.17: 82058461-82058461 on Build GRCh38
rs757142360 Chr.17: 82058516-82058516 on Build GRCh38
rs766551972 Chr.17: 82058458-82058458 on Build GRCh38
rs779548111 Chr.17: 82058703-82058703 on Build GRCh38
rs766329154 Chr.17: 82058593-82058593 on Build GRCh38
rs914496349 Chr.17: 82058694-82058694 on Build GRCh38
rs760319739 Chr.17: 82058397-82058397 on Build GRCh38
rs563952657 Chr.17: 82058707-82058707 on Build GRCh38
rs541870122 Chr.17: 82058333-82058333 on Build GRCh38
rs531072642 Chr.17: 82058302-82058302 on Build GRCh38
rs778251482 Chr.17: 82058610-82058610 on Build GRCh38
rs899380876 Chr.17: 82058424-82058424 on Build GRCh38
rs758294938 Chr.17: 82058358-82058358 on Build GRCh38
rs759872390 Chr.17: 82058309-82058309 on Build GRCh38
rs762782949 Chr.17: 82058496-82058496 on Build GRCh38
rs367654816 Chr.17: 82058279-82058279 on Build GRCh38
rs777957524 Chr.17: 82058691-82058691 on Build GRCh38
rs1047953297 Chr.17: 82058639-82058639 on Build GRCh38
rs750316624 Chr.17: 82058588-82058588 on Build GRCh38
rs768784916 Chr.17: 82058594-82058595 on Build GRCh38
rs761655171 Chr.17: 82058669-82058669 on Build GRCh38
rs977379746 Chr.17: 82058730-82058730 on Build GRCh38
rs779307705 Chr.17: 82058509-82058514 on Build GRCh38
rs977737533 Chr.17: 82058558-82058558 on Build GRCh38
rs368394562 Chr.17: 82058297-82058297 on Build GRCh38
rs769221897 Chr.17: 82058395-82058395 on Build GRCh38
rs539331883 Chr.17: 82058386-82058386 on Build GRCh38
rs754134112 Chr.17: 82058595-82058595 on Build GRCh38
rs780382603 Chr.17: 82058527-82058527 on Build GRCh38
rs768234900 Chr.17: 82058664-82058664 on Build GRCh38
rs779325097 Chr.17: 82058470-82058493 on Build GRCh38
rs372944774 Chr.17: 82058463-82058463 on Build GRCh38
rs762555310 Chr.17: 82058577-82058577 on Build GRCh38
rs776826110 Chr.17: 82058305-82058305 on Build GRCh38
rs765609064 Chr.17: 82058688-82058688 on Build GRCh38
rs756730573 Chr.17: 82058578-82058578 on Build GRCh38
rs200669716 Chr.17: 82058321-82058321 on Build GRCh38
rs1028076865 Chr.17: 82058456-82058456 on Build GRCh38
rs753620879 Chr.17: 82058287-82058287 on Build GRCh38
rs775037311 Chr.17: 82058466-82058466 on Build GRCh38
rs929599634 Chr.17: 82058633-82058633 on Build GRCh38
rs776148682 Chr.17: 82058482-82058482 on Build GRCh38
rs774994519 Chr.17: 82058396-82058396 on Build GRCh38
rs534334913 Chr.17: 82058584-82058584 on Build GRCh38
rs747439326 Chr.17: 82058638-82058638 on Build GRCh38
rs148506045 Chr.17: 82058491-82058491 on Build GRCh38
rs372912714 Chr.17: 82058410-82058410 on Build GRCh38
rs773243176 Chr.17: 82058672-82058672 on Build GRCh38
rs377326703 Chr.17: 82058716-82058716 on Build GRCh38
rs987514673 Chr.17: 82058308-82058308 on Build GRCh38
rs543202095 Chr.17: 82058724-82058724 on Build GRCh38
rs915627463 Chr.17: 82058600-82058600 on Build GRCh38
rs745437588 Chr.17: 82058575-82058575 on Build GRCh38
rs753464381 Chr.17: 82058414-82058414 on Build GRCh38
rs140545906 Chr.17: 82058720-82058720 on Build GRCh38
rs980000746 Chr.17: 82058706-82058706 on Build GRCh38
rs528501942 Chr.17: 82058332-82058332 on Build GRCh38
rs759111560 Chr.17: 82058490-82058490 on Build GRCh38
rs755016619 Chr.17: 82058467-82058467 on Build GRCh38
rs1011398123 Chr.17: 82058698-82058698 on Build GRCh38
rs760479427 Chr.17: 82058677-82058677 on Build GRCh38
rs751473507 Chr.17: 82058439-82058439 on Build GRCh38
rs749730139 Chr.17: 82058462-82058462 on Build GRCh38
rs1037907180 Chr.17: 82058620-82058620 on Build GRCh38
rs781193608 Chr.17: 82058503-82058503 on Build GRCh38
rs770587807 Chr.17: 82058480-82058480 on Build GRCh38
rs779492736 Chr.17: 82058652-82058652 on Build GRCh38
rs368733115 Chr.17: 82058389-82058389 on Build GRCh38
rs374612561 Chr.17: 82058292-82058292 on Build GRCh38
rs780010842 Chr.17: 82058282-82058282 on Build GRCh38
rs372697085 Chr.17: 82058322-82058322 on Build GRCh38
rs763848578 Chr.17: 82058499-82058499 on Build GRCh38
rs994149017 Chr.17: 82058434-82058434 on Build GRCh38
rs970260572 Chr.17: 82058705-82058705 on Build GRCh38
rs1007878115 Chr.17: 82058683-82058683 on Build GRCh38
rs770840231 Chr.17: 82058737-82058737 on Build GRCh38
rs757370937 Chr.17: 82058441-82058441 on Build GRCh38
rs145282065 Chr.17: 82058374-82058374 on Build GRCh38
rs760859623 Chr.17: 82058592-82058592 on Build GRCh38
rs749500949 Chr.17: 82058562-82058562 on Build GRCh38
rs754799152 Chr.17: 82058288-82058288 on Build GRCh38
rs540824953 Chr.17: 82058592-82058593 on Build GRCh38
rs4280314 Chr.17: 82058576-82058576 on Build GRCh38
rs546981229 Chr.17: 82058335-82058335 on Build GRCh38
rs755193983 Chr.17: 82058602-82058602 on Build GRCh38
rs773584473 Chr.17: 82058574-82058574 on Build GRCh38
rs377062150 Chr.17: 82058588-82058589 on Build GRCh38
rs147637107 Chr.17: 82058387-82058387 on Build GRCh38
rs755423201 Chr.17: 82058564-82058564 on Build GRCh38
rs761826323 Chr.17: 82058685-82058687 on Build GRCh38
rs746115129 Chr.17: 82058566-82058566 on Build GRCh38
rs563712584 Chr.17: 82058293-82058293 on Build GRCh38
rs778391841 Chr.17: 82058637-82058637 on Build GRCh38
rs199715310 Chr.17: 82058289-82058289 on Build GRCh38

View Full Product Details