Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 82058703-82059209 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUS1L
Entrez Gene ID
64118
Gene Name
dihydrouridine synthase 1 like
Gene Aliases
DUS1, PP3111
UniGene
Hs.514599
Gene Chromosome Location
Chr.17: 82057872-82065821 on Build GRCh38
Transcripts
NM_022156.4 XM_005256393.2 XM_005256394.2 XM_006722288.2 XM_006722289.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6417151 c.1206+6T>A p.? Chr.17: 82058775-82058775 on Build GRCh38
5041515 c.1181A>C p.K394T Chr.17: 82058806-82058806 on Build GRCh38
1740406 c.1193G>A p.C398Y Chr.17: 82058794-82058794 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs983462426 Chr.17: 82058990-82058990 on Build GRCh38
rs776063404 Chr.17: 82059174-82059174 on Build GRCh38
rs376200459 Chr.17: 82058783-82058783 on Build GRCh38
rs1051959722 Chr.17: 82058873-82058873 on Build GRCh38
rs374739810 Chr.17: 82059175-82059175 on Build GRCh38
rs767838401 Chr.17: 82058852-82058852 on Build GRCh38
rs530441773 Chr.17: 82059113-82059113 on Build GRCh38
rs779696797 Chr.17: 82058977-82058977 on Build GRCh38
rs1039042486 Chr.17: 82058901-82058901 on Build GRCh38
rs561602068 Chr.17: 82059092-82059092 on Build GRCh38
rs772822388 Chr.17: 82058733-82058733 on Build GRCh38
rs777477372 Chr.17: 82058800-82058800 on Build GRCh38
rs771922140 Chr.17: 82058732-82058732 on Build GRCh38
rs533298610 Chr.17: 82058994-82058994 on Build GRCh38
rs756554306 Chr.17: 82058863-82058863 on Build GRCh38
rs776692588 Chr.17: 82058739-82058739 on Build GRCh38
rs371117778 Chr.17: 82058794-82058794 on Build GRCh38
rs189369433 Chr.17: 82058995-82058995 on Build GRCh38
rs370035035 Chr.17: 82058862-82058862 on Build GRCh38
rs552969660 Chr.17: 82059072-82059072 on Build GRCh38
rs770823323 Chr.17: 82058804-82058804 on Build GRCh38
rs373764869 Chr.17: 82058769-82058769 on Build GRCh38
rs569886152 Chr.17: 82059014-82059014 on Build GRCh38
rs763040376 Chr.17: 82058754-82058754 on Build GRCh38
rs1011254451 Chr.17: 82058897-82058897 on Build GRCh38
rs932278120 Chr.17: 82059032-82059032 on Build GRCh38
rs942029451 Chr.17: 82058997-82058997 on Build GRCh38
rs959980248 Chr.17: 82059010-82059010 on Build GRCh38
rs879760072 Chr.17: 82059006-82059006 on Build GRCh38
rs944996384 Chr.17: 82059079-82059079 on Build GRCh38
rs557378736 Chr.17: 82059141-82059141 on Build GRCh38
rs754525560 Chr.17: 82058775-82058775 on Build GRCh38
rs1036021562 Chr.17: 82059147-82059148 on Build GRCh38
rs942040716 Chr.17: 82058801-82058802 on Build GRCh38
rs113440348 Chr.17: 82058913-82058913 on Build GRCh38
rs907975759 Chr.17: 82058991-82058991 on Build GRCh38
rs762134622 Chr.17: 82058843-82058843 on Build GRCh38
rs372002075 Chr.17: 82058889-82058889 on Build GRCh38
rs895634724 Chr.17: 82059162-82059162 on Build GRCh38
rs759735255 Chr.17: 82058748-82058748 on Build GRCh38
rs113000111 Chr.17: 82058885-82058885 on Build GRCh38
rs1039241246 Chr.17: 82059080-82059080 on Build GRCh38
rs373366357 Chr.17: 82058818-82058818 on Build GRCh38
rs760666251 Chr.17: 82058736-82058736 on Build GRCh38
rs900497381 Chr.17: 82058953-82058953 on Build GRCh38
rs1052819326 Chr.17: 82059061-82059062 on Build GRCh38
rs890605288 Chr.17: 82059042-82059042 on Build GRCh38
rs530630069 Chr.17: 82059042-82059042 on Build GRCh38
rs757920275 Chr.17: 82058796-82058796 on Build GRCh38
rs770012885 Chr.17: 82058815-82058815 on Build GRCh38
rs752215475 Chr.17: 82058868-82058868 on Build GRCh38
rs764534827 Chr.17: 82058755-82058755 on Build GRCh38
rs567047488 Chr.17: 82059069-82059069 on Build GRCh38
rs773030874 Chr.17: 82058824-82058831 on Build GRCh38
rs762618607 Chr.17: 82058851-82058852 on Build GRCh38
rs184751787 Chr.17: 82059110-82059110 on Build GRCh38
rs375743917 Chr.17: 82058837-82058837 on Build GRCh38
rs757640736 Chr.17: 82058767-82058767 on Build GRCh38
rs565120799 Chr.17: 82058985-82058985 on Build GRCh38
rs765374342 Chr.17: 82058750-82058750 on Build GRCh38
rs180864289 Chr.17: 82059070-82059070 on Build GRCh38
rs1001435731 Chr.17: 82059088-82059088 on Build GRCh38
rs1052457662 Chr.17: 82059041-82059041 on Build GRCh38
rs573409377 Chr.17: 82059156-82059156 on Build GRCh38
rs977379746 Chr.17: 82058730-82058730 on Build GRCh38
rs1022774162 Chr.17: 82058978-82058978 on Build GRCh38
rs970024229 Chr.17: 82058912-82058912 on Build GRCh38
rs946686047 Chr.17: 82059130-82059130 on Build GRCh38
rs189155853 Chr.17: 82059150-82059150 on Build GRCh38
rs764729633 Chr.17: 82058865-82058865 on Build GRCh38
rs775376635 Chr.17: 82058835-82058835 on Build GRCh38
rs921983625 Chr.17: 82059096-82059096 on Build GRCh38
rs369610213 Chr.17: 82058768-82058768 on Build GRCh38
rs970116525 Chr.17: 82059023-82059023 on Build GRCh38
rs879921594 Chr.17: 82059073-82059073 on Build GRCh38
rs920800306 Chr.17: 82058884-82058884 on Build GRCh38
rs543265382 Chr.17: 82059147-82059147 on Build GRCh38
rs1022662290 Chr.17: 82059021-82059021 on Build GRCh38
rs571150818 Chr.17: 82059087-82059087 on Build GRCh38
rs999606092 Chr.17: 82059149-82059149 on Build GRCh38
rs774623441 Chr.17: 82058840-82058840 on Build GRCh38
rs922157684 Chr.17: 82059018-82059018 on Build GRCh38
rs771442630 Chr.17: 82058811-82058811 on Build GRCh38
rs201131425 Chr.17: 82059057-82059057 on Build GRCh38
rs904376656 Chr.17: 82058979-82058979 on Build GRCh38
rs575974224 Chr.17: 82059142-82059142 on Build GRCh38
rs1031603873 Chr.17: 82059003-82059003 on Build GRCh38
rs543202095 Chr.17: 82058724-82058724 on Build GRCh38
rs377110757 Chr.17: 82058761-82058761 on Build GRCh38
rs528567843 Chr.17: 82058900-82058900 on Build GRCh38
rs944714810 Chr.17: 82058896-82058896 on Build GRCh38
rs1014210049 Chr.17: 82059163-82059163 on Build GRCh38
rs769524053 Chr.17: 82058806-82058806 on Build GRCh38
rs912051486 Chr.17: 82059078-82059078 on Build GRCh38
rs548734936 Chr.17: 82059058-82059058 on Build GRCh38
rs775883868 Chr.17: 82058753-82058753 on Build GRCh38
rs1043822691 Chr.17: 82059138-82059138 on Build GRCh38
rs770840231 Chr.17: 82058737-82058737 on Build GRCh38
rs1021960418 Chr.17: 82058909-82058909 on Build GRCh38
rs776745658 Chr.17: 82058808-82058808 on Build GRCh38
rs865822198 Chr.17: 82059048-82059048 on Build GRCh38
rs928459164 Chr.17: 82058938-82058938 on Build GRCh38
rs372250092 Chr.17: 82059171-82059171 on Build GRCh38
rs983283938 Chr.17: 82059151-82059151 on Build GRCh38
rs367779231 Chr.17: 82058888-82058888 on Build GRCh38

View Full Product Details