Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 82127602-82128096 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC57
Entrez Gene ID
284001
Gene Name
coiled-coil domain containing 57
Gene Aliases
-
UniGene
Hs.631724
Gene Chromosome Location
Chr.17: 82101458-82212892 on Build GRCh38
Transcripts
NM_198082.2 XM_011523545.2 XM_011523546.2 XM_011523548.2 XM_011523549.2 XM_011523551.2 XM_011523552.2 XM_011523555.2 XM_011523557.2 XM_017024462.1 XM_017024463.1 XM_017024464.1 XM_017024465.1 XM_017024466.1 XM_017024467.1 XM_017024468.1 XM_017024469.1 XM_017024470.1 XM_017024471.1 XM_017024472.1 XM_017024473.1 XM_017024474.1 XM_017024475.1 XM_017024476.1 XM_017024477.1 XM_017024478.1 XM_017024479.1 XM_017024483.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
437822 c.2501T>C p.M834T Chr.17: 82127757-82127757 on Build GRCh38
3821043 c.2488A>C p.T830P Chr.17: 82127770-82127770 on Build GRCh38
6622273 c.2452C>T p.R818C Chr.17: 82127806-82127806 on Build GRCh38
6189993 c.2405G>T p.R802M Chr.17: 82127853-82127853 on Build GRCh38
6622266 c.371G>A p.R124H Chr.17: 82127805-82127805 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746493044 Chr.17: 82127794-82127794 on Build GRCh38
rs750790520 Chr.17: 82127763-82127763 on Build GRCh38
rs996405601 Chr.17: 82127770-82127770 on Build GRCh38
rs770618095 Chr.17: 82127799-82127799 on Build GRCh38
rs772369654 Chr.17: 82128072-82128072 on Build GRCh38
rs11077969 Chr.17: 82127757-82127757 on Build GRCh38
rs375113072 Chr.17: 82127673-82127673 on Build GRCh38
rs772576742 Chr.17: 82127670-82127670 on Build GRCh38
rs755573619 Chr.17: 82127862-82127862 on Build GRCh38
rs917099787 Chr.17: 82127949-82127949 on Build GRCh38
rs370334205 Chr.17: 82127646-82127646 on Build GRCh38
rs11077970 Chr.17: 82128040-82128040 on Build GRCh38
rs755410467 Chr.17: 82127788-82127788 on Build GRCh38
rs996313723 Chr.17: 82128077-82128077 on Build GRCh38
rs745693145 Chr.17: 82127806-82127806 on Build GRCh38
rs548959567 Chr.17: 82127838-82127838 on Build GRCh38
rs770488811 Chr.17: 82127681-82127681 on Build GRCh38
rs766477657 Chr.17: 82127714-82127714 on Build GRCh38
rs754137716 Chr.17: 82127860-82127860 on Build GRCh38
rs1040102909 Chr.17: 82128054-82128054 on Build GRCh38
rs747689416 Chr.17: 82127650-82127650 on Build GRCh38
rs756431589 Chr.17: 82127767-82127767 on Build GRCh38
rs779173860 Chr.17: 82127974-82127974 on Build GRCh38
rs771562622 Chr.17: 82127661-82127661 on Build GRCh38
rs766777108 Chr.17: 82127842-82127842 on Build GRCh38
rs369190828 Chr.17: 82127925-82127925 on Build GRCh38
rs763007061 Chr.17: 82127817-82127817 on Build GRCh38
rs369275557 Chr.17: 82127997-82127997 on Build GRCh38
rs773252035 Chr.17: 82127941-82127941 on Build GRCh38
rs775405823 Chr.17: 82127814-82127814 on Build GRCh38
rs746538712 Chr.17: 82127872-82127872 on Build GRCh38
rs769559832 Chr.17: 82127896-82127896 on Build GRCh38
rs7213172 Chr.17: 82127827-82127827 on Build GRCh38
rs936586909 Chr.17: 82127986-82127986 on Build GRCh38
rs774494657 Chr.17: 82127823-82127823 on Build GRCh38
rs757556785 Chr.17: 82127756-82127756 on Build GRCh38
rs894983154 Chr.17: 82128002-82128002 on Build GRCh38
rs777411364 Chr.17: 82127789-82127789 on Build GRCh38
rs932939058 Chr.17: 82127625-82127625 on Build GRCh38
rs1004632325 Chr.17: 82127738-82127738 on Build GRCh38
rs879184189 Chr.17: 82127908-82127908 on Build GRCh38
rs780063876 Chr.17: 82127901-82127901 on Build GRCh38
rs748970320 Chr.17: 82127924-82127924 on Build GRCh38
rs530754735 Chr.17: 82127831-82127831 on Build GRCh38
rs761982946 Chr.17: 82127933-82127933 on Build GRCh38
rs374792018 Chr.17: 82127889-82127889 on Build GRCh38
rs1049208615 Chr.17: 82128019-82128019 on Build GRCh38
rs752238822 Chr.17: 82127645-82127645 on Build GRCh38
rs1000768868 Chr.17: 82127643-82127643 on Build GRCh38
rs78509330 Chr.17: 82127772-82127772 on Build GRCh38
rs1042112546 Chr.17: 82127977-82127977 on Build GRCh38
rs759350359 Chr.17: 82127689-82127689 on Build GRCh38
rs371380244 Chr.17: 82127888-82127888 on Build GRCh38
rs902463401 Chr.17: 82127635-82127635 on Build GRCh38
rs370764424 Chr.17: 82127706-82127706 on Build GRCh38
rs368060305 Chr.17: 82127682-82127682 on Build GRCh38
rs376747467 Chr.17: 82127820-82127820 on Build GRCh38
rs372964713 Chr.17: 82127955-82127955 on Build GRCh38
rs779217850 Chr.17: 82127869-82127869 on Build GRCh38
rs764271285 Chr.17: 82127715-82127715 on Build GRCh38
rs989053145 Chr.17: 82127629-82127629 on Build GRCh38
rs886095746 Chr.17: 82128037-82128037 on Build GRCh38
rs774107709 Chr.17: 82127926-82127926 on Build GRCh38
rs1019563497 Chr.17: 82127747-82127747 on Build GRCh38
rs1048696903 Chr.17: 82127697-82127697 on Build GRCh38
rs143558954 Chr.17: 82128013-82128013 on Build GRCh38
rs76663260 Chr.17: 82127979-82127979 on Build GRCh38
rs964535278 Chr.17: 82127904-82127905 on Build GRCh38
rs1053681285 Chr.17: 82127989-82127989 on Build GRCh38
rs776361582 Chr.17: 82127805-82127805 on Build GRCh38
rs765292615 Chr.17: 82127699-82127699 on Build GRCh38
rs374247944 Chr.17: 82127711-82127711 on Build GRCh38
rs573257555 Chr.17: 82127630-82127630 on Build GRCh38
rs763660437 Chr.17: 82127917-82127917 on Build GRCh38
rs749844215 Chr.17: 82127776-82127776 on Build GRCh38
rs772178729 Chr.17: 82127937-82127937 on Build GRCh38

View Full Product Details