Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 82740612-82741157 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FN3K
Entrez Gene ID
64122
Gene Name
fructosamine 3 kinase
Gene Aliases
-
UniGene
Hs.151135
Gene Chromosome Location
Chr.17: 82735576-82751197 on Build GRCh38
Transcripts
NM_022158.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
986167 c.357G>T p.E119D Chr.17: 82740826-82740826 on Build GRCh38
4489718 c.351C>T p.L117L Chr.17: 82740820-82740820 on Build GRCh38
3665247 c.334C>T p.L112F Chr.17: 82740803-82740803 on Build GRCh38
6338245 c.333T>C p.H111H Chr.17: 82740802-82740802 on Build GRCh38
6338725 c.315G>C p.E105D Chr.17: 82740784-82740784 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs111799031 Chr.17: 82741119-82741119 on Build GRCh38
rs534389539 Chr.17: 82741094-82741094 on Build GRCh38
rs777643195 Chr.17: 82740868-82740868 on Build GRCh38
rs749861928 Chr.17: 82740845-82740845 on Build GRCh38
rs778678575 Chr.17: 82740846-82740846 on Build GRCh38
rs745436501 Chr.17: 82740712-82740712 on Build GRCh38
rs752059225 Chr.17: 82740801-82740801 on Build GRCh38
rs567796038 Chr.17: 82741030-82741030 on Build GRCh38
rs770721895 Chr.17: 82740891-82740891 on Build GRCh38
rs149684982 Chr.17: 82740964-82740964 on Build GRCh38
rs547939833 Chr.17: 82740855-82740855 on Build GRCh38
rs746912854 Chr.17: 82740734-82740737 on Build GRCh38
rs774219553 Chr.17: 82740741-82740741 on Build GRCh38
rs559000346 Chr.17: 82740764-82740764 on Build GRCh38
rs919485096 Chr.17: 82740790-82740790 on Build GRCh38
rs79442878 Chr.17: 82740944-82740944 on Build GRCh38
rs774959876 Chr.17: 82740947-82740947 on Build GRCh38
rs372789922 Chr.17: 82740731-82740731 on Build GRCh38
rs371100076 Chr.17: 82740745-82740745 on Build GRCh38
rs374127367 Chr.17: 82740755-82740755 on Build GRCh38
rs998750355 Chr.17: 82741107-82741107 on Build GRCh38
rs190928766 Chr.17: 82740794-82740794 on Build GRCh38
rs1042389846 Chr.17: 82741036-82741036 on Build GRCh38
rs80113076 Chr.17: 82740945-82740945 on Build GRCh38
rs377756170 Chr.17: 82740821-82740821 on Build GRCh38
rs776513213 Chr.17: 82740758-82740758 on Build GRCh38
rs1054212701 Chr.17: 82741124-82741124 on Build GRCh38
rs563257242 Chr.17: 82740672-82740672 on Build GRCh38
rs776801225 Chr.17: 82740892-82740892 on Build GRCh38
rs767716820 Chr.17: 82741063-82741063 on Build GRCh38
rs768138182 Chr.17: 82740737-82740737 on Build GRCh38
rs757642134 Chr.17: 82740802-82740802 on Build GRCh38
rs779630733 Chr.17: 82740803-82740803 on Build GRCh38
rs377202968 Chr.17: 82740736-82740736 on Build GRCh38
rs753097460 Chr.17: 82740787-82740787 on Build GRCh38
rs570335020 Chr.17: 82740929-82740929 on Build GRCh38
rs771536239 Chr.17: 82740898-82740898 on Build GRCh38
rs201547124 Chr.17: 82740901-82740901 on Build GRCh38
rs901936004 Chr.17: 82740985-82740985 on Build GRCh38
rs560970710 Chr.17: 82740757-82740757 on Build GRCh38
rs747097517 Chr.17: 82740722-82740725 on Build GRCh38
rs776411489 Chr.17: 82741073-82741073 on Build GRCh38
rs371202212 Chr.17: 82740827-82740827 on Build GRCh38
rs139392875 Chr.17: 82740792-82740792 on Build GRCh38
rs75537818 Chr.17: 82740711-82740711 on Build GRCh38
rs879200738 Chr.17: 82740902-82740902 on Build GRCh38
rs570797027 Chr.17: 82741128-82741128 on Build GRCh38
rs1032834190 Chr.17: 82740687-82740687 on Build GRCh38
rs77084831 Chr.17: 82740666-82740666 on Build GRCh38
rs552066263 Chr.17: 82740921-82740921 on Build GRCh38
rs759824342 Chr.17: 82740769-82740769 on Build GRCh38
rs773269165 Chr.17: 82740746-82740746 on Build GRCh38
rs765332597 Chr.17: 82740773-82740773 on Build GRCh38
rs747913394 Chr.17: 82740742-82740742 on Build GRCh38
rs771988218 Chr.17: 82740864-82740864 on Build GRCh38
rs565874822 Chr.17: 82740701-82740701 on Build GRCh38
rs537392692 Chr.17: 82740959-82740959 on Build GRCh38
rs559755944 Chr.17: 82740879-82740879 on Build GRCh38

View Full Product Details