Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 657935-658472 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TYMS
Entrez Gene ID
7298
Gene Name
thymidylate synthetase
Gene Aliases
HST422, TMS, TS
UniGene
Hs.369762
Gene Chromosome Location
Chr.18: 657604-673499 on Build GRCh38
Transcripts
NM_001071.2 NM_001012716.2

Gene Symbol
TYMSOS
Entrez Gene ID
494514
Gene Name
TYMS opposite strand
Gene Aliases
C18orf56
UniGene
Hs.274959
Gene Chromosome Location
Chr.18: 649620-658340 on Build GRCh38
Transcripts
NM_001071.2 NM_001012716.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5648492 c.61C>T p.R21C Chr.18: 658187-658187 on Build GRCh38
5564780 c.49C>T p.R17W Chr.18: 658199-658199 on Build GRCh38
5017891 c.287delG p.G96fs*>28 Chr.18: 657961-657961 on Build GRCh38
6332622 c.203C>A p.T68N Chr.18: 658045-658045 on Build GRCh38
6317554 c.253C>A p.R85S Chr.18: 657995-657995 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765389767 Chr.18: 658281-658281 on Build GRCh38
rs775154346 Chr.18: 658126-658126 on Build GRCh38
rs371604300 Chr.18: 658149-658149 on Build GRCh38
rs1054515792 Chr.18: 658221-658221 on Build GRCh38
rs750633928 Chr.18: 657997-657997 on Build GRCh38
rs985686228 Chr.18: 658412-658412 on Build GRCh38
rs1037122446 Chr.18: 658263-658263 on Build GRCh38
rs1001773794 Chr.18: 658073-658073 on Build GRCh38
rs768186816 Chr.18: 658136-658136 on Build GRCh38
rs761703220 Chr.18: 658060-658060 on Build GRCh38
rs1014637784 Chr.18: 658118-658118 on Build GRCh38
rs745703243 Chr.18: 657969-657969 on Build GRCh38
rs368711614 Chr.18: 658066-658066 on Build GRCh38
rs936036150 Chr.18: 658043-658043 on Build GRCh38
rs370824496 Chr.18: 658057-658057 on Build GRCh38
rs772000195 Chr.18: 658238-658238 on Build GRCh38
rs550800489 Chr.18: 658265-658265 on Build GRCh38
rs758887422 Chr.18: 658116-658116 on Build GRCh38
rs1006035935 Chr.18: 658322-658322 on Build GRCh38
rs918390385 Chr.18: 658266-658266 on Build GRCh38
rs907108428 Chr.18: 658249-658249 on Build GRCh38
rs953164532 Chr.18: 658410-658410 on Build GRCh38
rs747334000 Chr.18: 658199-658199 on Build GRCh38
rs747627213 Chr.18: 658237-658237 on Build GRCh38
rs200637090 Chr.18: 658024-658024 on Build GRCh38
rs1041078250 Chr.18: 658370-658370 on Build GRCh38
rs772125266 Chr.18: 657992-657992 on Build GRCh38
rs536583615 Chr.18: 658301-658301 on Build GRCh38
rs755300515 Chr.18: 658178-658178 on Build GRCh38
rs376122364 Chr.18: 658333-658333 on Build GRCh38
rs578120349 Chr.18: 658026-658026 on Build GRCh38
rs779438042 Chr.18: 658113-658133 on Build GRCh38
rs576822037 Chr.18: 658435-658435 on Build GRCh38
rs28602966 Chr.18: 658170-658170 on Build GRCh38
rs957512941 Chr.18: 658422-658422 on Build GRCh38
rs758880712 Chr.18: 658196-658196 on Build GRCh38
rs756340417 Chr.18: 658084-658084 on Build GRCh38
rs889724531 Chr.18: 658360-658360 on Build GRCh38
rs560976325 Chr.18: 658087-658087 on Build GRCh38
rs563640714 Chr.18: 658038-658038 on Build GRCh38
rs778164405 Chr.18: 658197-658197 on Build GRCh38
rs764032292 Chr.18: 657964-657964 on Build GRCh38
rs2847612 Chr.18: 658430-658430 on Build GRCh38
rs975795511 Chr.18: 658145-658145 on Build GRCh38
rs780884826 Chr.18: 657988-657988 on Build GRCh38
rs773848513 Chr.18: 658233-658233 on Build GRCh38
rs757583882 Chr.18: 658205-658205 on Build GRCh38
rs946188016 Chr.18: 658452-658452 on Build GRCh38
rs1034181807 Chr.18: 658095-658095 on Build GRCh38
rs151221029 Chr.18: 658109-658109 on Build GRCh38
rs778130604 Chr.18: 658119-658119 on Build GRCh38
rs995436175 Chr.18: 657963-657963 on Build GRCh38
rs773316519 Chr.18: 658264-658264 on Build GRCh38
rs1048796379 Chr.18: 658089-658089 on Build GRCh38
rs773542884 Chr.18: 658125-658125 on Build GRCh38
rs767363359 Chr.18: 658152-658152 on Build GRCh38
rs766672286 Chr.18: 658167-658167 on Build GRCh38
rs762807813 Chr.18: 657961-657961 on Build GRCh38
rs867324318 Chr.18: 658049-658049 on Build GRCh38
rs757102809 Chr.18: 657970-657970 on Build GRCh38
rs182344736 Chr.18: 658147-658147 on Build GRCh38
rs760573111 Chr.18: 658164-658164 on Build GRCh38
rs897610784 Chr.18: 658349-658349 on Build GRCh38
rs759801692 Chr.18: 658274-658274 on Build GRCh38
rs572858775 Chr.18: 658019-658019 on Build GRCh38
rs755140967 Chr.18: 658102-658102 on Build GRCh38
rs752787982 Chr.18: 658191-658191 on Build GRCh38
rs776627638 Chr.18: 658273-658273 on Build GRCh38
rs769439371 Chr.18: 658039-658039 on Build GRCh38
rs773500709 Chr.18: 658030-658030 on Build GRCh38
rs201596180 Chr.18: 658121-658121 on Build GRCh38
rs1045772374 Chr.18: 658248-658248 on Build GRCh38
rs1006606844 Chr.18: 657982-657982 on Build GRCh38
rs1029962702 Chr.18: 658379-658379 on Build GRCh38
rs991259686 Chr.18: 658182-658182 on Build GRCh38
rs922904611 Chr.18: 658015-658015 on Build GRCh38
rs373697938 Chr.18: 658097-658097 on Build GRCh38
rs781582011 Chr.18: 658216-658216 on Build GRCh38
rs75047328 Chr.18: 658417-658417 on Build GRCh38
rs763894899 Chr.18: 658054-658054 on Build GRCh38
rs760695865 Chr.18: 658050-658050 on Build GRCh38
rs371383896 Chr.18: 658072-658072 on Build GRCh38
rs748975926 Chr.18: 658131-658131 on Build GRCh38
rs780279297 Chr.18: 658013-658013 on Build GRCh38
rs558592399 Chr.18: 658025-658025 on Build GRCh38
rs748922826 Chr.18: 658227-658227 on Build GRCh38
rs773086787 Chr.18: 658158-658158 on Build GRCh38
rs3891167 Chr.18: 658423-658423 on Build GRCh38
rs190076880 Chr.18: 658048-658048 on Build GRCh38
rs769387992 Chr.18: 658123-658123 on Build GRCh38
rs757776594 Chr.18: 658068-658069 on Build GRCh38
rs1001466540 Chr.18: 658442-658442 on Build GRCh38
rs751262220 Chr.18: 657966-657966 on Build GRCh38
rs369135185 Chr.18: 658447-658447 on Build GRCh38
rs142008740 Chr.18: 658287-658287 on Build GRCh38
rs1046048081 Chr.18: 658058-658058 on Build GRCh38
rs887590638 Chr.18: 658302-658302 on Build GRCh38
rs72634355 Chr.18: 658332-658332 on Build GRCh38
rs750740109 Chr.18: 658361-658361 on Build GRCh38
rs866565470 Chr.18: 658157-658157 on Build GRCh38
rs938046291 Chr.18: 658034-658034 on Build GRCh38
rs551390018 Chr.18: 658027-658027 on Build GRCh38
rs768256881 Chr.18: 658229-658229 on Build GRCh38
rs2853533 Chr.18: 658064-658064 on Build GRCh38
rs765347463 Chr.18: 658180-658180 on Build GRCh38
rs73366471 Chr.18: 658001-658001 on Build GRCh38
rs77400748 Chr.18: 658035-658035 on Build GRCh38
rs766254423 Chr.18: 658271-658271 on Build GRCh38

View Full Product Details