Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 14752621-14753119 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD30B
Entrez Gene ID
374860
Gene Name
ankyrin repeat domain 30B
Gene Aliases
NY-BR-1.1
UniGene
Hs.567889
Gene Chromosome Location
Chr.18: 14748240-14855465 on Build GRCh38
Transcripts
NM_001145029.1 XM_006722325.3 XM_011525662.2 XM_011525664.2 XM_011525665.2 XM_011525666.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3524358 c.304C>T p.L102F Chr.18: 14752648-14752648 on Build GRCh38
5515663 c.336+8G>T p.? Chr.18: 14752688-14752688 on Build GRCh38
7240222 c.439G>C p.V147L Chr.18: 14752941-14752941 on Build GRCh38
4661842 c.418delA p.N140fs*14 Chr.18: 14752920-14752920 on Build GRCh38
4962281 c.510+4G>T p.? Chr.18: 14753016-14753016 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763946877 Chr.18: 14752644-14752644 on Build GRCh38
rs187185365 Chr.18: 14753081-14753081 on Build GRCh38
rs780091376 Chr.18: 14753038-14753038 on Build GRCh38
rs374547236 Chr.18: 14752817-14752817 on Build GRCh38
rs756853799 Chr.18: 14752864-14752864 on Build GRCh38
rs756128884 Chr.18: 14752962-14752962 on Build GRCh38
rs887350971 Chr.18: 14752903-14752903 on Build GRCh38
rs776005399 Chr.18: 14753077-14753077 on Build GRCh38
rs752374686 Chr.18: 14752933-14752933 on Build GRCh38
rs939352410 Chr.18: 14752721-14752721 on Build GRCh38
rs559142364 Chr.18: 14752851-14752851 on Build GRCh38
rs764540544 Chr.18: 14752940-14752940 on Build GRCh38
rs753488886 Chr.18: 14752832-14752832 on Build GRCh38
rs768351086 Chr.18: 14752727-14752727 on Build GRCh38
rs146944735 Chr.18: 14752974-14752974 on Build GRCh38
rs199597228 Chr.18: 14752850-14752850 on Build GRCh38
rs746084131 Chr.18: 14752884-14752884 on Build GRCh38
rs757718328 Chr.18: 14752945-14752945 on Build GRCh38
rs774040196 Chr.18: 14752892-14752892 on Build GRCh38
rs368825805 Chr.18: 14752910-14752910 on Build GRCh38
rs757730136 Chr.18: 14753015-14753015 on Build GRCh38
rs747513542 Chr.18: 14752875-14752875 on Build GRCh38
rs556051000 Chr.18: 14752656-14752656 on Build GRCh38
rs749897804 Chr.18: 14752661-14752661 on Build GRCh38
rs1001858384 Chr.18: 14752771-14752771 on Build GRCh38
rs753495539 Chr.18: 14752672-14752672 on Build GRCh38
rs768121628 Chr.18: 14753027-14753027 on Build GRCh38
rs900948445 Chr.18: 14752791-14752791 on Build GRCh38
rs766404789 Chr.18: 14752979-14752979 on Build GRCh38
rs533482869 Chr.18: 14753067-14753067 on Build GRCh38
rs764893197 Chr.18: 14752927-14752927 on Build GRCh38
rs777383479 Chr.18: 14752746-14752746 on Build GRCh38
rs541728454 Chr.18: 14752924-14752924 on Build GRCh38
rs773196799 Chr.18: 14752909-14752909 on Build GRCh38
rs776785994 Chr.18: 14752707-14752707 on Build GRCh38
rs751631905 Chr.18: 14752979-14752991 on Build GRCh38
rs750867230 Chr.18: 14753028-14753028 on Build GRCh38
rs765818214 Chr.18: 14752911-14752911 on Build GRCh38
rs760214190 Chr.18: 14752820-14752820 on Build GRCh38
rs769222866 Chr.18: 14753031-14753031 on Build GRCh38
rs12959778 Chr.18: 14752724-14752724 on Build GRCh38
rs752127097 Chr.18: 14752941-14752941 on Build GRCh38
rs556937555 Chr.18: 14752807-14752807 on Build GRCh38
rs572652697 Chr.18: 14752855-14752855 on Build GRCh38
rs747990388 Chr.18: 14752896-14752896 on Build GRCh38
rs776026510 Chr.18: 14752991-14752991 on Build GRCh38
rs759791878 Chr.18: 14752711-14752711 on Build GRCh38
rs576848402 Chr.18: 14752774-14752774 on Build GRCh38
rs761682028 Chr.18: 14752657-14752657 on Build GRCh38
rs755123712 Chr.18: 14753058-14753058 on Build GRCh38
rs988122916 Chr.18: 14752737-14752737 on Build GRCh38
rs574383616 Chr.18: 14752705-14752705 on Build GRCh38
rs757471506 Chr.18: 14752883-14752883 on Build GRCh38
rs984288163 Chr.18: 14752658-14752658 on Build GRCh38
rs771104933 Chr.18: 14752706-14752706 on Build GRCh38
rs771819072 Chr.18: 14752900-14752900 on Build GRCh38
rs201536919 Chr.18: 14752987-14752987 on Build GRCh38
rs758899148 Chr.18: 14752934-14752934 on Build GRCh38
rs746952025 Chr.18: 14752982-14752982 on Build GRCh38
rs767020498 Chr.18: 14752660-14752660 on Build GRCh38
rs370471349 Chr.18: 14752977-14752977 on Build GRCh38
rs754069528 Chr.18: 14753052-14753052 on Build GRCh38
rs764621240 Chr.18: 14752996-14752996 on Build GRCh38
rs763188433 Chr.18: 14752938-14752938 on Build GRCh38
rs762637500 Chr.18: 14752998-14752998 on Build GRCh38
rs751935007 Chr.18: 14753009-14753009 on Build GRCh38
rs780368020 Chr.18: 14752972-14752972 on Build GRCh38
rs764558965 Chr.18: 14753004-14753004 on Build GRCh38
rs1039427827 Chr.18: 14752790-14752790 on Build GRCh38
rs767398172 Chr.18: 14752814-14752814 on Build GRCh38
rs45487996 Chr.18: 14752740-14752740 on Build GRCh38
rs12457503 Chr.18: 14752958-14752958 on Build GRCh38
rs779726394 Chr.18: 14752670-14752670 on Build GRCh38
rs774131799 Chr.18: 14752805-14752805 on Build GRCh38
rs946720944 Chr.18: 14753076-14753076 on Build GRCh38
rs1042961820 Chr.18: 14752742-14752742 on Build GRCh38
rs748614523 Chr.18: 14752690-14752690 on Build GRCh38
rs913146746 Chr.18: 14752738-14752738 on Build GRCh38
rs772638879 Chr.18: 14752693-14752693 on Build GRCh38
rs138054870 Chr.18: 14752997-14752997 on Build GRCh38
rs759595560 Chr.18: 14753012-14753012 on Build GRCh38
rs942545723 Chr.18: 14752786-14752786 on Build GRCh38
rs960455753 Chr.18: 14753039-14753039 on Build GRCh38
rs546557444 Chr.18: 14752677-14752677 on Build GRCh38
rs778472408 Chr.18: 14752872-14752872 on Build GRCh38
rs910087295 Chr.18: 14752669-14752669 on Build GRCh38
rs765905553 Chr.18: 14752825-14752825 on Build GRCh38
rs781163240 Chr.18: 14752956-14752956 on Build GRCh38
rs755361447 Chr.18: 14752685-14752685 on Build GRCh38
rs779320927 Chr.18: 14752686-14752686 on Build GRCh38
rs45620343 Chr.18: 14752769-14752769 on Build GRCh38
rs770021117 Chr.18: 14752885-14752885 on Build GRCh38
rs755789677 Chr.18: 14752667-14752667 on Build GRCh38

View Full Product Details