Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 26915948-26916451 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AQP4-AS1
Entrez Gene ID
147429
Gene Name
AQP4 antisense RNA 1
Gene Aliases
C18orf16, CHST9-AS1
UniGene
Hs.559280
Gene Chromosome Location
Chr.18: 26865308-26935946 on Build GRCh38
Transcripts
NR_026908.1 NM_001256316.1 NM_031422.5 XM_005258362.3 XM_017026033.1 XM_017026034.1

Gene Symbol
CHST9
Entrez Gene ID
83539
Gene Name
carbohydrate sulfotransferase 9
Gene Aliases
GALNAC4ST-2, GalNAc4ST2
UniGene
Hs.44584
Gene Chromosome Location
Chr.18: 26908754-27185364 on Build GRCh38
Transcripts
NR_026908.1 NM_001256316.1 NM_031422.5 XM_005258362.3 XM_017026033.1 XM_017026034.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6900647 c.1245G>A p.K415K Chr.18: 26916346-26916346 on Build GRCh38
708221 c.928T>C p.F310L Chr.18: 26916408-26916408 on Build GRCh38
5577346 c.945_948delTTCC p.S316fs*9 Chr.18: 26916388-26916391 on Build GRCh38
4406269 c.1168C>T p.L390L Chr.18: 26916423-26916423 on Build GRCh38
5577345 c.1200_1203delTTCC p.S401fs*9 Chr.18: 26916388-26916391 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs901835824 Chr.18: 26916093-26916093 on Build GRCh38
rs764070699 Chr.18: 26916316-26916316 on Build GRCh38
rs752832621 Chr.18: 26916297-26916297 on Build GRCh38
rs201405526 Chr.18: 26916295-26916295 on Build GRCh38
rs751601546 Chr.18: 26916321-26916321 on Build GRCh38
rs763242570 Chr.18: 26916427-26916427 on Build GRCh38
rs760162019 Chr.18: 26916401-26916401 on Build GRCh38
rs1047659612 Chr.18: 26916119-26916119 on Build GRCh38
rs781374538 Chr.18: 26916153-26916153 on Build GRCh38
rs80190384 Chr.18: 26916412-26916412 on Build GRCh38
rs772750714 Chr.18: 26916371-26916371 on Build GRCh38
rs887669532 Chr.18: 26916144-26916144 on Build GRCh38
rs754740628 Chr.18: 26916356-26916356 on Build GRCh38
rs777620473 Chr.18: 26916263-26916263 on Build GRCh38
rs117939395 Chr.18: 26916049-26916049 on Build GRCh38
rs534240641 Chr.18: 26916272-26916272 on Build GRCh38
rs76121570 Chr.18: 26916043-26916043 on Build GRCh38
rs373838914 Chr.18: 26916346-26916346 on Build GRCh38
rs778719992 Chr.18: 26916358-26916358 on Build GRCh38
rs1037374602 Chr.18: 26916081-26916081 on Build GRCh38
rs776277592 Chr.18: 26916161-26916161 on Build GRCh38
rs761310940 Chr.18: 26916400-26916400 on Build GRCh38
rs979596742 Chr.18: 26916038-26916038 on Build GRCh38
rs771451674 Chr.18: 26916256-26916256 on Build GRCh38
rs773941111 Chr.18: 26916238-26916238 on Build GRCh38
rs1006562540 Chr.18: 26916173-26916173 on Build GRCh38
rs749242498 Chr.18: 26916353-26916353 on Build GRCh38
rs747879318 Chr.18: 26916243-26916243 on Build GRCh38
rs1047965145 Chr.18: 26916113-26916113 on Build GRCh38
rs905692489 Chr.18: 26916228-26916228 on Build GRCh38
rs181527568 Chr.18: 26916198-26916198 on Build GRCh38
rs772845513 Chr.18: 26916269-26916269 on Build GRCh38
rs937710324 Chr.18: 26916059-26916059 on Build GRCh38
rs370973380 Chr.18: 26916234-26916234 on Build GRCh38
rs548529042 Chr.18: 26916236-26916236 on Build GRCh38
rs765470557 Chr.18: 26916222-26916224 on Build GRCh38
rs946978688 Chr.18: 26915996-26915996 on Build GRCh38
rs980096769 Chr.18: 26916397-26916397 on Build GRCh38
rs59062970 Chr.18: 26916132-26916133 on Build GRCh38
rs943428487 Chr.18: 26916054-26916054 on Build GRCh38
rs771813887 Chr.18: 26916365-26916365 on Build GRCh38
rs981975493 Chr.18: 26916382-26916382 on Build GRCh38
rs929320431 Chr.18: 26916100-26916100 on Build GRCh38
rs753119683 Chr.18: 26916280-26916286 on Build GRCh38
rs929580937 Chr.18: 26916111-26916111 on Build GRCh38
rs777312052 Chr.18: 26916274-26916274 on Build GRCh38
rs568388273 Chr.18: 26916232-26916232 on Build GRCh38
rs747751233 Chr.18: 26916359-26916359 on Build GRCh38
rs200160632 Chr.18: 26916410-26916410 on Build GRCh38
rs1009079071 Chr.18: 26916185-26916185 on Build GRCh38
rs764697833 Chr.18: 26916367-26916367 on Build GRCh38
rs758764758 Chr.18: 26916317-26916320 on Build GRCh38
rs185798102 Chr.18: 26916270-26916270 on Build GRCh38
rs375278353 Chr.18: 26916388-26916388 on Build GRCh38
rs746718413 Chr.18: 26916320-26916320 on Build GRCh38
rs552704972 Chr.18: 26916377-26916377 on Build GRCh38
rs982331154 Chr.18: 26916010-26916010 on Build GRCh38
rs371234835 Chr.18: 26916364-26916364 on Build GRCh38
rs777256953 Chr.18: 26916393-26916393 on Build GRCh38
rs926407341 Chr.18: 26916018-26916018 on Build GRCh38
rs749162270 Chr.18: 26916300-26916300 on Build GRCh38
rs1040899595 Chr.18: 26916078-26916078 on Build GRCh38
rs759933019 Chr.18: 26916281-26916281 on Build GRCh38
rs757330342 Chr.18: 26916333-26916333 on Build GRCh38
rs757374443 Chr.18: 26915982-26915982 on Build GRCh38
rs768328169 Chr.18: 26916387-26916387 on Build GRCh38
rs779983593 Chr.18: 26916221-26916221 on Build GRCh38

View Full Product Details