Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 37067059-37067563 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA1328
Entrez Gene ID
57536
Gene Name
KIAA1328
Gene Aliases
-
UniGene
Hs.4045
Gene Chromosome Location
Chr.18: 36828929-37232127 on Build GRCh38
Transcripts
NM_001322327.1 NM_020776.1 NR_136303.1 NR_136304.1 NR_136305.1 XM_005258315.4 XM_005258317.3 XM_005258318.4 XM_011526099.2 XM_011526100.2 XM_017025876.1 XM_017025877.1 XM_017025878.1 XM_017025879.1 XM_017025880.1 XM_017025881.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
709586 c.1048G>A p.G350R Chr.18: 37067361-37067361 on Build GRCh38
4275094 c.195T>G p.G65G Chr.18: 37067360-37067360 on Build GRCh38
4912908 c.975G>C p.K325N Chr.18: 37067300-37067300 on Build GRCh38
7352473 c.1034G>T p.G345V Chr.18: 37067359-37067359 on Build GRCh38
4580405 c.58C>A p.Q20K Chr.18: 37067223-37067223 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200193060 Chr.18: 37067090-37067090 on Build GRCh38
rs531868906 Chr.18: 37067329-37067329 on Build GRCh38
rs561248590 Chr.18: 37067144-37067144 on Build GRCh38
rs756607035 Chr.18: 37067422-37067422 on Build GRCh38
rs765861567 Chr.18: 37067444-37067444 on Build GRCh38
rs758828744 Chr.18: 37067102-37067102 on Build GRCh38
rs748905439 Chr.18: 37067342-37067342 on Build GRCh38
rs766522920 Chr.18: 37067174-37067174 on Build GRCh38
rs556086527 Chr.18: 37067103-37067103 on Build GRCh38
rs750978464 Chr.18: 37067403-37067403 on Build GRCh38
rs1055607667 Chr.18: 37067517-37067517 on Build GRCh38
rs755698231 Chr.18: 37067336-37067336 on Build GRCh38
rs774479080 Chr.18: 37067146-37067146 on Build GRCh38
rs761789868 Chr.18: 37067259-37067259 on Build GRCh38
rs372936367 Chr.18: 37067269-37067269 on Build GRCh38
rs769325693 Chr.18: 37067227-37067227 on Build GRCh38
rs764395931 Chr.18: 37067357-37067358 on Build GRCh38
rs370899316 Chr.18: 37067333-37067333 on Build GRCh38
rs12326301 Chr.18: 37067460-37067460 on Build GRCh38
rs772145664 Chr.18: 37067264-37067264 on Build GRCh38
rs767888785 Chr.18: 37067401-37067401 on Build GRCh38
rs764306641 Chr.18: 37067375-37067375 on Build GRCh38
rs745464096 Chr.18: 37067120-37067120 on Build GRCh38
rs757696470 Chr.18: 37067328-37067328 on Build GRCh38
rs763105137 Chr.18: 37067366-37067366 on Build GRCh38
rs777464467 Chr.18: 37067478-37067478 on Build GRCh38
rs542158748 Chr.18: 37067136-37067136 on Build GRCh38
rs774315647 Chr.18: 37067258-37067258 on Build GRCh38
rs772125014 Chr.18: 37067358-37067358 on Build GRCh38
rs376208853 Chr.18: 37067235-37067235 on Build GRCh38
rs181674796 Chr.18: 37067530-37067530 on Build GRCh38
rs1047053698 Chr.18: 37067542-37067542 on Build GRCh38
rs770469851 Chr.18: 37067117-37067117 on Build GRCh38
rs1014196773 Chr.18: 37067245-37067245 on Build GRCh38
rs556288231 Chr.18: 37067426-37067426 on Build GRCh38
rs199661890 Chr.18: 37067290-37067290 on Build GRCh38
rs949911937 Chr.18: 37067532-37067532 on Build GRCh38
rs763061028 Chr.18: 37067347-37067356 on Build GRCh38
rs548499817 Chr.18: 37067356-37067356 on Build GRCh38
rs1032455531 Chr.18: 37067388-37067388 on Build GRCh38
rs746504413 Chr.18: 37067109-37067109 on Build GRCh38
rs751931041 Chr.18: 37067458-37067459 on Build GRCh38
rs753971381 Chr.18: 37067176-37067176 on Build GRCh38
rs754696394 Chr.18: 37067346-37067346 on Build GRCh38
rs762221829 Chr.18: 37067397-37067397 on Build GRCh38
rs776573185 Chr.18: 37067285-37067285 on Build GRCh38
rs375543657 Chr.18: 37067100-37067100 on Build GRCh38
rs201864375 Chr.18: 37067461-37067461 on Build GRCh38
rs373293089 Chr.18: 37067215-37067215 on Build GRCh38
rs200769332 Chr.18: 37067371-37067371 on Build GRCh38
rs760802041 Chr.18: 37067153-37067153 on Build GRCh38
rs867319007 Chr.18: 37067134-37067134 on Build GRCh38
rs762271838 Chr.18: 37067466-37067467 on Build GRCh38
rs764394519 Chr.18: 37067327-37067327 on Build GRCh38
rs775574588 Chr.18: 37067529-37067529 on Build GRCh38
rs778790935 Chr.18: 37067351-37067351 on Build GRCh38
rs142778459 Chr.18: 37067254-37067254 on Build GRCh38
rs886967519 Chr.18: 37067292-37067292 on Build GRCh38
rs765359136 Chr.18: 37067303-37067303 on Build GRCh38
rs775454436 Chr.18: 37067364-37067364 on Build GRCh38
rs912903031 Chr.18: 37067113-37067113 on Build GRCh38
rs536436186 Chr.18: 37067253-37067253 on Build GRCh38
rs779541198 Chr.18: 37067338-37067338 on Build GRCh38
rs189282803 Chr.18: 37067408-37067408 on Build GRCh38
rs758815921 Chr.18: 37067186-37067186 on Build GRCh38
rs1006049917 Chr.18: 37067276-37067276 on Build GRCh38
rs925468418 Chr.18: 37067091-37067091 on Build GRCh38
rs753394860 Chr.18: 37067495-37067495 on Build GRCh38
rs368650756 Chr.18: 37067381-37067381 on Build GRCh38
rs752994435 Chr.18: 37067185-37067185 on Build GRCh38
rs778223324 Chr.18: 37067210-37067210 on Build GRCh38
rs775674730 Chr.18: 37067231-37067234 on Build GRCh38
rs769501238 Chr.18: 37067123-37067123 on Build GRCh38
rs3747895 Chr.18: 37067213-37067213 on Build GRCh38
rs753410065 Chr.18: 37067459-37067459 on Build GRCh38
rs569368453 Chr.18: 37067320-37067320 on Build GRCh38
rs770787335 Chr.18: 37067513-37067513 on Build GRCh38
rs779814978 Chr.18: 37067229-37067229 on Build GRCh38
rs576257098 Chr.18: 37067118-37067118 on Build GRCh38
rs539179533 Chr.18: 37067418-37067418 on Build GRCh38
rs530142922 Chr.18: 37067183-37067183 on Build GRCh38
rs745386624 Chr.18: 37067217-37067217 on Build GRCh38
rs753007246 Chr.18: 37067305-37067305 on Build GRCh38
rs72890602 Chr.18: 37067359-37067359 on Build GRCh38
rs200614954 Chr.18: 37067214-37067214 on Build GRCh38
rs978899645 Chr.18: 37067424-37067424 on Build GRCh38
rs879873322 Chr.18: 37067236-37067236 on Build GRCh38
rs3747896 Chr.18: 37067360-37067360 on Build GRCh38

View Full Product Details