Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 59296389-59296900 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPLX4
Entrez Gene ID
339302
Gene Name
complexin 4
Gene Aliases
CPX-IV, CPXIV
UniGene
Hs.449884
Gene Chromosome Location
Chr.18: 59295402-59318649 on Build GRCh38
Transcripts
NM_181654.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1718495 c.286G>A p.A96T Chr.18: 59296895-59296895 on Build GRCh38
6083517 c.447C>A p.I149I Chr.18: 59296734-59296734 on Build GRCh38
170105 c.458C>T p.A153V Chr.18: 59296723-59296723 on Build GRCh38
6358480 c.418G>C p.E140Q Chr.18: 59296763-59296763 on Build GRCh38
280104 c.361G>T p.D121Y Chr.18: 59296820-59296820 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756534722 Chr.18: 59296755-59296755 on Build GRCh38
rs771426097 Chr.18: 59296674-59296674 on Build GRCh38
rs985227766 Chr.18: 59296511-59296511 on Build GRCh38
rs148430146 Chr.18: 59296854-59296854 on Build GRCh38
rs890935895 Chr.18: 59296830-59296830 on Build GRCh38
rs766191153 Chr.18: 59296848-59296848 on Build GRCh38
rs1028126659 Chr.18: 59296600-59296600 on Build GRCh38
rs9954747 Chr.18: 59296670-59296670 on Build GRCh38
rs764708029 Chr.18: 59296554-59296554 on Build GRCh38
rs573065812 Chr.18: 59296729-59296729 on Build GRCh38
rs773866910 Chr.18: 59296817-59296817 on Build GRCh38
rs1056187408 Chr.18: 59296681-59296681 on Build GRCh38
rs775187540 Chr.18: 59296691-59296691 on Build GRCh38
rs764820442 Chr.18: 59296640-59296640 on Build GRCh38
rs888520772 Chr.18: 59296822-59296822 on Build GRCh38
rs748906033 Chr.18: 59296799-59296799 on Build GRCh38
rs939078991 Chr.18: 59296429-59296429 on Build GRCh38
rs564929455 Chr.18: 59296491-59296491 on Build GRCh38
rs774164945 Chr.18: 59296695-59296695 on Build GRCh38
rs745547819 Chr.18: 59296689-59296689 on Build GRCh38
rs747546607 Chr.18: 59296671-59296671 on Build GRCh38
rs775579082 Chr.18: 59296849-59296849 on Build GRCh38
rs965292917 Chr.18: 59296841-59296841 on Build GRCh38
rs949604552 Chr.18: 59296722-59296722 on Build GRCh38
rs746286693 Chr.18: 59296743-59296743 on Build GRCh38
rs549121916 Chr.18: 59296584-59296584 on Build GRCh38
rs770930522 Chr.18: 59296594-59296594 on Build GRCh38
rs139886644 Chr.18: 59296604-59296604 on Build GRCh38
rs374236510 Chr.18: 59296769-59296769 on Build GRCh38
rs778993856 Chr.18: 59296659-59296659 on Build GRCh38
rs764112572 Chr.18: 59296859-59296859 on Build GRCh38
rs965263702 Chr.18: 59296541-59296541 on Build GRCh38
rs772035562 Chr.18: 59296826-59296826 on Build GRCh38
rs762431120 Chr.18: 59296692-59296692 on Build GRCh38
rs768211175 Chr.18: 59296693-59296693 on Build GRCh38
rs752949677 Chr.18: 59296712-59296712 on Build GRCh38
rs4940456 Chr.18: 59296510-59296510 on Build GRCh38
rs373720376 Chr.18: 59296823-59296823 on Build GRCh38
rs35068407 Chr.18: 59296803-59296803 on Build GRCh38
rs376337443 Chr.18: 59296838-59296838 on Build GRCh38
rs767318677 Chr.18: 59296698-59296698 on Build GRCh38
rs781649955 Chr.18: 59296679-59296679 on Build GRCh38
rs202101844 Chr.18: 59296706-59296706 on Build GRCh38
rs752019452 Chr.18: 59296723-59296723 on Build GRCh38
rs75707948 Chr.18: 59296465-59296465 on Build GRCh38
rs750174236 Chr.18: 59296700-59296700 on Build GRCh38
rs17696504 Chr.18: 59296620-59296620 on Build GRCh38
rs750368635 Chr.18: 59296707-59296707 on Build GRCh38
rs538625675 Chr.18: 59296643-59296643 on Build GRCh38
rs557235805 Chr.18: 59296647-59296647 on Build GRCh38
rs149408519 Chr.18: 59296707-59296707 on Build GRCh38
rs767517665 Chr.18: 59296681-59296682 on Build GRCh38
rs752757172 Chr.18: 59296660-59296660 on Build GRCh38
rs779642789 Chr.18: 59296765-59296765 on Build GRCh38
rs144069269 Chr.18: 59296861-59296861 on Build GRCh38
rs767236960 Chr.18: 59296649-59296649 on Build GRCh38
rs773108748 Chr.18: 59296832-59296832 on Build GRCh38
rs750134087 Chr.18: 59296650-59296650 on Build GRCh38
rs956180513 Chr.18: 59296634-59296634 on Build GRCh38
rs370860769 Chr.18: 59296862-59296862 on Build GRCh38
rs531020927 Chr.18: 59296581-59296581 on Build GRCh38
rs368826625 Chr.18: 59296690-59296690 on Build GRCh38
rs994537668 Chr.18: 59296774-59296774 on Build GRCh38
rs182607438 Chr.18: 59296635-59296635 on Build GRCh38
rs781137378 Chr.18: 59296782-59296782 on Build GRCh38
rs781755340 Chr.18: 59296742-59296742 on Build GRCh38
rs750964905 Chr.18: 59296866-59296866 on Build GRCh38
rs147289072 Chr.18: 59296476-59296476 on Build GRCh38
rs761684060 Chr.18: 59296697-59296697 on Build GRCh38
rs563796406 Chr.18: 59296580-59296580 on Build GRCh38
rs756554813 Chr.18: 59296868-59296868 on Build GRCh38
rs534834836 Chr.18: 59296613-59296613 on Build GRCh38
rs1027462910 Chr.18: 59296508-59296508 on Build GRCh38
rs996802012 Chr.18: 59296585-59296585 on Build GRCh38
rs928767785 Chr.18: 59296667-59296667 on Build GRCh38
rs371133746 Chr.18: 59296717-59296717 on Build GRCh38
rs764166992 Chr.18: 59296718-59296718 on Build GRCh38
rs1008291309 Chr.18: 59296608-59296608 on Build GRCh38
rs17769193 Chr.18: 59296535-59296535 on Build GRCh38

View Full Product Details