Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 68714816-68715357 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC102B
Entrez Gene ID
79839
Gene Name
coiled-coil domain containing 102B
Gene Aliases
ACY1L, AN, C18orf14, HsT1731
UniGene
Hs.280781
Gene Chromosome Location
Chr.18: 68715221-69214350 on Build GRCh38
Transcripts
NM_001093729.1 XM_017025973.1 XM_017025975.1 XM_017025976.1 XM_017025979.1 XM_017025980.1 NM_019022.3 XM_005266714.2 XM_011526033.2 XM_011526034.2 XM_011526035.2

Gene Symbol
TMX3
Entrez Gene ID
54495
Gene Name
thioredoxin related transmembrane protein 3
Gene Aliases
PDIA13, TXNDC10
UniGene
Hs.440534
Gene Chromosome Location
Chr.18: 68673688-68715116 on Build GRCh38
Transcripts
NM_001093729.1 XM_017025973.1 XM_017025975.1 XM_017025976.1 XM_017025979.1 XM_017025980.1 NM_019022.3 XM_005266714.2 XM_011526033.2 XM_011526034.2 XM_011526035.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1684987 c.17G>T p.S6I Chr.18: 68714965-68714965 on Build GRCh38
4421359 c.8C>T p.A3V Chr.18: 68714974-68714974 on Build GRCh38
6326906 c.15G>A p.K5K Chr.18: 68714967-68714967 on Build GRCh38
5505419 c.19T>C p.W7R Chr.18: 68714963-68714963 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs964977424 Chr.18: 68715228-68715228 on Build GRCh38
rs181088195 Chr.18: 68715330-68715330 on Build GRCh38
rs767923692 Chr.18: 68714891-68714891 on Build GRCh38
rs768754708 Chr.18: 68715266-68715266 on Build GRCh38
rs545060751 Chr.18: 68715011-68715012 on Build GRCh38
rs762260660 Chr.18: 68714887-68714887 on Build GRCh38
rs867676004 Chr.18: 68714938-68714938 on Build GRCh38
rs972173313 Chr.18: 68715139-68715140 on Build GRCh38
rs930525019 Chr.18: 68714901-68714901 on Build GRCh38
rs546985305 Chr.18: 68715286-68715286 on Build GRCh38
rs370611762 Chr.18: 68714921-68714921 on Build GRCh38
rs139484151 Chr.18: 68715241-68715241 on Build GRCh38
rs996897327 Chr.18: 68715050-68715050 on Build GRCh38
rs971790186 Chr.18: 68715239-68715239 on Build GRCh38
rs1055746786 Chr.18: 68715077-68715077 on Build GRCh38
rs768064428 Chr.18: 68715008-68715008 on Build GRCh38
rs570898827 Chr.18: 68715243-68715243 on Build GRCh38
rs558928095 Chr.18: 68715194-68715194 on Build GRCh38
rs764639499 Chr.18: 68715069-68715069 on Build GRCh38
rs572320949 Chr.18: 68715111-68715111 on Build GRCh38
rs369181680 Chr.18: 68714911-68714911 on Build GRCh38
rs113574586 Chr.18: 68715294-68715294 on Build GRCh38
rs774643124 Chr.18: 68715005-68715005 on Build GRCh38
rs952665016 Chr.18: 68715180-68715181 on Build GRCh38
rs932381337 Chr.18: 68715269-68715269 on Build GRCh38
rs865983306 Chr.18: 68715058-68715058 on Build GRCh38
rs780312969 Chr.18: 68714910-68714910 on Build GRCh38
rs141697037 Chr.18: 68715134-68715134 on Build GRCh38
rs889385618 Chr.18: 68715209-68715209 on Build GRCh38
rs902948364 Chr.18: 68715101-68715101 on Build GRCh38
rs538910176 Chr.18: 68714837-68714837 on Build GRCh38
rs977513393 Chr.18: 68715123-68715123 on Build GRCh38
rs530335258 Chr.18: 68714931-68714931 on Build GRCh38
rs970991565 Chr.18: 68715080-68715080 on Build GRCh38
rs112163304 Chr.18: 68715032-68715032 on Build GRCh38
rs976606665 Chr.18: 68715128-68715128 on Build GRCh38
rs940721657 Chr.18: 68714856-68714856 on Build GRCh38
rs1049875275 Chr.18: 68715281-68715281 on Build GRCh38
rs959488244 Chr.18: 68715100-68715100 on Build GRCh38
rs899949219 Chr.18: 68714854-68714854 on Build GRCh38
rs770405183 Chr.18: 68714975-68714975 on Build GRCh38
rs570530481 Chr.18: 68715245-68715245 on Build GRCh38
rs776241442 Chr.18: 68714986-68714986 on Build GRCh38
rs188158033 Chr.18: 68715188-68715188 on Build GRCh38
rs866505028 Chr.18: 68715075-68715075 on Build GRCh38
rs1004592916 Chr.18: 68715061-68715061 on Build GRCh38
rs1033855107 Chr.18: 68715090-68715090 on Build GRCh38
rs576171290 Chr.18: 68715082-68715082 on Build GRCh38
rs536570061 Chr.18: 68715115-68715115 on Build GRCh38
rs373822949 Chr.18: 68714899-68714899 on Build GRCh38
rs760424705 Chr.18: 68714989-68714989 on Build GRCh38
rs199638611 Chr.18: 68714964-68714964 on Build GRCh38
rs903034395 Chr.18: 68714842-68714842 on Build GRCh38
rs968909570 Chr.18: 68715212-68715212 on Build GRCh38
rs865988166 Chr.18: 68714957-68714957 on Build GRCh38
rs1031308541 Chr.18: 68715227-68715227 on Build GRCh38
rs888473812 Chr.18: 68715060-68715060 on Build GRCh38
rs867506615 Chr.18: 68714967-68714967 on Build GRCh38
rs947017789 Chr.18: 68715011-68715011 on Build GRCh38
rs1033907591 Chr.18: 68715170-68715170 on Build GRCh38
rs543585906 Chr.18: 68715160-68715160 on Build GRCh38
rs868836497 Chr.18: 68715321-68715321 on Build GRCh38
rs774731403 Chr.18: 68714925-68714926 on Build GRCh38
rs984385199 Chr.18: 68715238-68715238 on Build GRCh38
rs752131491 Chr.18: 68715191-68715191 on Build GRCh38
rs766185629 Chr.18: 68715022-68715022 on Build GRCh38
rs117138050 Chr.18: 68714963-68714963 on Build GRCh38
rs146246189 Chr.18: 68715006-68715006 on Build GRCh38
rs943938752 Chr.18: 68715024-68715024 on Build GRCh38
rs113071377 Chr.18: 68715149-68715149 on Build GRCh38
rs309249 Chr.18: 68714873-68714873 on Build GRCh38
rs759297596 Chr.18: 68715063-68715063 on Build GRCh38
rs868069641 Chr.18: 68714962-68714962 on Build GRCh38
rs78924743 Chr.18: 68715260-68715260 on Build GRCh38
rs1003795706 Chr.18: 68715086-68715086 on Build GRCh38
rs982307169 Chr.18: 68714954-68714954 on Build GRCh38
rs924296160 Chr.18: 68715125-68715125 on Build GRCh38
rs757397075 Chr.18: 68715037-68715037 on Build GRCh38
rs1037079199 Chr.18: 68715244-68715244 on Build GRCh38
rs309248 Chr.18: 68715237-68715237 on Build GRCh38
rs542133234 Chr.18: 68715221-68715221 on Build GRCh38
rs543165825 Chr.18: 68714845-68714845 on Build GRCh38
rs953713210 Chr.18: 68715019-68715019 on Build GRCh38
rs771983599 Chr.18: 68714943-68714943 on Build GRCh38
rs895422102 Chr.18: 68715290-68715290 on Build GRCh38
rs914246174 Chr.18: 68715010-68715010 on Build GRCh38
rs868293159 Chr.18: 68714951-68714951 on Build GRCh38
rs527240627 Chr.18: 68715229-68715229 on Build GRCh38
rs199817898 Chr.18: 68715238-68715239 on Build GRCh38
rs1037479364 Chr.18: 68715154-68715154 on Build GRCh38
rs113880168 Chr.18: 68715148-68715148 on Build GRCh38
rs996465802 Chr.18: 68715120-68715120 on Build GRCh38
rs375368590 Chr.18: 68714923-68714923 on Build GRCh38
rs750638663 Chr.18: 68715105-68715105 on Build GRCh38
rs867265479 Chr.18: 68714979-68714979 on Build GRCh38
rs576762582 Chr.18: 68715162-68715162 on Build GRCh38
rs943989665 Chr.18: 68715126-68715126 on Build GRCh38
rs1032615544 Chr.18: 68714965-68714965 on Build GRCh38
rs756218821 Chr.18: 68714906-68714906 on Build GRCh38
rs770707104 Chr.18: 68715307-68715307 on Build GRCh38
rs976418490 Chr.18: 68715332-68715332 on Build GRCh38
rs76594176 Chr.18: 68715038-68715038 on Build GRCh38
rs750450973 Chr.18: 68714893-68714893 on Build GRCh38
rs1019602791 Chr.18: 68715220-68715220 on Build GRCh38
rs115125462 Chr.18: 68714868-68714868 on Build GRCh38
rs1022857223 Chr.18: 68715148-68715166 on Build GRCh38
rs1004472312 Chr.18: 68714884-68714884 on Build GRCh38
rs1055117224 Chr.18: 68715271-68715271 on Build GRCh38
rs370046451 Chr.18: 68715150-68715150 on Build GRCh38
rs376675449 Chr.18: 68715020-68715020 on Build GRCh38
rs35101496 Chr.18: 68715320-68715321 on Build GRCh38
rs865889107 Chr.18: 68715246-68715246 on Build GRCh38
rs932464854 Chr.18: 68715144-68715144 on Build GRCh38
rs548329439 Chr.18: 68714936-68714936 on Build GRCh38
rs113692845 Chr.18: 68715179-68715179 on Build GRCh38
rs74917552 Chr.18: 68715206-68715206 on Build GRCh38
rs764396768 Chr.18: 68714926-68714926 on Build GRCh38
rs532038381 Chr.18: 68714862-68714862 on Build GRCh38
rs117720303 Chr.18: 68715295-68715295 on Build GRCh38
rs1045527536 Chr.18: 68715099-68715099 on Build GRCh38
rs781626048 Chr.18: 68715230-68715230 on Build GRCh38
rs116186173 Chr.18: 68714853-68714853 on Build GRCh38
rs952791165 Chr.18: 68714878-68714878 on Build GRCh38
rs576189681 Chr.18: 68715137-68715137 on Build GRCh38
rs1015583756 Chr.18: 68715071-68715071 on Build GRCh38
rs1016239028 Chr.18: 68714916-68714916 on Build GRCh38
rs906524966 Chr.18: 68715157-68715157 on Build GRCh38
rs534758219 Chr.18: 68715102-68715102 on Build GRCh38
rs1029234616 Chr.18: 68715122-68715122 on Build GRCh38
rs779979251 Chr.18: 68715242-68715242 on Build GRCh38
rs760790303 Chr.18: 68715025-68715025 on Build GRCh38
rs554101218 Chr.18: 68715108-68715108 on Build GRCh38
rs924549989 Chr.18: 68715023-68715023 on Build GRCh38
rs769459920 Chr.18: 68714998-68714998 on Build GRCh38
rs367625691 Chr.18: 68714942-68714942 on Build GRCh38
rs759225063 Chr.18: 68714994-68714994 on Build GRCh38
rs914276361 Chr.18: 68715103-68715103 on Build GRCh38
rs571970251 Chr.18: 68715153-68715154 on Build GRCh38
rs150531716 Chr.18: 68715232-68715232 on Build GRCh38
rs902644017 Chr.18: 68715298-68715298 on Build GRCh38

View Full Product Details