Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 74446514-74447009 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM69C
Entrez Gene ID
125704
Gene Name
family with sequence similarity 69 member C
Gene Aliases
C18orf51
UniGene
Hs.371690
Gene Chromosome Location
Chr.18: 74432575-74464646 on Build GRCh38
Transcripts
NM_001044369.2 XM_011525816.2 XM_017025551.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4798191 c.723C>T p.A241A Chr.18: 74446759-74446759 on Build GRCh38
4854415 c.705C>T p.L235L Chr.18: 74446777-74446777 on Build GRCh38
6049471 c.475C>T p.L159L Chr.18: 74447007-74447007 on Build GRCh38
4889149 c.845C>A p.P282Q Chr.18: 74446637-74446637 on Build GRCh38
4932815 c.499A>T p.S167C Chr.18: 74446983-74446983 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763679336 Chr.18: 74446620-74446620 on Build GRCh38
rs778579435 Chr.18: 74446976-74446976 on Build GRCh38
rs200748260 Chr.18: 74446648-74446648 on Build GRCh38
rs891305303 Chr.18: 74446830-74446830 on Build GRCh38
rs552295990 Chr.18: 74446784-74446784 on Build GRCh38
rs941744661 Chr.18: 74446956-74446956 on Build GRCh38
rs752901203 Chr.18: 74446833-74446833 on Build GRCh38
rs887556442 Chr.18: 74446955-74446955 on Build GRCh38
rs1037320631 Chr.18: 74446801-74446801 on Build GRCh38
rs980670198 Chr.18: 74446774-74446774 on Build GRCh38
rs988849992 Chr.18: 74446710-74446710 on Build GRCh38
rs574324944 Chr.18: 74446845-74446845 on Build GRCh38
rs749121805 Chr.18: 74446749-74446749 on Build GRCh38
rs760411454 Chr.18: 74446606-74446606 on Build GRCh38
rs985617104 Chr.18: 74446935-74446935 on Build GRCh38
rs372816832 Chr.18: 74446950-74446950 on Build GRCh38
rs1022865306 Chr.18: 74446647-74446647 on Build GRCh38
rs929053174 Chr.18: 74446819-74446819 on Build GRCh38
rs1035587861 Chr.18: 74446982-74446982 on Build GRCh38
rs777206927 Chr.18: 74446957-74446957 on Build GRCh38
rs201867846 Chr.18: 74446694-74446694 on Build GRCh38
rs372573789 Chr.18: 74446590-74446590 on Build GRCh38
rs928934172 Chr.18: 74446951-74446951 on Build GRCh38
rs1025452767 Chr.18: 74446972-74446973 on Build GRCh38
rs779781210 Chr.18: 74446725-74446725 on Build GRCh38
rs754127109 Chr.18: 74446576-74446576 on Build GRCh38
rs1047526153 Chr.18: 74446952-74446952 on Build GRCh38
rs767581441 Chr.18: 74446915-74446915 on Build GRCh38
rs761432761 Chr.18: 74446588-74446588 on Build GRCh38
rs551035156 Chr.18: 74446722-74446722 on Build GRCh38
rs867129671 Chr.18: 74446644-74446644 on Build GRCh38
rs200975934 Chr.18: 74446764-74446764 on Build GRCh38
rs993257993 Chr.18: 74446614-74446614 on Build GRCh38
rs1003213489 Chr.18: 74446980-74446980 on Build GRCh38
rs111448276 Chr.18: 74446953-74446953 on Build GRCh38
rs529170679 Chr.18: 74446615-74446615 on Build GRCh38
rs970405937 Chr.18: 74446848-74446848 on Build GRCh38
rs993660637 Chr.18: 74446804-74446804 on Build GRCh38
rs1008495653 Chr.18: 74446653-74446653 on Build GRCh38
rs540465275 Chr.18: 74446973-74446973 on Build GRCh38
rs956807243 Chr.18: 74446695-74446695 on Build GRCh38
rs556607927 Chr.18: 74446863-74446863 on Build GRCh38
rs926508330 Chr.18: 74446778-74446778 on Build GRCh38
rs750529271 Chr.18: 74446936-74446936 on Build GRCh38
rs758668760 Chr.18: 74446836-74446836 on Build GRCh38
rs951826134 Chr.18: 74446619-74446619 on Build GRCh38
rs201797177 Chr.18: 74446803-74446803 on Build GRCh38
rs576473771 Chr.18: 74446896-74446896 on Build GRCh38
rs764498424 Chr.18: 74446834-74446834 on Build GRCh38
rs747604907 Chr.18: 74446769-74446769 on Build GRCh38
rs914628171 Chr.18: 74446924-74446924 on Build GRCh38
rs944321376 Chr.18: 74446941-74446941 on Build GRCh38
rs140849087 Chr.18: 74446820-74446820 on Build GRCh38
rs563652633 Chr.18: 74446662-74446662 on Build GRCh38
rs79862102 Chr.18: 74446597-74446597 on Build GRCh38
rs572436317 Chr.18: 74446963-74446963 on Build GRCh38
rs956033802 Chr.18: 74446886-74446886 on Build GRCh38
rs970596624 Chr.18: 74446972-74446972 on Build GRCh38
rs748061835 Chr.18: 74446988-74446988 on Build GRCh38
rs375429028 Chr.18: 74446690-74446690 on Build GRCh38
rs757417615 Chr.18: 74446680-74446680 on Build GRCh38
rs537562844 Chr.18: 74446714-74446714 on Build GRCh38
rs376456185 Chr.18: 74446706-74446706 on Build GRCh38
rs536902218 Chr.18: 74446857-74446857 on Build GRCh38
rs568472946 Chr.18: 74446664-74446664 on Build GRCh38
rs900083854 Chr.18: 74446858-74446858 on Build GRCh38
rs968758704 Chr.18: 74446928-74446928 on Build GRCh38
rs1007606537 Chr.18: 74446558-74446558 on Build GRCh38
rs751571074 Chr.18: 74446668-74446668 on Build GRCh38
rs3752101 Chr.18: 74446946-74446946 on Build GRCh38
rs545396559 Chr.18: 74446940-74446940 on Build GRCh38
rs1002792589 Chr.18: 74446849-74446849 on Build GRCh38
rs759199668 Chr.18: 74446579-74446579 on Build GRCh38
rs8082945 Chr.18: 74446763-74446763 on Build GRCh38
rs546458957 Chr.18: 74446559-74446559 on Build GRCh38
rs534438209 Chr.18: 74446802-74446802 on Build GRCh38

View Full Product Details