Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 828151-828591 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AZU1
Entrez Gene ID
566
Gene Name
azurocidin 1
Gene Aliases
AZAMP, AZU, CAP37, HBP, HUMAZUR, NAZC, hHBP
UniGene
Hs.72885
Gene Chromosome Location
Chr.19: 827826-832018 on Build GRCh38
Transcripts
NM_001700.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1680451 c.89G>A p.G30D Chr.19: 828260-828260 on Build GRCh38
6580359 c.174T>C p.H58H Chr.19: 828345-828345 on Build GRCh38
5594248 c.139C>T p.Q47* Chr.19: 828310-828310 on Build GRCh38
6586243 c.59-5C>A p.? Chr.19: 828225-828225 on Build GRCh38
1003497 c.212G>A p.S71N Chr.19: 828383-828383 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs975294900 Chr.19: 828368-828368 on Build GRCh38
rs559592689 Chr.19: 828275-828275 on Build GRCh38
rs746644189 Chr.19: 828288-828288 on Build GRCh38
rs558206275 Chr.19: 828178-828178 on Build GRCh38
rs113510125 Chr.19: 828332-828332 on Build GRCh38
rs377499337 Chr.19: 828371-828371 on Build GRCh38
rs777814627 Chr.19: 828426-828426 on Build GRCh38
rs757611252 Chr.19: 828416-828417 on Build GRCh38
rs781577673 Chr.19: 828272-828272 on Build GRCh38
rs746066408 Chr.19: 828222-828222 on Build GRCh38
rs776408993 Chr.19: 828327-828327 on Build GRCh38
rs191231045 Chr.19: 828171-828171 on Build GRCh38
rs761153158 Chr.19: 828407-828407 on Build GRCh38
rs758446551 Chr.19: 828380-828387 on Build GRCh38
rs184057116 Chr.19: 828258-828258 on Build GRCh38
rs926581387 Chr.19: 828462-828462 on Build GRCh38
rs773012210 Chr.19: 828287-828287 on Build GRCh38
rs1056726698 Chr.19: 828554-828556 on Build GRCh38
rs758299163 Chr.19: 828363-828363 on Build GRCh38
rs767784098 Chr.19: 828347-828347 on Build GRCh38
rs780129133 Chr.19: 828223-828223 on Build GRCh38
rs755091033 Chr.19: 828360-828360 on Build GRCh38
rs1027757053 Chr.19: 828425-828425 on Build GRCh38
rs982639715 Chr.19: 828293-828293 on Build GRCh38
rs144130932 Chr.19: 828263-828263 on Build GRCh38
rs757628322 Chr.19: 828221-828221 on Build GRCh38
rs746878090 Chr.19: 828365-828365 on Build GRCh38
rs762234556 Chr.19: 828396-828396 on Build GRCh38
rs761657439 Chr.19: 828231-828231 on Build GRCh38
rs755041641 Chr.19: 828278-828278 on Build GRCh38
rs754875458 Chr.19: 828344-828357 on Build GRCh38
rs751421360 Chr.19: 828434-828434 on Build GRCh38
rs1028099898 Chr.19: 828274-828274 on Build GRCh38
rs377079085 Chr.19: 828527-828527 on Build GRCh38
rs375253461 Chr.19: 828214-828214 on Build GRCh38
rs370446425 Chr.19: 828318-828318 on Build GRCh38
rs938026132 Chr.19: 828522-828522 on Build GRCh38
rs976534448 Chr.19: 828564-828564 on Build GRCh38
rs765179806 Chr.19: 828374-828391 on Build GRCh38
rs754090107 Chr.19: 828410-828410 on Build GRCh38
rs777718209 Chr.19: 828389-828390 on Build GRCh38
rs753330643 Chr.19: 828262-828262 on Build GRCh38
rs369485730 Chr.19: 828224-828224 on Build GRCh38
rs1042682777 Chr.19: 828188-828188 on Build GRCh38
rs894037669 Chr.19: 828176-828176 on Build GRCh38
rs766699351 Chr.19: 828408-828408 on Build GRCh38
rs759165177 Chr.19: 828328-828328 on Build GRCh38
rs771796857 Chr.19: 828286-828286 on Build GRCh38
rs531710662 Chr.19: 828422-828422 on Build GRCh38
rs753680041 Chr.19: 828216-828217 on Build GRCh38
rs1015455815 Chr.19: 828524-828524 on Build GRCh38
rs770827243 Chr.19: 828366-828366 on Build GRCh38
rs745474843 Chr.19: 828372-828372 on Build GRCh38
rs921017716 Chr.19: 828350-828350 on Build GRCh38
rs769448939 Chr.19: 828376-828376 on Build GRCh38
rs201991986 Chr.19: 828399-828399 on Build GRCh38
rs1055725872 Chr.19: 828414-828414 on Build GRCh38
rs965304064 Chr.19: 828538-828538 on Build GRCh38
rs376406973 Chr.19: 828269-828269 on Build GRCh38
rs976587095 Chr.19: 828342-828342 on Build GRCh38
rs549929895 Chr.19: 828515-828515 on Build GRCh38
rs747231172 Chr.19: 828412-828412 on Build GRCh38
rs112635330 Chr.19: 828246-828246 on Build GRCh38
rs772781020 Chr.19: 828252-828252 on Build GRCh38
rs200732632 Chr.19: 828230-828230 on Build GRCh38
rs757145221 Chr.19: 828437-828437 on Build GRCh38
rs758393187 Chr.19: 828423-828423 on Build GRCh38
rs766144640 Chr.19: 828198-828199 on Build GRCh38
rs561308327 Chr.19: 828420-828421 on Build GRCh38
rs371419465 Chr.19: 828392-828392 on Build GRCh38
rs533210773 Chr.19: 828349-828349 on Build GRCh38
rs755146107 Chr.19: 828415-828415 on Build GRCh38
rs764755144 Chr.19: 828268-828268 on Build GRCh38
rs979542797 Chr.19: 828448-828448 on Build GRCh38
rs754040928 Chr.19: 828355-828355 on Build GRCh38
rs893621873 Chr.19: 828216-828216 on Build GRCh38
rs771879307 Chr.19: 828249-828249 on Build GRCh38
rs1028602180 Chr.19: 828570-828570 on Build GRCh38
rs748088127 Chr.19: 828281-828281 on Build GRCh38
rs531365895 Chr.19: 828218-828218 on Build GRCh38
rs757599211 Chr.19: 828270-828270 on Build GRCh38
rs373430982 Chr.19: 828212-828212 on Build GRCh38
rs758917067 Chr.19: 828179-828179 on Build GRCh38
rs760337322 Chr.19: 828253-828253 on Build GRCh38
rs267605769 Chr.19: 828292-828292 on Build GRCh38
rs546084154 Chr.19: 828173-828173 on Build GRCh38
rs777566662 Chr.19: 828364-828364 on Build GRCh38
rs764805887 Chr.19: 828331-828331 on Build GRCh38
rs778924546 Chr.19: 828362-828362 on Build GRCh38
rs201217134 Chr.19: 828405-828405 on Build GRCh38
rs541188510 Chr.19: 828273-828273 on Build GRCh38
rs36002066 Chr.19: 828237-828237 on Build GRCh38
rs752597585 Chr.19: 828375-828376 on Build GRCh38
rs765315617 Chr.19: 828416-828416 on Build GRCh38

View Full Product Details