Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 830622-831069 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AZU1
Entrez Gene ID
566
Gene Name
azurocidin 1
Gene Aliases
AZAMP, AZU, CAP37, HBP, HUMAZUR, NAZC, hHBP
UniGene
Hs.72885
Gene Chromosome Location
Chr.19: 827826-832018 on Build GRCh38
Transcripts
NM_001700.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6557195 c.426G>T p.T142T Chr.19: 830773-830773 on Build GRCh38
1740482 c.457G>A p.G153S Chr.19: 830804-830804 on Build GRCh38
475502 c.514G>A p.V172M Chr.19: 830861-830861 on Build GRCh38
216482 c.542G>A p.R181H Chr.19: 830889-830889 on Build GRCh38
6609191 c.395C>T p.T132M Chr.19: 830742-830742 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs61732996 Chr.19: 830841-830841 on Build GRCh38
rs755484616 Chr.19: 830809-830809 on Build GRCh38
rs78165561 Chr.19: 830774-830774 on Build GRCh38
rs932998085 Chr.19: 830703-830703 on Build GRCh38
rs140987452 Chr.19: 830742-830742 on Build GRCh38
rs774697507 Chr.19: 831026-831026 on Build GRCh38
rs778716358 Chr.19: 830988-830988 on Build GRCh38
rs909026435 Chr.19: 830662-830662 on Build GRCh38
rs1026075091 Chr.19: 831041-831041 on Build GRCh38
rs749185505 Chr.19: 830924-830924 on Build GRCh38
rs573452523 Chr.19: 830990-830990 on Build GRCh38
rs113354700 Chr.19: 831025-831025 on Build GRCh38
rs543133240 Chr.19: 830951-830951 on Build GRCh38
rs550259328 Chr.19: 831046-831046 on Build GRCh38
rs757429833 Chr.19: 830900-830900 on Build GRCh38
rs772354159 Chr.19: 830758-830758 on Build GRCh38
rs137867622 Chr.19: 830932-830932 on Build GRCh38
rs1051889574 Chr.19: 830715-830715 on Build GRCh38
rs947947978 Chr.19: 830981-830981 on Build GRCh38
rs936054551 Chr.19: 830996-830996 on Build GRCh38
rs747759846 Chr.19: 830927-830927 on Build GRCh38
rs935027036 Chr.19: 830963-830963 on Build GRCh38
rs771744178 Chr.19: 830992-830992 on Build GRCh38
rs770090217 Chr.19: 830745-830750 on Build GRCh38
rs371203310 Chr.19: 830909-830909 on Build GRCh38
rs904809336 Chr.19: 830898-830898 on Build GRCh38
rs776294256 Chr.19: 831035-831035 on Build GRCh38
rs932026533 Chr.19: 830656-830656 on Build GRCh38
rs201821197 Chr.19: 830875-830875 on Build GRCh38
rs745794088 Chr.19: 830839-830839 on Build GRCh38
rs762731829 Chr.19: 830783-830783 on Build GRCh38
rs595844 Chr.19: 830854-830854 on Build GRCh38
rs371000929 Chr.19: 830805-830805 on Build GRCh38
rs368496402 Chr.19: 830819-830819 on Build GRCh38
rs201652896 Chr.19: 830888-830888 on Build GRCh38
rs1027553314 Chr.19: 830960-830960 on Build GRCh38
rs758863448 Chr.19: 830871-830871 on Build GRCh38
rs780004498 Chr.19: 830921-830921 on Build GRCh38
rs753183696 Chr.19: 830813-830813 on Build GRCh38
rs751200199 Chr.19: 830798-830798 on Build GRCh38
rs145363133 Chr.19: 830861-830861 on Build GRCh38
rs756163889 Chr.19: 830912-830912 on Build GRCh38
rs28626600 Chr.19: 830820-830820 on Build GRCh38
rs759108068 Chr.19: 831000-831000 on Build GRCh38
rs151205721 Chr.19: 830724-830724 on Build GRCh38
rs141563771 Chr.19: 830773-830773 on Build GRCh38
rs753236836 Chr.19: 830867-830867 on Build GRCh38
rs1053830763 Chr.19: 830966-830966 on Build GRCh38
rs776160226 Chr.19: 830943-830943 on Build GRCh38
rs750611158 Chr.19: 830967-830967 on Build GRCh38
rs1014964088 Chr.19: 830941-830941 on Build GRCh38
rs752023955 Chr.19: 830955-830955 on Build GRCh38
rs765959714 Chr.19: 830936-830936 on Build GRCh38
rs376967352 Chr.19: 830857-830857 on Build GRCh38
rs1054695185 Chr.19: 831015-831015 on Build GRCh38
rs138374662 Chr.19: 830925-830925 on Build GRCh38
rs141614223 Chr.19: 830860-830860 on Build GRCh38
rs376157351 Chr.19: 830659-830659 on Build GRCh38
rs781271870 Chr.19: 830838-830838 on Build GRCh38
rs771010406 Chr.19: 830769-830769 on Build GRCh38
rs771828264 Chr.19: 830930-830930 on Build GRCh38
rs369142349 Chr.19: 830666-830666 on Build GRCh38
rs906250875 Chr.19: 830918-830918 on Build GRCh38
rs764375369 Chr.19: 830910-830910 on Build GRCh38
rs749238516 Chr.19: 830979-830979 on Build GRCh38
rs368212768 Chr.19: 830743-830743 on Build GRCh38
rs372031493 Chr.19: 830753-830753 on Build GRCh38
rs570804979 Chr.19: 830840-830840 on Build GRCh38
rs776619088 Chr.19: 830678-830678 on Build GRCh38
rs941870770 Chr.19: 830663-830663 on Build GRCh38
rs1053310212 Chr.19: 830907-830907 on Build GRCh38
rs142354223 Chr.19: 830942-830942 on Build GRCh38
rs149180756 Chr.19: 830832-830832 on Build GRCh38
rs767015057 Chr.19: 830803-830803 on Build GRCh38
rs760294050 Chr.19: 830961-830976 on Build GRCh38
rs772730104 Chr.19: 830931-830931 on Build GRCh38
rs61732997 Chr.19: 830829-830829 on Build GRCh38
rs977541952 Chr.19: 830982-830982 on Build GRCh38
rs915165097 Chr.19: 830971-830971 on Build GRCh38
rs771548104 Chr.19: 830807-830808 on Build GRCh38
rs376150747 Chr.19: 830738-830738 on Build GRCh38
rs761271952 Chr.19: 830800-830800 on Build GRCh38
rs757548506 Chr.19: 830959-830959 on Build GRCh38
rs763493733 Chr.19: 830945-830945 on Build GRCh38
rs372443381 Chr.19: 830908-830908 on Build GRCh38
rs772652509 Chr.19: 830824-830825 on Build GRCh38
rs773907749 Chr.19: 830855-830855 on Build GRCh38
rs754324176 Chr.19: 830804-830804 on Build GRCh38
rs774067158 Chr.19: 830889-830889 on Build GRCh38
rs920446246 Chr.19: 830644-830644 on Build GRCh38
rs774138133 Chr.19: 830926-830926 on Build GRCh38
rs540843317 Chr.19: 830993-830993 on Build GRCh38
rs776211421 Chr.19: 830994-830994 on Build GRCh38
rs766058397 Chr.19: 831034-831035 on Build GRCh38
rs766294858 Chr.19: 830677-830677 on Build GRCh38
rs373338227 Chr.19: 830700-830700 on Build GRCh38
rs919044111 Chr.19: 830690-830690 on Build GRCh38
rs924600461 Chr.19: 830983-830983 on Build GRCh38
rs556744148 Chr.19: 830859-830859 on Build GRCh38
rs749776715 Chr.19: 830783-830783 on Build GRCh38
rs1014603397 Chr.19: 830794-830794 on Build GRCh38
rs759287867 Chr.19: 830681-830681 on Build GRCh38
rs147872987 Chr.19: 830772-830772 on Build GRCh38
rs376674305 Chr.19: 830767-830767 on Build GRCh38
rs568736368 Chr.19: 830952-830952 on Build GRCh38
rs752367946 Chr.19: 830714-830714 on Build GRCh38
rs985899614 Chr.19: 830977-830977 on Build GRCh38
rs140313087 Chr.19: 830980-830980 on Build GRCh38
rs756215262 Chr.19: 830970-830970 on Build GRCh38
rs28664760 Chr.19: 830768-830768 on Build GRCh38
rs773367600 Chr.19: 830660-830660 on Build GRCh38
rs1041962705 Chr.19: 830683-830683 on Build GRCh38
rs547297811 Chr.19: 830735-830735 on Build GRCh38
rs763793172 Chr.19: 830787-830787 on Build GRCh38
rs779429626 Chr.19: 830810-830810 on Build GRCh38
rs768217830 Chr.19: 830691-830691 on Build GRCh38
rs776019769 Chr.19: 830766-830785 on Build GRCh38
rs112932668 Chr.19: 831040-831040 on Build GRCh38
rs773136690 Chr.19: 830781-830781 on Build GRCh38
rs375688702 Chr.19: 830978-830978 on Build GRCh38
rs762276684 Chr.19: 831027-831027 on Build GRCh38
rs762784855 Chr.19: 830845-830845 on Build GRCh38
rs756775340 Chr.19: 830737-830737 on Build GRCh38

View Full Product Details