Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 1230794-1231313 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CBARP
Entrez Gene ID
255057
Gene Name
CACN beta subunit associated regulatory protein
Gene Aliases
BARP, C19orf26, DOS
UniGene
Hs.346575
Gene Chromosome Location
Chr.19: 1228287-1238512 on Build GRCh38
Transcripts
NM_152769.2 XM_017026555.1 XM_017026556.1 XM_017026557.1 XM_017026558.1 XM_017026559.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6202248 c.989G>T p.R330L Chr.19: 1231248-1231248 on Build GRCh38
2815553 c.1313C>G p.T438S Chr.19: 1230924-1230924 on Build GRCh38
7301060 c.1220delC p.P407fs*>47 Chr.19: 1231035-1231035 on Build GRCh38
4667137 c.1202delC p.P401fs*>47 Chr.19: 1231035-1231035 on Build GRCh38
7414090 c.1252G>A p.A418T Chr.19: 1230985-1230985 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766926207 Chr.19: 1231042-1231042 on Build GRCh38
rs549881842 Chr.19: 1231037-1231037 on Build GRCh38
rs749222203 Chr.19: 1230859-1230859 on Build GRCh38
rs753413344 Chr.19: 1231044-1231044 on Build GRCh38
rs777411929 Chr.19: 1231017-1231017 on Build GRCh38
rs771267092 Chr.19: 1231135-1231135 on Build GRCh38
rs774741354 Chr.19: 1231137-1231137 on Build GRCh38
rs146809339 Chr.19: 1231263-1231263 on Build GRCh38
rs762688472 Chr.19: 1231217-1231217 on Build GRCh38
rs772696313 Chr.19: 1231139-1231139 on Build GRCh38
rs778821034 Chr.19: 1231035-1231035 on Build GRCh38
rs144889893 Chr.19: 1231223-1231223 on Build GRCh38
rs373982099 Chr.19: 1230995-1230995 on Build GRCh38
rs772263901 Chr.19: 1230963-1230963 on Build GRCh38
rs758588663 Chr.19: 1231294-1231294 on Build GRCh38
rs763362020 Chr.19: 1231041-1231041 on Build GRCh38
rs776627990 Chr.19: 1231285-1231285 on Build GRCh38
rs769048353 Chr.19: 1230975-1230975 on Build GRCh38
rs529073514 Chr.19: 1231052-1231052 on Build GRCh38
rs112943778 Chr.19: 1230849-1230849 on Build GRCh38
rs747873410 Chr.19: 1230854-1230854 on Build GRCh38
rs140733006 Chr.19: 1231209-1231209 on Build GRCh38
rs370323804 Chr.19: 1230976-1230976 on Build GRCh38
rs1010266157 Chr.19: 1230820-1230820 on Build GRCh38
rs779237736 Chr.19: 1231172-1231172 on Build GRCh38
rs554460406 Chr.19: 1231242-1231242 on Build GRCh38
rs749663010 Chr.19: 1231072-1231072 on Build GRCh38
rs749556660 Chr.19: 1230867-1230869 on Build GRCh38
rs916118639 Chr.19: 1231286-1231286 on Build GRCh38
rs779114359 Chr.19: 1231249-1231249 on Build GRCh38
rs747561926 Chr.19: 1231192-1231192 on Build GRCh38
rs202062486 Chr.19: 1231083-1231083 on Build GRCh38
rs757027814 Chr.19: 1231029-1231029 on Build GRCh38
rs750635988 Chr.19: 1231293-1231293 on Build GRCh38
rs769469325 Chr.19: 1231034-1231035 on Build GRCh38
rs777769050 Chr.19: 1231167-1231167 on Build GRCh38
rs368744096 Chr.19: 1230989-1230989 on Build GRCh38
rs1001634165 Chr.19: 1230833-1230833 on Build GRCh38
rs766979616 Chr.19: 1230993-1230993 on Build GRCh38
rs752648961 Chr.19: 1231163-1231163 on Build GRCh38
rs752661766 Chr.19: 1231232-1231232 on Build GRCh38
rs779202931 Chr.19: 1230934-1230935 on Build GRCh38
rs751040900 Chr.19: 1230897-1230897 on Build GRCh38
rs915150202 Chr.19: 1231176-1231176 on Build GRCh38
rs75463186 Chr.19: 1231043-1231043 on Build GRCh38
rs750118611 Chr.19: 1231105-1231105 on Build GRCh38
rs756229000 Chr.19: 1231126-1231126 on Build GRCh38
rs755004029 Chr.19: 1231116-1231116 on Build GRCh38
rs1003819494 Chr.19: 1231159-1231159 on Build GRCh38
rs534213272 Chr.19: 1231118-1231118 on Build GRCh38
rs777517023 Chr.19: 1230918-1230918 on Build GRCh38
rs775865702 Chr.19: 1230869-1230869 on Build GRCh38
rs748578658 Chr.19: 1230960-1230960 on Build GRCh38
rs765709603 Chr.19: 1230981-1230981 on Build GRCh38
rs764939260 Chr.19: 1231048-1231048 on Build GRCh38
rs764157694 Chr.19: 1231241-1231241 on Build GRCh38
rs8110590 Chr.19: 1231143-1231143 on Build GRCh38
rs373552551 Chr.19: 1230900-1230900 on Build GRCh38
rs372008248 Chr.19: 1231208-1231208 on Build GRCh38
rs770117663 Chr.19: 1231070-1231070 on Build GRCh38
rs769591833 Chr.19: 1230855-1230855 on Build GRCh38
rs761359927 Chr.19: 1231091-1231091 on Build GRCh38
rs538068170 Chr.19: 1230815-1230815 on Build GRCh38
rs771515759 Chr.19: 1231034-1231034 on Build GRCh38
rs777136090 Chr.19: 1230969-1230969 on Build GRCh38
rs745899331 Chr.19: 1230957-1230957 on Build GRCh38
rs946312034 Chr.19: 1231032-1231032 on Build GRCh38
rs747045240 Chr.19: 1231055-1231055 on Build GRCh38
rs368745333 Chr.19: 1231166-1231166 on Build GRCh38
rs755130689 Chr.19: 1231059-1231059 on Build GRCh38
rs756711336 Chr.19: 1230914-1230914 on Build GRCh38
rs774435933 Chr.19: 1230861-1230861 on Build GRCh38
rs1024874901 Chr.19: 1230856-1230856 on Build GRCh38
rs760947065 Chr.19: 1230970-1230970 on Build GRCh38
rs772395227 Chr.19: 1230985-1230985 on Build GRCh38
rs370841902 Chr.19: 1230884-1230884 on Build GRCh38
rs974539910 Chr.19: 1230928-1230928 on Build GRCh38
rs778056754 Chr.19: 1231134-1231134 on Build GRCh38
rs745357441 Chr.19: 1231035-1231035 on Build GRCh38
rs200100448 Chr.19: 1230987-1230987 on Build GRCh38
rs780254035 Chr.19: 1230858-1230859 on Build GRCh38
rs761912877 Chr.19: 1231289-1231289 on Build GRCh38
rs933506268 Chr.19: 1230998-1230998 on Build GRCh38
rs770638939 Chr.19: 1231275-1231275 on Build GRCh38
rs764956513 Chr.19: 1231099-1231099 on Build GRCh38
rs143744896 Chr.19: 1231180-1231180 on Build GRCh38
rs770857450 Chr.19: 1230947-1230947 on Build GRCh38
rs764548299 Chr.19: 1230883-1230883 on Build GRCh38
rs1034807872 Chr.19: 1230844-1230844 on Build GRCh38
rs1013394493 Chr.19: 1231069-1231069 on Build GRCh38
rs764757468 Chr.19: 1231145-1231145 on Build GRCh38
rs535630461 Chr.19: 1231189-1231189 on Build GRCh38
rs780642066 Chr.19: 1231185-1231185 on Build GRCh38
rs746371959 Chr.19: 1231138-1231138 on Build GRCh38
rs367849189 Chr.19: 1231004-1231004 on Build GRCh38
rs542551827 Chr.19: 1230826-1230826 on Build GRCh38
rs762186919 Chr.19: 1231038-1231038 on Build GRCh38
rs757545091 Chr.19: 1231171-1231171 on Build GRCh38
rs762116218 Chr.19: 1230977-1230977 on Build GRCh38
rs750747430 Chr.19: 1231250-1231250 on Build GRCh38
rs574917073 Chr.19: 1230821-1230821 on Build GRCh38
rs199653057 Chr.19: 1231080-1231080 on Build GRCh38
rs894614553 Chr.19: 1231111-1231111 on Build GRCh38
rs780767215 Chr.19: 1230907-1230907 on Build GRCh38
rs781379578 Chr.19: 1231066-1231066 on Build GRCh38
rs201218608 Chr.19: 1231036-1231036 on Build GRCh38
rs773662961 Chr.19: 1231040-1231040 on Build GRCh38
rs757759533 Chr.19: 1230892-1230892 on Build GRCh38
rs775259962 Chr.19: 1231113-1231146 on Build GRCh38
rs746549975 Chr.19: 1230846-1230846 on Build GRCh38
rs1028163269 Chr.19: 1231170-1231170 on Build GRCh38
rs766368928 Chr.19: 1231113-1231113 on Build GRCh38
rs757414200 Chr.19: 1231243-1231243 on Build GRCh38
rs751518232 Chr.19: 1231115-1231115 on Build GRCh38
rs762743530 Chr.19: 1231151-1231151 on Build GRCh38
rs760650145 Chr.19: 1231284-1231284 on Build GRCh38
rs755315849 Chr.19: 1230913-1230914 on Build GRCh38
rs749089918 Chr.19: 1230942-1230942 on Build GRCh38
rs759376967 Chr.19: 1231224-1231224 on Build GRCh38
rs867091868 Chr.19: 1230934-1230934 on Build GRCh38
rs758813156 Chr.19: 1231259-1231259 on Build GRCh38
rs1004808615 Chr.19: 1231085-1231085 on Build GRCh38
rs202174072 Chr.19: 1230875-1230875 on Build GRCh38
rs753633360 Chr.19: 1231018-1231018 on Build GRCh38
rs201160402 Chr.19: 1231162-1231162 on Build GRCh38
rs1005097755 Chr.19: 1231274-1231274 on Build GRCh38
rs894750835 Chr.19: 1230824-1230824 on Build GRCh38
rs759561839 Chr.19: 1230863-1230863 on Build GRCh38
rs377468857 Chr.19: 1230857-1230857 on Build GRCh38
rs748285437 Chr.19: 1231068-1231068 on Build GRCh38
rs756271515 Chr.19: 1230854-1230868 on Build GRCh38
rs560621209 Chr.19: 1230837-1230837 on Build GRCh38
rs556364899 Chr.19: 1230817-1230817 on Build GRCh38
rs1048624784 Chr.19: 1231144-1231144 on Build GRCh38
rs766244135 Chr.19: 1231156-1231156 on Build GRCh38
rs982499202 Chr.19: 1230864-1230864 on Build GRCh38
rs760087675 Chr.19: 1231081-1231081 on Build GRCh38
rs767313930 Chr.19: 1231231-1231231 on Build GRCh38
rs765395408 Chr.19: 1231290-1231290 on Build GRCh38
rs761033508 Chr.19: 1230880-1230880 on Build GRCh38
rs759375801 Chr.19: 1231160-1231160 on Build GRCh38
rs1043332238 Chr.19: 1231194-1231194 on Build GRCh38
rs759199749 Chr.19: 1231280-1231280 on Build GRCh38
rs374648311 Chr.19: 1231157-1231157 on Build GRCh38
rs773433082 Chr.19: 1231071-1231071 on Build GRCh38
rs758353354 Chr.19: 1231050-1231050 on Build GRCh38
rs748817445 Chr.19: 1231270-1231270 on Build GRCh38
rs777406939 Chr.19: 1231268-1231268 on Build GRCh38
rs756977356 Chr.19: 1231047-1231047 on Build GRCh38
rs769107400 Chr.19: 1231264-1231264 on Build GRCh38
rs758299866 Chr.19: 1231106-1231106 on Build GRCh38
rs762645325 Chr.19: 1231210-1231212 on Build GRCh38
rs765685504 Chr.19: 1230896-1230896 on Build GRCh38
rs771357082 Chr.19: 1231074-1231074 on Build GRCh38
rs746245251 Chr.19: 1231174-1231174 on Build GRCh38
rs545836096 Chr.19: 1231283-1231283 on Build GRCh38
rs552122460 Chr.19: 1231119-1231119 on Build GRCh38
rs754032618 Chr.19: 1231169-1231169 on Build GRCh38
rs752695821 Chr.19: 1230830-1230830 on Build GRCh38
rs774174156 Chr.19: 1231279-1231279 on Build GRCh38
rs780330992 Chr.19: 1231262-1231262 on Build GRCh38
rs1026642384 Chr.19: 1230827-1230827 on Build GRCh38
rs778716668 Chr.19: 1230955-1230955 on Build GRCh38
rs895942094 Chr.19: 1231248-1231248 on Build GRCh38
rs773021394 Chr.19: 1231147-1231147 on Build GRCh38
rs950616299 Chr.19: 1230859-1230859 on Build GRCh38
rs200691204 Chr.19: 1230865-1230865 on Build GRCh38
rs760573404 Chr.19: 1231240-1231240 on Build GRCh38
rs375974361 Chr.19: 1230910-1230910 on Build GRCh38

View Full Product Details